Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2881777-2882291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD1
Entrez Gene ID
118
Gene Name
adducin 1
Gene Aliases
ADDA
UniGene
Hs.183706
Gene Chromosome Location
Chr.4: 2843732-2930076 on Build GRCh38
Transcripts
NM_001119.4 NM_001286645.1 NM_014189.3 NM_014190.3 NM_176801.2 XM_005247933.2 XM_005247934.2 XM_005247935.2 XM_005247936.2 XM_005247937.2 XM_017007703.1 XM_017007704.1 XM_017007705.1 XM_017007706.1 XM_017007707.1 XM_017007708.1 XM_017007709.1 XM_017007710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3134005 c.329C>T p.P110L Chr.4: 2882031-2882031 on Build GRCh38
6595106 c.228G>A p.Q76Q Chr.4: 2881930-2881930 on Build GRCh38
3134004 c.328C>T p.P110S Chr.4: 2882030-2882030 on Build GRCh38
4332107 c.208G>T p.E70* Chr.4: 2881910-2881910 on Build GRCh38
3993637 c.326C>A p.A109D Chr.4: 2882028-2882028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200543663 Chr.4: 2881865-2881865 on Build GRCh38
rs753061853 Chr.4: 2881887-2881887 on Build GRCh38
rs780023838 Chr.4: 2881964-2881964 on Build GRCh38
rs764396737 Chr.4: 2882027-2882027 on Build GRCh38
rs970147300 Chr.4: 2882045-2882045 on Build GRCh38
rs376198993 Chr.4: 2882011-2882011 on Build GRCh38
rs910860920 Chr.4: 2882143-2882143 on Build GRCh38
rs770419678 Chr.4: 2882108-2882108 on Build GRCh38
rs754093766 Chr.4: 2882031-2882031 on Build GRCh38
rs758495974 Chr.4: 2882050-2882050 on Build GRCh38
rs192299459 Chr.4: 2882138-2882138 on Build GRCh38
rs760130500 Chr.4: 2881820-2881820 on Build GRCh38
rs1039625971 Chr.4: 2881847-2881847 on Build GRCh38
rs776416895 Chr.4: 2881853-2881853 on Build GRCh38
rs757352719 Chr.4: 2881903-2881903 on Build GRCh38
rs147906331 Chr.4: 2881886-2881886 on Build GRCh38
rs113259674 Chr.4: 2881952-2881952 on Build GRCh38
rs965404808 Chr.4: 2882142-2882142 on Build GRCh38
rs1007387312 Chr.4: 2882131-2882131 on Build GRCh38
rs985447920 Chr.4: 2881811-2881811 on Build GRCh38
rs923473763 Chr.4: 2882204-2882204 on Build GRCh38
rs200647918 Chr.4: 2881920-2881920 on Build GRCh38
rs757515922 Chr.4: 2882032-2882032 on Build GRCh38
rs1025924620 Chr.4: 2882158-2882158 on Build GRCh38
rs942442859 Chr.4: 2881815-2881815 on Build GRCh38
rs952407485 Chr.4: 2882124-2882124 on Build GRCh38
rs374828271 Chr.4: 2881991-2881991 on Build GRCh38
rs780475507 Chr.4: 2882200-2882200 on Build GRCh38
rs529207799 Chr.4: 2882235-2882235 on Build GRCh38
rs746863295 Chr.4: 2881966-2881966 on Build GRCh38
rs1080566 Chr.4: 2881818-2881818 on Build GRCh38
rs372131271 Chr.4: 2882099-2882099 on Build GRCh38
rs911229589 Chr.4: 2881822-2881822 on Build GRCh38
rs115217936 Chr.4: 2882159-2882159 on Build GRCh38
rs570518249 Chr.4: 2881997-2881999 on Build GRCh38
rs747752144 Chr.4: 2881985-2881985 on Build GRCh38
rs1053363015 Chr.4: 2881894-2881894 on Build GRCh38
rs776433418 Chr.4: 2881984-2881984 on Build GRCh38
rs901976736 Chr.4: 2881850-2881850 on Build GRCh38
rs779858510 Chr.4: 2882071-2882071 on Build GRCh38
rs769374673 Chr.4: 2881997-2881997 on Build GRCh38
rs910059957 Chr.4: 2882196-2882196 on Build GRCh38
rs79280082 Chr.4: 2882105-2882105 on Build GRCh38
rs762511882 Chr.4: 2882002-2882002 on Build GRCh38
rs113362280 Chr.4: 2881995-2881995 on Build GRCh38
rs1021888613 Chr.4: 2882121-2882121 on Build GRCh38
rs772637954 Chr.4: 2881998-2881998 on Build GRCh38
rs752128069 Chr.4: 2881857-2881857 on Build GRCh38
rs891836805 Chr.4: 2881958-2881958 on Build GRCh38
rs764781475 Chr.4: 2881855-2881855 on Build GRCh38
rs965616983 Chr.4: 2881840-2881840 on Build GRCh38
rs777977683 Chr.4: 2881899-2881899 on Build GRCh38
rs745933841 Chr.4: 2881908-2881908 on Build GRCh38
rs768496845 Chr.4: 2881972-2881972 on Build GRCh38
rs748473867 Chr.4: 2881842-2881842 on Build GRCh38
rs750522808 Chr.4: 2882049-2882049 on Build GRCh38
rs754003824 Chr.4: 2881900-2881900 on Build GRCh38
rs984277886 Chr.4: 2882184-2882184 on Build GRCh38
rs755065282 Chr.4: 2882237-2882237 on Build GRCh38
rs756466627 Chr.4: 2881893-2881893 on Build GRCh38
rs767924054 Chr.4: 2882015-2882015 on Build GRCh38
rs772914237 Chr.4: 2882103-2882103 on Build GRCh38
rs767976701 Chr.4: 2882004-2882004 on Build GRCh38
rs778820874 Chr.4: 2881905-2881905 on Build GRCh38
rs371281109 Chr.4: 2881928-2881928 on Build GRCh38
rs573310707 Chr.4: 2882085-2882085 on Build GRCh38
rs747919792 Chr.4: 2882091-2882091 on Build GRCh38
rs16843535 Chr.4: 2882224-2882224 on Build GRCh38
rs926973594 Chr.4: 2882229-2882229 on Build GRCh38
rs774948008 Chr.4: 2881809-2881809 on Build GRCh38
rs750824150 Chr.4: 2882155-2882155 on Build GRCh38
rs918494052 Chr.4: 2882113-2882113 on Build GRCh38
rs746881980 Chr.4: 2882073-2882073 on Build GRCh38
rs754305003 Chr.4: 2881957-2881957 on Build GRCh38
rs553553817 Chr.4: 2882046-2882046 on Build GRCh38
rs768035833 Chr.4: 2881874-2881874 on Build GRCh38
rs781092667 Chr.4: 2882086-2882086 on Build GRCh38
rs761042261 Chr.4: 2882012-2882012 on Build GRCh38

View Full Product Details