Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94127631-94128210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH17
Entrez Gene ID
1015
Gene Name
cadherin 17
Gene Aliases
CDH16, HPT-1, HPT1
UniGene
Hs.591853
Gene Chromosome Location
Chr.8: 94127162-94217303 on Build GRCh38
Transcripts
NM_001144663.1 NM_004063.3 XM_011516790.1

SNP Detail

SNP ID Chromosome Location
rs981319278 Chr.8: 94127837-94127837 on Build GRCh38
rs1052466162 Chr.8: 94127900-94127900 on Build GRCh38
rs548046229 Chr.8: 94127791-94127791 on Build GRCh38
rs532733284 Chr.8: 94127722-94127722 on Build GRCh38
rs377610823 Chr.8: 94127720-94127720 on Build GRCh38
rs1004268186 Chr.8: 94127970-94127970 on Build GRCh38
rs1019242241 Chr.8: 94127696-94127696 on Build GRCh38
rs183321980 Chr.8: 94128133-94128133 on Build GRCh38
rs777868197 Chr.8: 94127913-94127913 on Build GRCh38
rs534499964 Chr.8: 94127867-94127867 on Build GRCh38
rs562446436 Chr.8: 94128160-94128160 on Build GRCh38
rs769808340 Chr.8: 94127836-94127836 on Build GRCh38
rs1041696406 Chr.8: 94127922-94127922 on Build GRCh38
rs915708698 Chr.8: 94127833-94127833 on Build GRCh38
rs535963510 Chr.8: 94127927-94127927 on Build GRCh38
rs542056751 Chr.8: 94128159-94128159 on Build GRCh38
rs944268312 Chr.8: 94127921-94127921 on Build GRCh38
rs902110807 Chr.8: 94127967-94127967 on Build GRCh38
rs925677848 Chr.8: 94127847-94127847 on Build GRCh38
rs375126839 Chr.8: 94128032-94128032 on Build GRCh38
rs16916576 Chr.8: 94128176-94128176 on Build GRCh38
rs1040558738 Chr.8: 94128166-94128166 on Build GRCh38
rs773636619 Chr.8: 94128036-94128037 on Build GRCh38
rs959956634 Chr.8: 94127809-94127809 on Build GRCh38
rs993163415 Chr.8: 94127831-94127831 on Build GRCh38
rs573412175 Chr.8: 94128149-94128149 on Build GRCh38
rs889309559 Chr.8: 94127668-94127668 on Build GRCh38
rs1038726928 Chr.8: 94128165-94128165 on Build GRCh38
rs915727607 Chr.8: 94127832-94127832 on Build GRCh38
rs1015567262 Chr.8: 94127803-94127803 on Build GRCh38
rs551831196 Chr.8: 94127779-94127779 on Build GRCh38
rs996957882 Chr.8: 94127938-94127938 on Build GRCh38
rs1027134017 Chr.8: 94127948-94127948 on Build GRCh38
rs577261443 Chr.8: 94128072-94128072 on Build GRCh38
rs966064521 Chr.8: 94127697-94127697 on Build GRCh38
rs890130088 Chr.8: 94127919-94127919 on Build GRCh38
rs948560898 Chr.8: 94127834-94127834 on Build GRCh38
rs971758619 Chr.8: 94127705-94127705 on Build GRCh38
rs889277033 Chr.8: 94127954-94127954 on Build GRCh38
rs565557660 Chr.8: 94127863-94127863 on Build GRCh38
rs866865696 Chr.8: 94127935-94127935 on Build GRCh38
rs531122199 Chr.8: 94128161-94128161 on Build GRCh38
rs537492031 Chr.8: 94128055-94128055 on Build GRCh38
rs569270514 Chr.8: 94127716-94127716 on Build GRCh38
rs186673641 Chr.8: 94128175-94128175 on Build GRCh38
rs781177018 Chr.8: 94127778-94127778 on Build GRCh38
rs992884439 Chr.8: 94128024-94128024 on Build GRCh38
rs774040043 Chr.8: 94127960-94127960 on Build GRCh38
rs934369372 Chr.8: 94128082-94128082 on Build GRCh38
rs955968646 Chr.8: 94128089-94128089 on Build GRCh38
rs989156358 Chr.8: 94128104-94128104 on Build GRCh38
rs888508770 Chr.8: 94127957-94127957 on Build GRCh38
rs747989741 Chr.8: 94127802-94127802 on Build GRCh38
rs546341553 Chr.8: 94128084-94128084 on Build GRCh38
rs1019575002 Chr.8: 94127958-94127958 on Build GRCh38
rs371037813 Chr.8: 94128025-94128025 on Build GRCh38
rs989212202 Chr.8: 94127760-94127772 on Build GRCh38
rs191319530 Chr.8: 94127701-94127701 on Build GRCh38
rs939311093 Chr.8: 94127880-94127880 on Build GRCh38
rs928521093 Chr.8: 94128074-94128074 on Build GRCh38
rs554873498 Chr.8: 94128005-94128005 on Build GRCh38
rs574856739 Chr.8: 94127991-94128022 on Build GRCh38
rs1032948485 Chr.8: 94127761-94127761 on Build GRCh38
rs556726251 Chr.8: 94127761-94127762 on Build GRCh38
rs1026458729 Chr.8: 94127748-94127748 on Build GRCh38
rs1007809661 Chr.8: 94127689-94127689 on Build GRCh38
rs1015562724 Chr.8: 94127991-94127991 on Build GRCh38
rs748766994 Chr.8: 94127914-94127914 on Build GRCh38
rs557399856 Chr.8: 94128071-94128071 on Build GRCh38

View Full Product Details