Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149751902-149752440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD2
Entrez Gene ID
51122
Gene Name
COMM domain containing 2
Gene Aliases
HSPC042
UniGene
Hs.432729
Gene Chromosome Location
Chr.3: 149737160-149752507 on Build GRCh38
Transcripts
NM_016094.3 NR_135149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6511649 c.83G>T p.G28V Chr.3: 149752272-149752272 on Build GRCh38
1617046 c.73G>A p.A25T Chr.3: 149752282-149752282 on Build GRCh38
5580691 c.45C>T p.F15F Chr.3: 149752400-149752400 on Build GRCh38
4584030 c.24G>A p.E8E Chr.3: 149752421-149752421 on Build GRCh38
445789 c.100_102delTTC p.F34delF Chr.3: 149752253-149752255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781465324 Chr.3: 149752301-149752301 on Build GRCh38
rs761278149 Chr.3: 149752350-149752350 on Build GRCh38
rs528522353 Chr.3: 149751982-149751982 on Build GRCh38
rs998237873 Chr.3: 149752067-149752067 on Build GRCh38
rs113267620 Chr.3: 149752198-149752198 on Build GRCh38
rs913495728 Chr.3: 149751940-149751940 on Build GRCh38
rs979878543 Chr.3: 149752065-149752066 on Build GRCh38
rs927084604 Chr.3: 149752076-149752076 on Build GRCh38
rs781184439 Chr.3: 149752213-149752213 on Build GRCh38
rs1043104975 Chr.3: 149751934-149751934 on Build GRCh38
rs775311488 Chr.3: 149752253-149752253 on Build GRCh38
rs1015897298 Chr.3: 149752336-149752336 on Build GRCh38
rs751625679 Chr.3: 149752184-149752185 on Build GRCh38
rs776855706 Chr.3: 149752037-149752037 on Build GRCh38
rs946038314 Chr.3: 149752145-149752145 on Build GRCh38
rs760274591 Chr.3: 149752395-149752395 on Build GRCh38
rs1039661836 Chr.3: 149752316-149752316 on Build GRCh38
rs1005507864 Chr.3: 149752025-149752025 on Build GRCh38
rs572175781 Chr.3: 149752291-149752291 on Build GRCh38
rs140884434 Chr.3: 149752080-149752080 on Build GRCh38
rs761381804 Chr.3: 149752262-149752262 on Build GRCh38
rs773768894 Chr.3: 149752216-149752216 on Build GRCh38
rs766124022 Chr.3: 149752295-149752295 on Build GRCh38
rs753815576 Chr.3: 149752177-149752177 on Build GRCh38
rs754408116 Chr.3: 149752391-149752391 on Build GRCh38
rs746274570 Chr.3: 149752399-149752399 on Build GRCh38
rs756191730 Chr.3: 149752306-149752306 on Build GRCh38
rs367995657 Chr.3: 149752162-149752162 on Build GRCh38
rs369408183 Chr.3: 149752393-149752393 on Build GRCh38
rs751416299 Chr.3: 149752367-149752367 on Build GRCh38
rs776061197 Chr.3: 149752260-149752260 on Build GRCh38
rs1046893730 Chr.3: 149751997-149751997 on Build GRCh38
rs778160708 Chr.3: 149752311-149752311 on Build GRCh38
rs1047329372 Chr.3: 149752299-149752299 on Build GRCh38
rs888312061 Chr.3: 149752252-149752252 on Build GRCh38
rs750353015 Chr.3: 149752275-149752275 on Build GRCh38
rs201913747 Chr.3: 149752210-149752210 on Build GRCh38
rs759350681 Chr.3: 149752051-149752051 on Build GRCh38
rs777839249 Chr.3: 149752382-149752382 on Build GRCh38
rs956625722 Chr.3: 149752121-149752121 on Build GRCh38
rs762811636 Chr.3: 149752360-149752360 on Build GRCh38
rs899720076 Chr.3: 149752325-149752325 on Build GRCh38
rs766175798 Chr.3: 149752361-149752361 on Build GRCh38
rs747797574 Chr.3: 149752409-149752409 on Build GRCh38
rs747186120 Chr.3: 149752206-149752206 on Build GRCh38
rs759573474 Chr.3: 149752134-149752134 on Build GRCh38
rs752790023 Chr.3: 149752303-149752303 on Build GRCh38
rs780305111 Chr.3: 149752205-149752205 on Build GRCh38
rs963772081 Chr.3: 149752050-149752050 on Build GRCh38
rs904644853 Chr.3: 149751942-149751942 on Build GRCh38
rs768488736 Chr.3: 149752344-149752344 on Build GRCh38
rs769298818 Chr.3: 149752359-149752359 on Build GRCh38
rs556451921 Chr.3: 149752180-149752180 on Build GRCh38
rs763709780 Chr.3: 149752254-149752254 on Build GRCh38
rs759325062 Chr.3: 149752368-149752368 on Build GRCh38
rs779556816 Chr.3: 149752397-149752397 on Build GRCh38
rs373377514 Chr.3: 149752396-149752396 on Build GRCh38
rs774130363 Chr.3: 149752100-149752100 on Build GRCh38
rs776314530 Chr.3: 149752345-149752345 on Build GRCh38
rs201007726 Chr.3: 149752184-149752184 on Build GRCh38
rs867344006 Chr.3: 149752270-149752270 on Build GRCh38
rs751786660 Chr.3: 149752296-149752296 on Build GRCh38
rs767675952 Chr.3: 149752369-149752369 on Build GRCh38
rs375191073 Chr.3: 149752268-149752268 on Build GRCh38
rs535681632 Chr.3: 149752200-149752200 on Build GRCh38
rs557801773 Chr.3: 149752375-149752375 on Build GRCh38
rs770264317 Chr.3: 149752215-149752215 on Build GRCh38
rs771423951 Chr.3: 149752326-149752326 on Build GRCh38
rs992649966 Chr.3: 149752404-149752404 on Build GRCh38
rs993129073 Chr.3: 149752114-149752114 on Build GRCh38
rs905907486 Chr.3: 149751969-149751969 on Build GRCh38
rs146422588 Chr.3: 149751964-149751964 on Build GRCh38
rs150156056 Chr.3: 149752331-149752331 on Build GRCh38
rs199947632 Chr.3: 149752169-149752169 on Build GRCh38
rs775070099 Chr.3: 149752324-149752324 on Build GRCh38
rs768046794 Chr.3: 149752339-149752340 on Build GRCh38
rs766927842 Chr.3: 149752141-149752141 on Build GRCh38
rs771368903 Chr.3: 149752239-149752239 on Build GRCh38
rs200332959 Chr.3: 149752378-149752378 on Build GRCh38
rs1029704666 Chr.3: 149752353-149752353 on Build GRCh38
rs994027406 Chr.3: 149751996-149751996 on Build GRCh38
rs1047173552 Chr.3: 149752224-149752224 on Build GRCh38
rs753140224 Chr.3: 149752406-149752407 on Build GRCh38
rs759566209 Chr.3: 149752200-149752200 on Build GRCh38
rs769193180 Chr.3: 149752415-149752415 on Build GRCh38
rs111677174 Chr.3: 149752048-149752048 on Build GRCh38
rs202123794 Chr.3: 149752164-149752164 on Build GRCh38
rs543028868 Chr.3: 149752408-149752408 on Build GRCh38
rs979011878 Chr.3: 149752149-149752149 on Build GRCh38
rs913441959 Chr.3: 149752133-149752133 on Build GRCh38
rs989357124 Chr.3: 149751929-149751929 on Build GRCh38
rs375733158 Chr.3: 149751976-149751976 on Build GRCh38
rs549304629 Chr.3: 149751965-149751965 on Build GRCh38
rs750251517 Chr.3: 149752189-149752189 on Build GRCh38
rs756175795 Chr.3: 149752379-149752379 on Build GRCh38
rs1057239692 Chr.3: 149751980-149751980 on Build GRCh38
rs963496881 Chr.3: 149752055-149752055 on Build GRCh38
rs372134103 Chr.3: 149752223-149752223 on Build GRCh38
rs888365902 Chr.3: 149752015-149752015 on Build GRCh38
rs753790050 Chr.3: 149752108-149752109 on Build GRCh38
rs927517155 Chr.3: 149752153-149752153 on Build GRCh38
rs755138058 Chr.3: 149752300-149752300 on Build GRCh38
rs748705578 Chr.3: 149752214-149752214 on Build GRCh38
rs537034390 Chr.3: 149752124-149752124 on Build GRCh38
rs937325032 Chr.3: 149751973-149751973 on Build GRCh38
rs942742168 Chr.3: 149752201-149752201 on Build GRCh38

View Full Product Details