Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148844247-148844738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPB1
Entrez Gene ID
1360
Gene Name
carboxypeptidase B1
Gene Aliases
CPB, PASP, PCPB
UniGene
Hs.477891
Gene Chromosome Location
Chr.3: 148827801-148860187 on Build GRCh38
Transcripts
NM_001871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4972637 c.656A>C p.N219T Chr.3: 148844557-148844557 on Build GRCh38
4972636 c.642C>A p.V214V Chr.3: 148844543-148844543 on Build GRCh38
5886258 c.598G>A p.E200K Chr.3: 148844499-148844499 on Build GRCh38
1039667 c.711C>T p.R237R Chr.3: 148844700-148844700 on Build GRCh38
1039666 c.691C>T p.R231* Chr.3: 148844680-148844680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763101412 Chr.3: 148844702-148844702 on Build GRCh38
rs1059502 Chr.3: 148844523-148844523 on Build GRCh38
rs750694031 Chr.3: 148844701-148844701 on Build GRCh38
rs201609971 Chr.3: 148844586-148844586 on Build GRCh38
rs758976092 Chr.3: 148844439-148844439 on Build GRCh38
rs772900682 Chr.3: 148844589-148844589 on Build GRCh38
rs948029993 Chr.3: 148844362-148844362 on Build GRCh38
rs73866671 Chr.3: 148844683-148844683 on Build GRCh38
rs761703950 Chr.3: 148844698-148844698 on Build GRCh38
rs145781572 Chr.3: 148844477-148844477 on Build GRCh38
rs766411111 Chr.3: 148844710-148844710 on Build GRCh38
rs13318853 Chr.3: 148844612-148844612 on Build GRCh38
rs756420574 Chr.3: 148844440-148844440 on Build GRCh38
rs771244519 Chr.3: 148844578-148844578 on Build GRCh38
rs767341161 Chr.3: 148844436-148844436 on Build GRCh38
rs1036524509 Chr.3: 148844449-148844449 on Build GRCh38
rs373288927 Chr.3: 148844633-148844633 on Build GRCh38
rs765515836 Chr.3: 148844699-148844699 on Build GRCh38
rs755339841 Chr.3: 148844635-148844635 on Build GRCh38
rs751522493 Chr.3: 148844427-148844427 on Build GRCh38
rs781478430 Chr.3: 148844636-148844636 on Build GRCh38
rs147755306 Chr.3: 148844522-148844522 on Build GRCh38
rs771635241 Chr.3: 148844427-148844427 on Build GRCh38
rs768923237 Chr.3: 148844580-148844580 on Build GRCh38
rs759210795 Chr.3: 148844484-148844484 on Build GRCh38
rs747357035 Chr.3: 148844572-148844572 on Build GRCh38
rs13318851 Chr.3: 148844610-148844610 on Build GRCh38
rs141004994 Chr.3: 148844528-148844528 on Build GRCh38
rs376486004 Chr.3: 148844680-148844680 on Build GRCh38
rs77369355 Chr.3: 148844645-148844645 on Build GRCh38
rs750408411 Chr.3: 148844380-148844380 on Build GRCh38
rs779538001 Chr.3: 148844461-148844461 on Build GRCh38
rs754084157 Chr.3: 148844613-148844613 on Build GRCh38
rs771506964 Chr.3: 148844666-148844666 on Build GRCh38
rs752103864 Chr.3: 148844718-148844718 on Build GRCh38
rs780429134 Chr.3: 148844565-148844565 on Build GRCh38
rs531638338 Chr.3: 148844458-148844458 on Build GRCh38
rs759130928 Chr.3: 148844606-148844607 on Build GRCh38
rs757379478 Chr.3: 148844540-148844540 on Build GRCh38
rs370907426 Chr.3: 148844681-148844681 on Build GRCh38
rs574451489 Chr.3: 148844647-148844647 on Build GRCh38
rs940772117 Chr.3: 148844669-148844669 on Build GRCh38
rs770424800 Chr.3: 148844594-148844594 on Build GRCh38
rs375699145 Chr.3: 148844664-148844664 on Build GRCh38
rs558918468 Chr.3: 148844632-148844632 on Build GRCh38
rs546131776 Chr.3: 148844298-148844298 on Build GRCh38
rs368659092 Chr.3: 148844555-148844555 on Build GRCh38
rs977605122 Chr.3: 148844545-148844545 on Build GRCh38
rs1045059248 Chr.3: 148844317-148844317 on Build GRCh38
rs749325370 Chr.3: 148844457-148844457 on Build GRCh38
rs563612174 Chr.3: 148844614-148844614 on Build GRCh38
rs142008836 Chr.3: 148844504-148844504 on Build GRCh38
rs774014207 Chr.3: 148844582-148844583 on Build GRCh38
rs755307416 Chr.3: 148844559-148844559 on Build GRCh38
rs562230201 Chr.3: 148844433-148844433 on Build GRCh38
rs1056940083 Chr.3: 148844406-148844406 on Build GRCh38
rs759063700 Chr.3: 148844602-148844602 on Build GRCh38
rs372764833 Chr.3: 148844591-148844591 on Build GRCh38
rs9835098 Chr.3: 148844638-148844638 on Build GRCh38
rs150241322 Chr.3: 148844517-148844517 on Build GRCh38
rs541135098 Chr.3: 148844616-148844616 on Build GRCh38
rs746360549 Chr.3: 148844556-148844556 on Build GRCh38
rs762180005 Chr.3: 148844618-148844618 on Build GRCh38
rs375031841 Chr.3: 148844634-148844634 on Build GRCh38
rs764338741 Chr.3: 148844534-148844534 on Build GRCh38
rs771615902 Chr.3: 148844620-148844620 on Build GRCh38
rs754031887 Chr.3: 148844535-148844535 on Build GRCh38
rs777875530 Chr.3: 148844453-148844453 on Build GRCh38
rs748986444 Chr.3: 148844595-148844595 on Build GRCh38
rs775977182 Chr.3: 148844481-148844481 on Build GRCh38
rs372662546 Chr.3: 148844656-148844656 on Build GRCh38
rs772768988 Chr.3: 148844496-148844496 on Build GRCh38
rs774243114 Chr.3: 148844503-148844503 on Build GRCh38
rs573286439 Chr.3: 148844358-148844358 on Build GRCh38
rs1046389936 Chr.3: 148844379-148844379 on Build GRCh38
rs765375879 Chr.3: 148844368-148844368 on Build GRCh38
rs145346983 Chr.3: 148844485-148844485 on Build GRCh38
rs386666920 Chr.3: 148844610-148844612 on Build GRCh38
rs113411601 Chr.3: 148844553-148844553 on Build GRCh38

View Full Product Details