Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 216503586-216504157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRRG
Entrez Gene ID
2104
Gene Name
estrogen related receptor gamma
Gene Aliases
ERR3, ERRgamma, NR3B3
UniGene
Hs.444225
Gene Chromosome Location
Chr.1: 216503246-217137757 on Build GRCh38
Transcripts
NM_001134285.2 NM_001243505.1 NM_001243506.1 NM_001243507.1 NM_001243509.1 NM_001243510.2 NM_001243511.2 NM_001243512.1 NM_001243513.1 NM_001243514.1 NM_001243515.1 NM_001243518.1 NM_001243519.1 NM_001438.3 NM_206594.2 NM_206595.2 XM_011509265.2 XM_011509266.2 XM_011509267.1 XM_011509268.2 XM_011509269.2 XM_011509270.1 XM_011509271.2 XM_011509274.1 XM_011509275.1 XM_011509276.1 XM_011509277.1 XM_011509278.1 XM_011509279.1 XM_011509280.2 XM_017000621.1 XM_017000622.1 XM_017000623.1 XM_017000624.1 XM_017000625.1 XM_017000626.1 XM_017000627.1 XM_017000628.1 XM_017000629.1 XM_017000630.1 XM_017000631.1 XM_017000632.1 XM_017000633.1 XM_017000634.1 XM_017000635.1 XM_017000636.1 XM_017000637.1 XM_017000638.1 XM_017000639.1 XM_017000640.1 XM_017000641.1 XM_017000642.1 XM_017000643.1 XM_017000644.1 XM_017000645.1 XM_017000646.1 XM_017000647.1 XM_017000648.1 XM_017000649.1

SNP Detail

SNP ID Chromosome Location
rs574518248 Chr.1: 216503944-216503944 on Build GRCh38
rs915908530 Chr.1: 216503717-216503717 on Build GRCh38
rs11572840 Chr.1: 216503969-216503969 on Build GRCh38
rs1026140176 Chr.1: 216503665-216503665 on Build GRCh38
rs546762074 Chr.1: 216503907-216503907 on Build GRCh38
rs982935631 Chr.1: 216503928-216503928 on Build GRCh38
rs755698568 Chr.1: 216503632-216503632 on Build GRCh38
rs951535962 Chr.1: 216503922-216503922 on Build GRCh38
rs951503652 Chr.1: 216503753-216503753 on Build GRCh38
rs560623597 Chr.1: 216503988-216503988 on Build GRCh38
rs771622441 Chr.1: 216503705-216503705 on Build GRCh38
rs941339850 Chr.1: 216503702-216503702 on Build GRCh38
rs867482022 Chr.1: 216503987-216503987 on Build GRCh38
rs963976963 Chr.1: 216504083-216504083 on Build GRCh38
rs748743220 Chr.1: 216503687-216503687 on Build GRCh38
rs764054247 Chr.1: 216503939-216503941 on Build GRCh38
rs747943310 Chr.1: 216503842-216503842 on Build GRCh38
rs1056414002 Chr.1: 216503990-216503990 on Build GRCh38
rs946477420 Chr.1: 216503955-216503955 on Build GRCh38
rs530654066 Chr.1: 216503985-216503985 on Build GRCh38
rs562030703 Chr.1: 216504087-216504087 on Build GRCh38
rs11572839 Chr.1: 216504015-216504015 on Build GRCh38
rs1005566033 Chr.1: 216504047-216504047 on Build GRCh38
rs964029390 Chr.1: 216503800-216503800 on Build GRCh38
rs963204566 Chr.1: 216504132-216504132 on Build GRCh38
rs768622413 Chr.1: 216503666-216503666 on Build GRCh38
rs984069337 Chr.1: 216503777-216503777 on Build GRCh38
rs1141268 Chr.1: 216503722-216503722 on Build GRCh38
rs1005937504 Chr.1: 216504110-216504110 on Build GRCh38
rs1141267 Chr.1: 216503862-216503862 on Build GRCh38
rs997219352 Chr.1: 216503904-216503904 on Build GRCh38
rs549141557 Chr.1: 216503989-216503989 on Build GRCh38
rs772522274 Chr.1: 216503940-216503940 on Build GRCh38
rs965795514 Chr.1: 216503634-216503639 on Build GRCh38
rs575095019 Chr.1: 216503678-216503678 on Build GRCh38
rs922547531 Chr.1: 216503834-216503834 on Build GRCh38
rs193238923 Chr.1: 216504103-216504103 on Build GRCh38
rs774079271 Chr.1: 216503789-216503789 on Build GRCh38
rs910164207 Chr.1: 216503672-216503672 on Build GRCh38
rs1007902462 Chr.1: 216503833-216503833 on Build GRCh38
rs975447508 Chr.1: 216504112-216504112 on Build GRCh38
rs186760462 Chr.1: 216503667-216503667 on Build GRCh38
rs577524315 Chr.1: 216504006-216504006 on Build GRCh38
rs542242828 Chr.1: 216504062-216504062 on Build GRCh38
rs1017237356 Chr.1: 216503843-216503843 on Build GRCh38
rs766306200 Chr.1: 216503649-216503652 on Build GRCh38
rs183531214 Chr.1: 216503641-216503641 on Build GRCh38
rs886165682 Chr.1: 216504090-216504090 on Build GRCh38
rs537866412 Chr.1: 216503941-216503941 on Build GRCh38
rs901509232 Chr.1: 216503880-216503880 on Build GRCh38
rs1016921153 Chr.1: 216503787-216503787 on Build GRCh38
rs903012038 Chr.1: 216503791-216503791 on Build GRCh38
rs201563931 Chr.1: 216503941-216503942 on Build GRCh38
rs1017066800 Chr.1: 216504123-216504123 on Build GRCh38
rs531585514 Chr.1: 216504051-216504051 on Build GRCh38
rs991479818 Chr.1: 216503709-216503709 on Build GRCh38
rs982987617 Chr.1: 216503664-216503664 on Build GRCh38
rs1001758071 Chr.1: 216503626-216503626 on Build GRCh38
rs1141266 Chr.1: 216503863-216503863 on Build GRCh38
rs1016995984 Chr.1: 216504048-216504048 on Build GRCh38
rs904836695 Chr.1: 216503977-216503977 on Build GRCh38
rs975280062 Chr.1: 216503820-216503820 on Build GRCh38
rs1017706685 Chr.1: 216503616-216503616 on Build GRCh38
rs997166325 Chr.1: 216504038-216504038 on Build GRCh38
rs2809287 Chr.1: 216504008-216504008 on Build GRCh38
rs774937214 Chr.1: 216503897-216503897 on Build GRCh38
rs558821807 Chr.1: 216503642-216503642 on Build GRCh38
rs1025833475 Chr.1: 216504010-216504010 on Build GRCh38
rs934511246 Chr.1: 216503799-216503799 on Build GRCh38
rs148636656 Chr.1: 216503943-216503943 on Build GRCh38

View Full Product Details