Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 40933239-40933833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN1
Entrez Gene ID
1272
Gene Name
contactin 1
Gene Aliases
F3, GP135, MYPCN
UniGene
Hs.143434
Gene Chromosome Location
Chr.12: 40692439-41072412 on Build GRCh38
Transcripts
NM_001256063.1 NM_001256064.1 NM_001843.3 NM_175038.2 XM_005268651.2 XM_006719241.2 XM_011537926.2 XM_011537927.2 XM_017018821.1 XM_017018822.1 XM_017018823.1 XM_017018824.1 XM_017018825.1 XM_017018826.1 XM_017018827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4207049 c.793G>T p.A265S Chr.12: 40933550-40933550 on Build GRCh38
6136667 c.877G>T p.A293S Chr.12: 40933770-40933770 on Build GRCh38
1628585 c.751G>T p.V251L Chr.12: 40933508-40933508 on Build GRCh38
6072348 c.755A>G p.Y252C Chr.12: 40933512-40933512 on Build GRCh38
6631702 c.752T>C p.V251A Chr.12: 40933509-40933509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749834632 Chr.12: 40933765-40933766 on Build GRCh38
rs778449042 Chr.12: 40933581-40933581 on Build GRCh38
rs765149060 Chr.12: 40933789-40933789 on Build GRCh38
rs552921973 Chr.12: 40933290-40933290 on Build GRCh38
rs796927032 Chr.12: 40933624-40933625 on Build GRCh38
rs758956439 Chr.12: 40933481-40933481 on Build GRCh38
rs747626974 Chr.12: 40933589-40933589 on Build GRCh38
rs773699933 Chr.12: 40933595-40933595 on Build GRCh38
rs144047921 Chr.12: 40933719-40933719 on Build GRCh38
rs957638697 Chr.12: 40933630-40933630 on Build GRCh38
rs574860448 Chr.12: 40933366-40933366 on Build GRCh38
rs557373219 Chr.12: 40933276-40933276 on Build GRCh38
rs997165991 Chr.12: 40933265-40933274 on Build GRCh38
rs983528939 Chr.12: 40933406-40933406 on Build GRCh38
rs145955800 Chr.12: 40933794-40933794 on Build GRCh38
rs1036256810 Chr.12: 40933735-40933735 on Build GRCh38
rs375392779 Chr.12: 40933575-40933575 on Build GRCh38
rs773748002 Chr.12: 40933415-40933415 on Build GRCh38
rs776868385 Chr.12: 40933610-40933610 on Build GRCh38
rs200145508 Chr.12: 40933670-40933670 on Build GRCh38
rs761007424 Chr.12: 40933436-40933436 on Build GRCh38
rs1001470110 Chr.12: 40933354-40933354 on Build GRCh38
rs937462249 Chr.12: 40933698-40933698 on Build GRCh38
rs897467981 Chr.12: 40933746-40933746 on Build GRCh38
rs1045867653 Chr.12: 40933319-40933319 on Build GRCh38
rs747859026 Chr.12: 40933708-40933708 on Build GRCh38
rs587776718 Chr.12: 40933764-40933765 on Build GRCh38
rs757218571 Chr.12: 40933509-40933509 on Build GRCh38
rs375708810 Chr.12: 40933676-40933676 on Build GRCh38
rs572715662 Chr.12: 40933616-40933616 on Build GRCh38
rs56323985 Chr.12: 40933625-40933625 on Build GRCh38
rs538243746 Chr.12: 40933332-40933332 on Build GRCh38
rs55976072 Chr.12: 40933624-40933624 on Build GRCh38
rs771359796 Chr.12: 40933721-40933721 on Build GRCh38
rs766198445 Chr.12: 40933781-40933781 on Build GRCh38
rs1021883816 Chr.12: 40933334-40933334 on Build GRCh38
rs746184282 Chr.12: 40933730-40933730 on Build GRCh38
rs900742739 Chr.12: 40933262-40933262 on Build GRCh38
rs779849920 Chr.12: 40933685-40933685 on Build GRCh38
rs796312692 Chr.12: 40933353-40933353 on Build GRCh38
rs752951944 Chr.12: 40933410-40933410 on Build GRCh38
rs1051401838 Chr.12: 40933752-40933752 on Build GRCh38
rs997199034 Chr.12: 40933516-40933516 on Build GRCh38
rs199647238 Chr.12: 40933623-40933623 on Build GRCh38
rs764970741 Chr.12: 40933683-40933683 on Build GRCh38
rs764202885 Chr.12: 40933763-40933763 on Build GRCh38
rs374647402 Chr.12: 40933662-40933662 on Build GRCh38
rs367865918 Chr.12: 40933783-40933783 on Build GRCh38
rs761240540 Chr.12: 40933598-40933598 on Build GRCh38
rs772600743 Chr.12: 40933433-40933433 on Build GRCh38
rs149305190 Chr.12: 40933487-40933487 on Build GRCh38
rs867956389 Chr.12: 40933472-40933472 on Build GRCh38
rs530752245 Chr.12: 40933380-40933380 on Build GRCh38
rs570716833 Chr.12: 40933300-40933300 on Build GRCh38
rs200022020 Chr.12: 40933766-40933766 on Build GRCh38
rs957883894 Chr.12: 40933284-40933284 on Build GRCh38
rs779778967 Chr.12: 40933543-40933543 on Build GRCh38
rs781242588 Chr.12: 40933511-40933511 on Build GRCh38
rs950873423 Chr.12: 40933393-40933393 on Build GRCh38
rs771622826 Chr.12: 40933593-40933593 on Build GRCh38
rs771256157 Chr.12: 40933603-40933603 on Build GRCh38
rs375008317 Chr.12: 40933450-40933450 on Build GRCh38
rs199622338 Chr.12: 40933701-40933701 on Build GRCh38
rs759678214 Chr.12: 40933464-40933464 on Build GRCh38
rs367549221 Chr.12: 40933478-40933478 on Build GRCh38
rs748913992 Chr.12: 40933574-40933574 on Build GRCh38
rs372328375 Chr.12: 40933513-40933513 on Build GRCh38
rs1042788910 Chr.12: 40933661-40933661 on Build GRCh38
rs770242044 Chr.12: 40933741-40933741 on Build GRCh38
rs967383840 Chr.12: 40933682-40933682 on Build GRCh38
rs992199884 Chr.12: 40933378-40933378 on Build GRCh38
rs775839275 Chr.12: 40933748-40933748 on Build GRCh38
rs778411720 Chr.12: 40933706-40933706 on Build GRCh38
rs1034297105 Chr.12: 40933365-40933365 on Build GRCh38
rs946081982 Chr.12: 40933318-40933318 on Build GRCh38
rs201982650 Chr.12: 40933498-40933498 on Build GRCh38
rs763333899 Chr.12: 40933759-40933759 on Build GRCh38
rs752740643 Chr.12: 40933507-40933507 on Build GRCh38
rs554314947 Chr.12: 40933612-40933612 on Build GRCh38
rs200759494 Chr.12: 40933675-40933675 on Build GRCh38
rs756134091 Chr.12: 40933680-40933680 on Build GRCh38
rs552483875 Chr.12: 40933458-40933458 on Build GRCh38
rs201607834 Chr.12: 40933720-40933720 on Build GRCh38
rs753864544 Chr.12: 40933782-40933782 on Build GRCh38
rs1007757023 Chr.12: 40933770-40933770 on Build GRCh38
rs575916673 Chr.12: 40933372-40933372 on Build GRCh38
rs369343021 Chr.12: 40933660-40933660 on Build GRCh38
rs556331560 Chr.12: 40933345-40933345 on Build GRCh38
rs548330075 Chr.12: 40933798-40933798 on Build GRCh38
rs887698166 Chr.12: 40933374-40933374 on Build GRCh38
rs758056182 Chr.12: 40933802-40933802 on Build GRCh38
rs56172264 Chr.12: 40933689-40933689 on Build GRCh38
rs762335667 Chr.12: 40933420-40933420 on Build GRCh38
rs374930846 Chr.12: 40933460-40933460 on Build GRCh38
rs374200408 Chr.12: 40933714-40933714 on Build GRCh38
rs200611219 Chr.12: 40933625-40933625 on Build GRCh38
rs139270901 Chr.12: 40933530-40933530 on Build GRCh38
rs1051869143 Chr.12: 40933623-40933623 on Build GRCh38
rs755959346 Chr.12: 40933517-40933517 on Build GRCh38
rs928683884 Chr.12: 40933697-40933697 on Build GRCh38
rs760650497 Chr.12: 40933775-40933775 on Build GRCh38
rs773751348 Chr.12: 40933434-40933434 on Build GRCh38

View Full Product Details