Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 57070824-57071187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB1
Entrez Gene ID
1600
Gene Name
DAB1, reelin adaptor protein
Gene Aliases
-
UniGene
Hs.477370
Gene Chromosome Location
Chr.1: 56997906-58250539 on Build GRCh38
Transcripts
NM_021080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4632101 c.577G>A p.V193I Chr.1: 57071043-57071043 on Build GRCh38
4874898 c.559-1G>T p.? Chr.1: 57071062-57071062 on Build GRCh38
911115 c.559-1G>T p.? Chr.1: 57071062-57071062 on Build GRCh38
3491294 c.567G>A p.L189L Chr.1: 57071053-57071053 on Build GRCh38
3491293 c.586C>T p.P196S Chr.1: 57071034-57071034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019010433 Chr.1: 57071096-57071096 on Build GRCh38
rs772896005 Chr.1: 57071000-57071000 on Build GRCh38
rs1039425957 Chr.1: 57070955-57070955 on Build GRCh38
rs937795658 Chr.1: 57070993-57070993 on Build GRCh38
rs751617345 Chr.1: 57070943-57070943 on Build GRCh38
rs780497825 Chr.1: 57070977-57070977 on Build GRCh38
rs984010095 Chr.1: 57070922-57070922 on Build GRCh38
rs4912426 Chr.1: 57070988-57070988 on Build GRCh38
rs765746752 Chr.1: 57070905-57070905 on Build GRCh38
rs756572527 Chr.1: 57071104-57071104 on Build GRCh38
rs199893970 Chr.1: 57071003-57071003 on Build GRCh38
rs544090295 Chr.1: 57070936-57070936 on Build GRCh38
rs565752438 Chr.1: 57071146-57071146 on Build GRCh38
rs776647567 Chr.1: 57071005-57071005 on Build GRCh38
rs746312877 Chr.1: 57070960-57070960 on Build GRCh38
rs17418461 Chr.1: 57070909-57070909 on Build GRCh38
rs757380881 Chr.1: 57071081-57071081 on Build GRCh38
rs919688312 Chr.1: 57070968-57070968 on Build GRCh38
rs534967456 Chr.1: 57070876-57070876 on Build GRCh38
rs975675228 Chr.1: 57070904-57070904 on Build GRCh38
rs770303067 Chr.1: 57071004-57071004 on Build GRCh38
rs763213115 Chr.1: 57071073-57071073 on Build GRCh38
rs751691975 Chr.1: 57071079-57071079 on Build GRCh38
rs747874746 Chr.1: 57070980-57070980 on Build GRCh38
rs781738144 Chr.1: 57070948-57070948 on Build GRCh38
rs764886343 Chr.1: 57070932-57070932 on Build GRCh38
rs201824114 Chr.1: 57071078-57071078 on Build GRCh38
rs1019481441 Chr.1: 57070882-57070882 on Build GRCh38
rs9803960 Chr.1: 57071072-57071072 on Build GRCh38
rs756514622 Chr.1: 57070964-57070964 on Build GRCh38
rs765086569 Chr.1: 57071055-57071055 on Build GRCh38
rs769146310 Chr.1: 57071093-57071093 on Build GRCh38
rs1021256972 Chr.1: 57071059-57071059 on Build GRCh38
rs757746572 Chr.1: 57070944-57070944 on Build GRCh38
rs752925636 Chr.1: 57070934-57070934 on Build GRCh38
rs904997824 Chr.1: 57071151-57071151 on Build GRCh38
rs371998081 Chr.1: 57071043-57071043 on Build GRCh38
rs931199926 Chr.1: 57070916-57070916 on Build GRCh38
rs750944305 Chr.1: 57071087-57071087 on Build GRCh38
rs908176441 Chr.1: 57070969-57070969 on Build GRCh38
rs762669081 Chr.1: 57070901-57070901 on Build GRCh38
rs767438515 Chr.1: 57071082-57071082 on Build GRCh38
rs778885982 Chr.1: 57070880-57070880 on Build GRCh38
rs1002466082 Chr.1: 57071155-57071155 on Build GRCh38

View Full Product Details