Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111312036-111312604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIA
Entrez Gene ID
27159
Gene Name
chitinase, acidic
Gene Aliases
AMCASE, CHIT2, TSA1902
UniGene
Hs.128814
Gene Chromosome Location
Chr.1: 111290852-111320566 on Build GRCh38
Transcripts
NM_001040623.2 NM_001258001.1 NM_001258002.1 NM_001258003.1 NM_001258004.1 NM_001258005.1 NM_021797.3 NM_201653.3 XM_006710577.3 XM_017001048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1732858 c.119G>A p.R40H Chr.1: 111312253-111312253 on Build GRCh38
462593 c.105G>A p.R35R Chr.1: 111312239-111312239 on Build GRCh38
6628369 c.201C>A p.T67T Chr.1: 111312335-111312335 on Build GRCh38
5443588 c.240C>T p.F80F Chr.1: 111312374-111312374 on Build GRCh38
674368 c.212G>T p.W71L Chr.1: 111312346-111312346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027883579 Chr.1: 111312506-111312506 on Build GRCh38
rs780786351 Chr.1: 111312401-111312401 on Build GRCh38
rs369742654 Chr.1: 111312217-111312217 on Build GRCh38
rs748311921 Chr.1: 111312233-111312233 on Build GRCh38
rs765842701 Chr.1: 111312306-111312306 on Build GRCh38
rs886766639 Chr.1: 111312482-111312482 on Build GRCh38
rs41282490 Chr.1: 111312119-111312119 on Build GRCh38
rs373325850 Chr.1: 111312252-111312252 on Build GRCh38
rs771992473 Chr.1: 111312236-111312236 on Build GRCh38
rs752677743 Chr.1: 111312342-111312342 on Build GRCh38
rs772997390 Chr.1: 111312290-111312290 on Build GRCh38
rs769569777 Chr.1: 111312420-111312420 on Build GRCh38
rs769930181 Chr.1: 111312143-111312143 on Build GRCh38
rs776231839 Chr.1: 111312186-111312186 on Build GRCh38
rs376456634 Chr.1: 111312260-111312260 on Build GRCh38
rs764153298 Chr.1: 111312309-111312309 on Build GRCh38
rs748020999 Chr.1: 111312168-111312168 on Build GRCh38
rs41282492 Chr.1: 111312267-111312267 on Build GRCh38
rs769888097 Chr.1: 111312277-111312277 on Build GRCh38
rs933716223 Chr.1: 111312480-111312480 on Build GRCh38
rs200537454 Chr.1: 111312326-111312326 on Build GRCh38
rs368959092 Chr.1: 111312370-111312370 on Build GRCh38
rs1051241328 Chr.1: 111312481-111312481 on Build GRCh38
rs61752461 Chr.1: 111312237-111312237 on Build GRCh38
rs149356213 Chr.1: 111312229-111312229 on Build GRCh38
rs1039148446 Chr.1: 111312453-111312453 on Build GRCh38
rs943559067 Chr.1: 111312403-111312403 on Build GRCh38
rs768877632 Chr.1: 111312156-111312156 on Build GRCh38
rs1042877044 Chr.1: 111312346-111312346 on Build GRCh38
rs553741768 Chr.1: 111312134-111312134 on Build GRCh38
rs778188110 Chr.1: 111312393-111312393 on Build GRCh38
rs748413001 Chr.1: 111312293-111312293 on Build GRCh38
rs202013630 Chr.1: 111312331-111312331 on Build GRCh38
rs74108971 Chr.1: 111312527-111312527 on Build GRCh38
rs905754758 Chr.1: 111312162-111312162 on Build GRCh38
rs745790378 Chr.1: 111312304-111312304 on Build GRCh38
rs749350753 Chr.1: 111312227-111312227 on Build GRCh38
rs770951786 Chr.1: 111312243-111312243 on Build GRCh38
rs759898807 Chr.1: 111312335-111312335 on Build GRCh38
rs576835560 Chr.1: 111312187-111312187 on Build GRCh38
rs186163063 Chr.1: 111312333-111312333 on Build GRCh38
rs188897015 Chr.1: 111312253-111312253 on Build GRCh38
rs41282494 Chr.1: 111312273-111312273 on Build GRCh38
rs753128380 Chr.1: 111312496-111312496 on Build GRCh38
rs913902553 Chr.1: 111312328-111312328 on Build GRCh38
rs10494133 Chr.1: 111312066-111312066 on Build GRCh38
rs983607276 Chr.1: 111312544-111312544 on Build GRCh38
rs900785606 Chr.1: 111312465-111312465 on Build GRCh38
rs746963750 Chr.1: 111312238-111312238 on Build GRCh38
rs927253771 Chr.1: 111312305-111312305 on Build GRCh38
rs61752527 Chr.1: 111312296-111312296 on Build GRCh38
rs767144908 Chr.1: 111312341-111312341 on Build GRCh38
rs4423003 Chr.1: 111312133-111312133 on Build GRCh38
rs757700023 Chr.1: 111312211-111312211 on Build GRCh38
rs1018953456 Chr.1: 111312417-111312417 on Build GRCh38
rs370883122 Chr.1: 111312302-111312302 on Build GRCh38
rs569468207 Chr.1: 111312398-111312398 on Build GRCh38
rs780134341 Chr.1: 111312226-111312226 on Build GRCh38
rs1014631783 Chr.1: 111312550-111312550 on Build GRCh38
rs760618080 Chr.1: 111312180-111312180 on Build GRCh38
rs41282496 Chr.1: 111312316-111312316 on Build GRCh38
rs779649610 Chr.1: 111312438-111312438 on Build GRCh38
rs767494822 Chr.1: 111312099-111312100 on Build GRCh38
rs979641265 Chr.1: 111312350-111312350 on Build GRCh38
rs756560380 Chr.1: 111312219-111312219 on Build GRCh38
rs746838864 Chr.1: 111312301-111312301 on Build GRCh38
rs758521259 Chr.1: 111312369-111312369 on Build GRCh38
rs766000740 Chr.1: 111312185-111312185 on Build GRCh38
rs375891484 Chr.1: 111312327-111312327 on Build GRCh38
rs773346687 Chr.1: 111312176-111312176 on Build GRCh38
rs766721992 Chr.1: 111312332-111312332 on Build GRCh38
rs754284897 Chr.1: 111312289-111312289 on Build GRCh38
rs779249634 Chr.1: 111312232-111312232 on Build GRCh38
rs745831611 Chr.1: 111312418-111312418 on Build GRCh38
rs771747939 Chr.1: 111312170-111312170 on Build GRCh38
rs749426433 Chr.1: 111312090-111312090 on Build GRCh38
rs1054323888 Chr.1: 111312317-111312317 on Build GRCh38
rs767864863 Chr.1: 111312212-111312212 on Build GRCh38
rs182022651 Chr.1: 111312303-111312303 on Build GRCh38
rs376426347 Chr.1: 111312577-111312577 on Build GRCh38
rs754985114 Chr.1: 111312230-111312230 on Build GRCh38
rs1047420587 Chr.1: 111312093-111312093 on Build GRCh38
rs893923292 Chr.1: 111312193-111312193 on Build GRCh38
rs141615278 Chr.1: 111312059-111312059 on Build GRCh38
rs759202544 Chr.1: 111312323-111312323 on Build GRCh38
rs921020515 Chr.1: 111312460-111312460 on Build GRCh38
rs796090026 Chr.1: 111312584-111312584 on Build GRCh38
rs759140330 Chr.1: 111312189-111312189 on Build GRCh38
rs555346981 Chr.1: 111312087-111312087 on Build GRCh38
rs766185173 Chr.1: 111312196-111312196 on Build GRCh38
rs925447190 Chr.1: 111312251-111312251 on Build GRCh38
rs751406036 Chr.1: 111312243-111312243 on Build GRCh38
rs780045344 Chr.1: 111312150-111312150 on Build GRCh38
rs1043911399 Chr.1: 111312242-111312242 on Build GRCh38
rs756648314 Chr.1: 111312276-111312276 on Build GRCh38
rs762447155 Chr.1: 111312272-111312272 on Build GRCh38
rs752270984 Chr.1: 111312199-111312199 on Build GRCh38
rs920358501 Chr.1: 111312081-111312081 on Build GRCh38
rs778788058 Chr.1: 111312291-111312291 on Build GRCh38
rs548844817 Chr.1: 111312583-111312583 on Build GRCh38
rs545496572 Chr.1: 111312191-111312191 on Build GRCh38
rs750741444 Chr.1: 111312275-111312275 on Build GRCh38
rs777741609 Chr.1: 111312300-111312300 on Build GRCh38
rs769351912 Chr.1: 111312313-111312313 on Build GRCh38
rs749528386 Chr.1: 111312151-111312151 on Build GRCh38

View Full Product Details