Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160830049-160830648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD244
Entrez Gene ID
51744
Gene Name
CD244 molecule
Gene Aliases
2B4, NAIL, NKR2B4, Nmrk, SLAMF4
UniGene
Hs.157872
Gene Chromosome Location
Chr.1: 160830160-160862902 on Build GRCh38
Transcripts
NM_001166663.1 NM_001166664.1 NM_016382.3 XM_011509623.2

SNP Detail

SNP ID Chromosome Location
rs994957524 Chr.1: 160830420-160830420 on Build GRCh38
rs111696936 Chr.1: 160830543-160830543 on Build GRCh38
rs1026772069 Chr.1: 160830625-160830625 on Build GRCh38
rs750483311 Chr.1: 160830319-160830319 on Build GRCh38
rs889784713 Chr.1: 160830129-160830129 on Build GRCh38
rs573638806 Chr.1: 160830450-160830450 on Build GRCh38
rs545131122 Chr.1: 160830593-160830593 on Build GRCh38
rs557000799 Chr.1: 160830522-160830522 on Build GRCh38
rs981155322 Chr.1: 160830344-160830344 on Build GRCh38
rs931232843 Chr.1: 160830116-160830116 on Build GRCh38
rs908472231 Chr.1: 160830223-160830223 on Build GRCh38
rs572990782 Chr.1: 160830100-160830100 on Build GRCh38
rs890716153 Chr.1: 160830372-160830372 on Build GRCh38
rs781236856 Chr.1: 160830419-160830419 on Build GRCh38
rs958280045 Chr.1: 160830284-160830284 on Build GRCh38
rs539450494 Chr.1: 160830559-160830559 on Build GRCh38
rs573581526 Chr.1: 160830524-160830524 on Build GRCh38
rs1053269273 Chr.1: 160830347-160830347 on Build GRCh38
rs565823873 Chr.1: 160830391-160830391 on Build GRCh38
rs952638970 Chr.1: 160830204-160830204 on Build GRCh38
rs573003338 Chr.1: 160830591-160830591 on Build GRCh38
rs935425027 Chr.1: 160830481-160830481 on Build GRCh38
rs775150468 Chr.1: 160830097-160830097 on Build GRCh38
rs534193169 Chr.1: 160830221-160830221 on Build GRCh38
rs939817848 Chr.1: 160830098-160830098 on Build GRCh38
rs961131059 Chr.1: 160830365-160830365 on Build GRCh38
rs995265366 Chr.1: 160830594-160830594 on Build GRCh38
rs1028164045 Chr.1: 160830421-160830421 on Build GRCh38
rs1012914693 Chr.1: 160830286-160830286 on Build GRCh38
rs181775292 Chr.1: 160830169-160830169 on Build GRCh38
rs987489730 Chr.1: 160830120-160830120 on Build GRCh38
rs1023929406 Chr.1: 160830304-160830304 on Build GRCh38
rs931202834 Chr.1: 160830427-160830427 on Build GRCh38
rs1049712327 Chr.1: 160830122-160830122 on Build GRCh38
rs760316019 Chr.1: 160830102-160830102 on Build GRCh38
rs112997964 Chr.1: 160830442-160830442 on Build GRCh38
rs755283775 Chr.1: 160830526-160830526 on Build GRCh38
rs552945240 Chr.1: 160830567-160830567 on Build GRCh38
rs1053841004 Chr.1: 160830512-160830512 on Build GRCh38
rs1009175659 Chr.1: 160830568-160830568 on Build GRCh38
rs902576227 Chr.1: 160830468-160830468 on Build GRCh38
rs964799219 Chr.1: 160830156-160830156 on Build GRCh38
rs928403015 Chr.1: 160830096-160830096 on Build GRCh38
rs534419941 Chr.1: 160830425-160830425 on Build GRCh38
rs915570759 Chr.1: 160830278-160830278 on Build GRCh38
rs563296548 Chr.1: 160830297-160830297 on Build GRCh38
rs985775428 Chr.1: 160830429-160830429 on Build GRCh38
rs565490611 Chr.1: 160830599-160830599 on Build GRCh38
rs948377424 Chr.1: 160830490-160830490 on Build GRCh38
rs1017580094 Chr.1: 160830130-160830130 on Build GRCh38
rs751820157 Chr.1: 160830402-160830402 on Build GRCh38
rs919791601 Chr.1: 160830180-160830180 on Build GRCh38
rs185116894 Chr.1: 160830437-160830437 on Build GRCh38
rs919761443 Chr.1: 160830403-160830403 on Build GRCh38
rs144853685 Chr.1: 160830252-160830252 on Build GRCh38
rs1041093935 Chr.1: 160830202-160830202 on Build GRCh38
rs57280672 Chr.1: 160830509-160830509 on Build GRCh38
rs528926615 Chr.1: 160830317-160830317 on Build GRCh38
rs58480178 Chr.1: 160830448-160830448 on Build GRCh38
rs190770003 Chr.1: 160830539-160830539 on Build GRCh38
rs915601924 Chr.1: 160830488-160830488 on Build GRCh38
rs757544762 Chr.1: 160830352-160830352 on Build GRCh38
rs964687461 Chr.1: 160830405-160830405 on Build GRCh38
rs961078490 Chr.1: 160830598-160830598 on Build GRCh38
rs1042700562 Chr.1: 160830523-160830523 on Build GRCh38
rs938561607 Chr.1: 160830238-160830238 on Build GRCh38
rs148555185 Chr.1: 160830336-160830336 on Build GRCh38
rs1002512114 Chr.1: 160830571-160830571 on Build GRCh38
rs78916399 Chr.1: 160830222-160830222 on Build GRCh38
rs765388552 Chr.1: 160830263-160830263 on Build GRCh38
rs79250720 Chr.1: 160830254-160830254 on Build GRCh38
rs919813606 Chr.1: 160830109-160830109 on Build GRCh38

View Full Product Details