Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 60459591-60460162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL11A
Entrez Gene ID
53335
Gene Name
B-cell CLL/lymphoma 11A
Gene Aliases
BCL11A-L, BCL11A-S, BCL11A-XL, BCL11a-M, CTIP1, EVI9, HBFQTL5, ZNF856
UniGene
Hs.370549
Gene Chromosome Location
Chr.2: 60451167-60553498 on Build GRCh38
Transcripts
NM_018014.3 NM_022893.3 NM_138559.1 XM_011532909.1 XM_011532910.1 XM_011532912.1 XM_017004333.1 XM_017004334.1 XM_017004335.1 XM_017004336.1 XM_017004337.1 XM_017004338.1

SNP Detail

SNP ID Chromosome Location
rs992787664 Chr.2: 60459736-60459736 on Build GRCh38
rs189586228 Chr.2: 60460100-60460100 on Build GRCh38
rs980375948 Chr.2: 60459768-60459768 on Build GRCh38
rs950899189 Chr.2: 60459947-60459950 on Build GRCh38
rs926051562 Chr.2: 60459791-60459791 on Build GRCh38
rs945238871 Chr.2: 60460103-60460103 on Build GRCh38
rs969087741 Chr.2: 60460068-60460068 on Build GRCh38
rs920371061 Chr.2: 60459764-60459764 on Build GRCh38
rs1015474601 Chr.2: 60459703-60459703 on Build GRCh38
rs996273627 Chr.2: 60459895-60459895 on Build GRCh38
rs950949738 Chr.2: 60459714-60459714 on Build GRCh38
rs181979594 Chr.2: 60459950-60459950 on Build GRCh38
rs927652654 Chr.2: 60459769-60459769 on Build GRCh38
rs1029234035 Chr.2: 60459654-60459654 on Build GRCh38
rs760683236 Chr.2: 60459829-60459829 on Build GRCh38
rs757415243 Chr.2: 60459934-60459934 on Build GRCh38
rs540452802 Chr.2: 60459849-60459849 on Build GRCh38
rs993069715 Chr.2: 60459948-60459948 on Build GRCh38
rs576925492 Chr.2: 60459919-60459919 on Build GRCh38
rs564815635 Chr.2: 60459877-60459877 on Build GRCh38
rs955098839 Chr.2: 60459699-60459699 on Build GRCh38
rs973573664 Chr.2: 60460002-60460002 on Build GRCh38
rs562096198 Chr.2: 60459963-60459963 on Build GRCh38
rs962272587 Chr.2: 60459718-60459718 on Build GRCh38
rs980525650 Chr.2: 60459981-60459981 on Build GRCh38
rs897410090 Chr.2: 60460108-60460108 on Build GRCh38
rs371548533 Chr.2: 60460059-60460059 on Build GRCh38
rs150974916 Chr.2: 60459702-60459702 on Build GRCh38
rs548438060 Chr.2: 60460025-60460025 on Build GRCh38
rs980492194 Chr.2: 60460073-60460073 on Build GRCh38
rs12623650 Chr.2: 60460044-60460044 on Build GRCh38
rs74493918 Chr.2: 60459806-60459806 on Build GRCh38
rs1039117117 Chr.2: 60460136-60460136 on Build GRCh38
rs141136901 Chr.2: 60459795-60459795 on Build GRCh38
rs962506231 Chr.2: 60459987-60459987 on Build GRCh38
rs764164303 Chr.2: 60459878-60459878 on Build GRCh38
rs1051666767 Chr.2: 60459812-60459812 on Build GRCh38
rs185460868 Chr.2: 60459984-60459984 on Build GRCh38
rs963559185 Chr.2: 60459628-60459628 on Build GRCh38
rs147346851 Chr.2: 60460017-60460018 on Build GRCh38
rs1026899119 Chr.2: 60459934-60459935 on Build GRCh38
rs570854019 Chr.2: 60460129-60460129 on Build GRCh38
rs946609784 Chr.2: 60459847-60459847 on Build GRCh38
rs566844089 Chr.2: 60460029-60460029 on Build GRCh38
rs1043144217 Chr.2: 60459854-60459854 on Build GRCh38
rs1017330837 Chr.2: 60459861-60459861 on Build GRCh38
rs199562853 Chr.2: 60460051-60460052 on Build GRCh38
rs1009437839 Chr.2: 60459811-60459812 on Build GRCh38
rs148734179 Chr.2: 60460104-60460104 on Build GRCh38
rs368389144 Chr.2: 60459810-60459810 on Build GRCh38
rs970024578 Chr.2: 60459978-60459978 on Build GRCh38
rs180821769 Chr.2: 60460124-60460124 on Build GRCh38
rs532272289 Chr.2: 60459936-60459936 on Build GRCh38
rs890390902 Chr.2: 60460119-60460119 on Build GRCh38
rs188924437 Chr.2: 60459767-60459767 on Build GRCh38
rs184557786 Chr.2: 60459758-60459758 on Build GRCh38
rs112094046 Chr.2: 60459930-60459930 on Build GRCh38
rs917250052 Chr.2: 60459748-60459748 on Build GRCh38
rs1027792124 Chr.2: 60460056-60460056 on Build GRCh38
rs973998615 Chr.2: 60459678-60459678 on Build GRCh38
rs1007203256 Chr.2: 60459860-60459860 on Build GRCh38

View Full Product Details