Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24299935-24300533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOP9
Entrez Gene ID
161424
Gene Name
NOP9 nucleolar protein
Gene Aliases
C14orf21
UniGene
Hs.708040
Gene Chromosome Location
Chr.14: 24299854-24309126 on Build GRCh38
Transcripts
NM_001286367.1 NM_174913.2 XM_005267385.1 XM_011536526.1 XM_011536527.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3386499 c.372C>T p.R124R Chr.14: 24300532-24300532 on Build GRCh38
6415770 c.140C>A p.S47* Chr.14: 24300094-24300094 on Build GRCh38
1179550 c.292C>T p.L98L Chr.14: 24300452-24300452 on Build GRCh38
3983212 c.241G>T p.E81* Chr.14: 24300195-24300195 on Build GRCh38
6386430 c.71G>A p.G24E Chr.14: 24300025-24300025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527573528 Chr.14: 24300119-24300119 on Build GRCh38
rs779701180 Chr.14: 24300120-24300120 on Build GRCh38
rs554554917 Chr.14: 24300423-24300423 on Build GRCh38
rs867141047 Chr.14: 24299992-24299992 on Build GRCh38
rs762205692 Chr.14: 24299958-24299958 on Build GRCh38
rs940891771 Chr.14: 24299993-24299993 on Build GRCh38
rs750670721 Chr.14: 24299966-24299966 on Build GRCh38
rs753890120 Chr.14: 24300076-24300076 on Build GRCh38
rs763104098 Chr.14: 24300047-24300047 on Build GRCh38
rs769154283 Chr.14: 24300138-24300138 on Build GRCh38
rs183868211 Chr.14: 24300016-24300016 on Build GRCh38
rs747616298 Chr.14: 24300137-24300137 on Build GRCh38
rs369013513 Chr.14: 24300497-24300497 on Build GRCh38
rs774511274 Chr.14: 24300177-24300177 on Build GRCh38
rs990455440 Chr.14: 24300265-24300265 on Build GRCh38
rs376742385 Chr.14: 24300384-24300384 on Build GRCh38
rs767707407 Chr.14: 24300112-24300112 on Build GRCh38
rs936159576 Chr.14: 24300260-24300260 on Build GRCh38
rs913476536 Chr.14: 24300269-24300269 on Build GRCh38
rs377258662 Chr.14: 24300425-24300425 on Build GRCh38
rs759825430 Chr.14: 24300111-24300111 on Build GRCh38
rs779444373 Chr.14: 24300391-24300391 on Build GRCh38
rs201249356 Chr.14: 24300182-24300182 on Build GRCh38
rs758022321 Chr.14: 24300088-24300088 on Build GRCh38
rs188210549 Chr.14: 24300322-24300322 on Build GRCh38
rs79529000 Chr.14: 24300094-24300094 on Build GRCh38
rs11848295 Chr.14: 24300105-24300105 on Build GRCh38
rs775747142 Chr.14: 24300185-24300185 on Build GRCh38
rs185864031 Chr.14: 24300440-24300440 on Build GRCh38
rs11158632 Chr.14: 24300457-24300457 on Build GRCh38
rs776509560 Chr.14: 24300438-24300438 on Build GRCh38
rs1055102990 Chr.14: 24300071-24300071 on Build GRCh38
rs138108727 Chr.14: 24300492-24300492 on Build GRCh38
rs1017264676 Chr.14: 24300166-24300166 on Build GRCh38
rs780699944 Chr.14: 24300131-24300131 on Build GRCh38
rs779607641 Chr.14: 24300484-24300484 on Build GRCh38
rs138618033 Chr.14: 24299974-24299974 on Build GRCh38
rs769534157 Chr.14: 24300228-24300228 on Build GRCh38
rs749741873 Chr.14: 24300149-24300149 on Build GRCh38
rs985283141 Chr.14: 24299989-24299989 on Build GRCh38
rs376446684 Chr.14: 24300108-24300108 on Build GRCh38
rs768124512 Chr.14: 24300029-24300029 on Build GRCh38
rs932075816 Chr.14: 24300331-24300331 on Build GRCh38
rs568672109 Chr.14: 24300376-24300376 on Build GRCh38
rs780009697 Chr.14: 24299968-24299968 on Build GRCh38
rs142900863 Chr.14: 24300419-24300419 on Build GRCh38
rs757196473 Chr.14: 24300079-24300079 on Build GRCh38
rs749212269 Chr.14: 24300003-24300003 on Build GRCh38
rs200199640 Chr.14: 24300478-24300478 on Build GRCh38
rs949771625 Chr.14: 24299976-24299976 on Build GRCh38
rs754654809 Chr.14: 24300233-24300233 on Build GRCh38
rs192494773 Chr.14: 24300441-24300441 on Build GRCh38
rs768214653 Chr.14: 24300095-24300095 on Build GRCh38
rs777894674 Chr.14: 24300001-24300001 on Build GRCh38
rs1040358648 Chr.14: 24300098-24300098 on Build GRCh38
rs773196067 Chr.14: 24300445-24300445 on Build GRCh38
rs953993650 Chr.14: 24300115-24300115 on Build GRCh38
rs560111090 Chr.14: 24300230-24300230 on Build GRCh38
rs369739999 Chr.14: 24300117-24300117 on Build GRCh38
rs201814151 Chr.14: 24300128-24300128 on Build GRCh38
rs766582037 Chr.14: 24300052-24300052 on Build GRCh38
rs777139996 Chr.14: 24300383-24300383 on Build GRCh38
rs35291524 Chr.14: 24300256-24300257 on Build GRCh38
rs375008436 Chr.14: 24300044-24300044 on Build GRCh38
rs762796426 Chr.14: 24300217-24300217 on Build GRCh38
rs541939642 Chr.14: 24300452-24300452 on Build GRCh38
rs199501119 Chr.14: 24300198-24300198 on Build GRCh38
rs771282026 Chr.14: 24300387-24300387 on Build GRCh38
rs866425700 Chr.14: 24300415-24300415 on Build GRCh38
rs148479739 Chr.14: 24300451-24300451 on Build GRCh38
rs771699726 Chr.14: 24300019-24300019 on Build GRCh38
rs1023897666 Chr.14: 24300192-24300192 on Build GRCh38
rs752324730 Chr.14: 24300357-24300357 on Build GRCh38
rs768158760 Chr.14: 24300115-24300126 on Build GRCh38
rs545911691 Chr.14: 24300089-24300089 on Build GRCh38
rs748666928 Chr.14: 24300100-24300100 on Build GRCh38
rs1016735269 Chr.14: 24300456-24300456 on Build GRCh38
rs577617515 Chr.14: 24300439-24300439 on Build GRCh38
rs1043379511 Chr.14: 24300393-24300393 on Build GRCh38
rs746685746 Chr.14: 24300011-24300011 on Build GRCh38
rs532653052 Chr.14: 24300114-24300114 on Build GRCh38
rs751697679 Chr.14: 24300054-24300054 on Build GRCh38
rs746731632 Chr.14: 24300126-24300126 on Build GRCh38
rs1050672721 Chr.14: 24300307-24300307 on Build GRCh38
rs879362255 Chr.14: 24300399-24300399 on Build GRCh38
rs776115625 Chr.14: 24300096-24300096 on Build GRCh38
rs746360271 Chr.14: 24300485-24300485 on Build GRCh38
rs902152251 Chr.14: 24300395-24300395 on Build GRCh38
rs902246755 Chr.14: 24300064-24300064 on Build GRCh38
rs569219407 Chr.14: 24300316-24300316 on Build GRCh38
rs760881723 Chr.14: 24300202-24300202 on Build GRCh38
rs777106325 Chr.14: 24300115-24300116 on Build GRCh38
rs767689193 Chr.14: 24300068-24300068 on Build GRCh38
rs368061098 Chr.14: 24300023-24300023 on Build GRCh38
rs771390834 Chr.14: 24300158-24300158 on Build GRCh38
rs770234990 Chr.14: 24300104-24300104 on Build GRCh38
rs755153399 Chr.14: 24300065-24300065 on Build GRCh38
rs777347694 Chr.14: 24300466-24300466 on Build GRCh38
rs923447349 Chr.14: 24300294-24300294 on Build GRCh38
rs201493718 Chr.14: 24300020-24300020 on Build GRCh38
rs548473650 Chr.14: 24300354-24300354 on Build GRCh38
rs746198829 Chr.14: 24300400-24300400 on Build GRCh38
rs144579665 Chr.14: 24300304-24300304 on Build GRCh38
rs770904987 Chr.14: 24300004-24300004 on Build GRCh38
rs753241472 Chr.14: 24300470-24300470 on Build GRCh38
rs756150491 Chr.14: 24299995-24299995 on Build GRCh38
rs745631280 Chr.14: 24300087-24300087 on Build GRCh38
rs532325791 Chr.14: 24300275-24300275 on Build GRCh38
rs563998141 Chr.14: 24300512-24300512 on Build GRCh38
rs747260921 Chr.14: 24300504-24300504 on Build GRCh38
rs768848919 Chr.14: 24300435-24300435 on Build GRCh38
rs976542454 Chr.14: 24300283-24300283 on Build GRCh38
rs201129797 Chr.14: 24300183-24300183 on Build GRCh38
rs927495856 Chr.14: 24300259-24300259 on Build GRCh38
rs773814064 Chr.14: 24300145-24300145 on Build GRCh38
rs543850916 Chr.14: 24299967-24299967 on Build GRCh38
rs940768849 Chr.14: 24300248-24300248 on Build GRCh38
rs765565601 Chr.14: 24299960-24299960 on Build GRCh38
rs768704589 Chr.14: 24300511-24300511 on Build GRCh38
rs768384595 Chr.14: 24299955-24299955 on Build GRCh38
rs764058624 Chr.14: 24300212-24300212 on Build GRCh38
rs778587952 Chr.14: 24300081-24300081 on Build GRCh38
rs901819165 Chr.14: 24300323-24300323 on Build GRCh38
rs762297365 Chr.14: 24300043-24300043 on Build GRCh38
rs561947101 Chr.14: 24300116-24300116 on Build GRCh38
rs371208153 Chr.14: 24300042-24300042 on Build GRCh38
rs143657838 Chr.14: 24300494-24300494 on Build GRCh38
rs200068328 Chr.14: 24300106-24300106 on Build GRCh38
rs750434480 Chr.14: 24300213-24300213 on Build GRCh38
rs780862988 Chr.14: 24300235-24300235 on Build GRCh38
rs766203493 Chr.14: 24300220-24300220 on Build GRCh38

View Full Product Details