Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73506279-73506727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT1
Entrez Gene ID
641371
Gene Name
acyl-CoA thioesterase 1
Gene Aliases
ACH2, CTE-1, LACH2
UniGene
Hs.446685
Gene Chromosome Location
Chr.14: 73493021-73543799 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_011536761.2 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_011536761.2 XM_017021289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4652536 c.1937G>A p.R646Q Chr.14: 73506516-73506516 on Build GRCh38
2137129 c.1967T>G p.I656S Chr.14: 73506486-73506486 on Build GRCh38
4052282 c.1806C>A p.A602A Chr.14: 73506506-73506506 on Build GRCh38
6236360 c.1893C>A p.H631Q Chr.14: 73506560-73506560 on Build GRCh38
1740134 c.1826T>A p.I609N Chr.14: 73506486-73506486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200485796 Chr.14: 73506686-73506686 on Build GRCh38
rs752604931 Chr.14: 73506535-73506535 on Build GRCh38
rs139632154 Chr.14: 73506489-73506489 on Build GRCh38
rs576219698 Chr.14: 73506513-73506513 on Build GRCh38
rs201421441 Chr.14: 73506684-73506684 on Build GRCh38
rs372980757 Chr.14: 73506516-73506516 on Build GRCh38
rs778061121 Chr.14: 73506614-73506614 on Build GRCh38
rs867910298 Chr.14: 73506685-73506685 on Build GRCh38
rs753804577 Chr.14: 73506591-73506591 on Build GRCh38
rs778922454 Chr.14: 73506508-73506508 on Build GRCh38
rs375723728 Chr.14: 73506490-73506490 on Build GRCh38
rs12894674 Chr.14: 73506432-73506432 on Build GRCh38
rs1003500474 Chr.14: 73506365-73506365 on Build GRCh38
rs758405437 Chr.14: 73506608-73506608 on Build GRCh38
rs933331036 Chr.14: 73506331-73506331 on Build GRCh38
rs994884017 Chr.14: 73506407-73506407 on Build GRCh38
rs910595349 Chr.14: 73506661-73506661 on Build GRCh38
rs933275454 Chr.14: 73506648-73506648 on Build GRCh38
rs772454432 Chr.14: 73506421-73506421 on Build GRCh38
rs772331853 Chr.14: 73506511-73506511 on Build GRCh38
rs755072557 Chr.14: 73506503-73506503 on Build GRCh38
rs370145878 Chr.14: 73506485-73506485 on Build GRCh38
rs773664740 Chr.14: 73506423-73506423 on Build GRCh38
rs138786560 Chr.14: 73506471-73506471 on Build GRCh38
rs12894816 Chr.14: 73506460-73506460 on Build GRCh38
rs12894399 Chr.14: 73506428-73506428 on Build GRCh38
rs950833459 Chr.14: 73506455-73506455 on Build GRCh38
rs75985060 Chr.14: 73506338-73506338 on Build GRCh38
rs373861116 Chr.14: 73506466-73506466 on Build GRCh38
rs144326822 Chr.14: 73506615-73506615 on Build GRCh38
rs370313641 Chr.14: 73506514-73506514 on Build GRCh38
rs199744072 Chr.14: 73506686-73506686 on Build GRCh38
rs895155592 Chr.14: 73506388-73506388 on Build GRCh38
rs267604048 Chr.14: 73506476-73506476 on Build GRCh38
rs760076062 Chr.14: 73506525-73506525 on Build GRCh38
rs191350723 Chr.14: 73506385-73506385 on Build GRCh38
rs777212501 Chr.14: 73506518-73506518 on Build GRCh38
rs966695362 Chr.14: 73506472-73506472 on Build GRCh38
rs766038976 Chr.14: 73506569-73506569 on Build GRCh38
rs59419201 Chr.14: 73506572-73506572 on Build GRCh38
rs35842680 Chr.14: 73506660-73506660 on Build GRCh38
rs771353866 Chr.14: 73506620-73506620 on Build GRCh38
rs886506655 Chr.14: 73506679-73506679 on Build GRCh38
rs186929344 Chr.14: 73506324-73506324 on Build GRCh38
rs760529326 Chr.14: 73506556-73506556 on Build GRCh38
rs748194777 Chr.14: 73506509-73506509 on Build GRCh38
rs778025966 Chr.14: 73506457-73506458 on Build GRCh38
rs59482129 Chr.14: 73506521-73506521 on Build GRCh38
rs553313019 Chr.14: 73506698-73506698 on Build GRCh38
rs375929384 Chr.14: 73506482-73506482 on Build GRCh38
rs1042610492 Chr.14: 73506543-73506543 on Build GRCh38
rs372430555 Chr.14: 73506486-73506486 on Build GRCh38
rs143649458 Chr.14: 73506550-73506550 on Build GRCh38
rs144130677 Chr.14: 73506643-73506643 on Build GRCh38
rs777001024 Chr.14: 73506470-73506470 on Build GRCh38
rs12894425 Chr.14: 73506526-73506526 on Build GRCh38
rs747473183 Chr.14: 73506515-73506515 on Build GRCh38
rs774047581 Chr.14: 73506546-73506546 on Build GRCh38
rs1050476440 Chr.14: 73506379-73506379 on Build GRCh38
rs778798021 Chr.14: 73506593-73506593 on Build GRCh38
rs146324119 Chr.14: 73506481-73506481 on Build GRCh38
rs150150454 Chr.14: 73506465-73506465 on Build GRCh38
rs12894400 Chr.14: 73506474-73506474 on Build GRCh38
rs199925819 Chr.14: 73506684-73506685 on Build GRCh38
rs761391001 Chr.14: 73506547-73506547 on Build GRCh38
rs768315194 Chr.14: 73506527-73506527 on Build GRCh38
rs766968201 Chr.14: 73506469-73506469 on Build GRCh38
rs200728814 Chr.14: 73506600-73506600 on Build GRCh38
rs749971982 Chr.14: 73506554-73506554 on Build GRCh38
rs550514633 Chr.14: 73506377-73506377 on Build GRCh38

View Full Product Details