Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2671969-2672504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM193A
Entrez Gene ID
8603
Gene Name
family with sequence similarity 193 member A
Gene Aliases
C4orf8, RES4-22
UniGene
Hs.652364
Gene Chromosome Location
Chr.4: 2596288-2732575 on Build GRCh38
Transcripts
NM_001256666.1 NM_001256667.1 NM_001256668.1 NM_003704.3 NR_046335.1 NR_046336.1 XM_006713930.2 XM_006713932.2 XM_011513590.1 XM_011513593.2 XM_017008788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659166 c.1362delC p.H454fs*75 Chr.4: 2672276-2672276 on Build GRCh38
1429343 c.1406G>A p.R469H Chr.4: 2672320-2672320 on Build GRCh38
6659165 c.1362delC p.H454fs*75 Chr.4: 2672276-2672276 on Build GRCh38
7101503 c.1210G>A p.E404K Chr.4: 2672124-2672124 on Build GRCh38
1429344 c.1421C>A p.P474H Chr.4: 2672335-2672335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780698106 Chr.4: 2672334-2672334 on Build GRCh38
rs554143533 Chr.4: 2672452-2672452 on Build GRCh38
rs753925462 Chr.4: 2672219-2672219 on Build GRCh38
rs376734061 Chr.4: 2672114-2672114 on Build GRCh38
rs373468021 Chr.4: 2672098-2672098 on Build GRCh38
rs750483565 Chr.4: 2672411-2672411 on Build GRCh38
rs146511820 Chr.4: 2672236-2672236 on Build GRCh38
rs142820570 Chr.4: 2672201-2672201 on Build GRCh38
rs761056396 Chr.4: 2672367-2672367 on Build GRCh38
rs376862761 Chr.4: 2672404-2672404 on Build GRCh38
rs757450850 Chr.4: 2672319-2672319 on Build GRCh38
rs960016867 Chr.4: 2672082-2672082 on Build GRCh38
rs760330626 Chr.4: 2672158-2672158 on Build GRCh38
rs775337271 Chr.4: 2672120-2672120 on Build GRCh38
rs759965495 Chr.4: 2672192-2672192 on Build GRCh38
rs1025309865 Chr.4: 2672115-2672115 on Build GRCh38
rs964874774 Chr.4: 2672181-2672181 on Build GRCh38
rs994202604 Chr.4: 2672101-2672101 on Build GRCh38
rs952413929 Chr.4: 2672126-2672126 on Build GRCh38
rs141153097 Chr.4: 2672329-2672329 on Build GRCh38
rs145745319 Chr.4: 2672185-2672185 on Build GRCh38
rs781012887 Chr.4: 2672342-2672342 on Build GRCh38
rs746787056 Chr.4: 2672340-2672340 on Build GRCh38
rs144767549 Chr.4: 2672341-2672341 on Build GRCh38
rs753074424 Chr.4: 2672197-2672197 on Build GRCh38
rs149778748 Chr.4: 2672002-2672002 on Build GRCh38
rs779783176 Chr.4: 2672252-2672252 on Build GRCh38
rs764302543 Chr.4: 2672316-2672316 on Build GRCh38
rs945930062 Chr.4: 2672159-2672159 on Build GRCh38
rs772755858 Chr.4: 2672304-2672304 on Build GRCh38
rs760289138 Chr.4: 2672144-2672144 on Build GRCh38
rs765175538 Chr.4: 2672369-2672369 on Build GRCh38
rs187661037 Chr.4: 2672356-2672356 on Build GRCh38
rs369298637 Chr.4: 2672351-2672351 on Build GRCh38
rs947420068 Chr.4: 2672220-2672220 on Build GRCh38
rs1015971981 Chr.4: 2672441-2672442 on Build GRCh38
rs760000921 Chr.4: 2672307-2672307 on Build GRCh38
rs769540793 Chr.4: 2672354-2672354 on Build GRCh38
rs887388518 Chr.4: 2672364-2672364 on Build GRCh38
rs779984639 Chr.4: 2672335-2672335 on Build GRCh38
rs994113942 Chr.4: 2672327-2672327 on Build GRCh38
rs746806828 Chr.4: 2672271-2672271 on Build GRCh38
rs760968624 Chr.4: 2672313-2672313 on Build GRCh38
rs201152034 Chr.4: 2672311-2672311 on Build GRCh38
rs568749555 Chr.4: 2672070-2672070 on Build GRCh38
rs1053506799 Chr.4: 2672196-2672196 on Build GRCh38
rs758438333 Chr.4: 2672424-2672424 on Build GRCh38
rs772020128 Chr.4: 2672248-2672248 on Build GRCh38
rs747850129 Chr.4: 2672350-2672350 on Build GRCh38
rs974973924 Chr.4: 2672182-2672182 on Build GRCh38
rs767821001 Chr.4: 2672193-2672193 on Build GRCh38
rs756463163 Chr.4: 2672062-2672063 on Build GRCh38
rs963798798 Chr.4: 2672438-2672438 on Build GRCh38
rs765218172 Chr.4: 2672100-2672101 on Build GRCh38
rs768060223 Chr.4: 2672309-2672309 on Build GRCh38
rs372731013 Chr.4: 2672368-2672368 on Build GRCh38
rs913248493 Chr.4: 2672207-2672207 on Build GRCh38
rs776338506 Chr.4: 2672153-2672153 on Build GRCh38
rs777954987 Chr.4: 2672202-2672202 on Build GRCh38
rs913132426 Chr.4: 2672136-2672136 on Build GRCh38
rs905996022 Chr.4: 2672267-2672267 on Build GRCh38
rs751970054 Chr.4: 2672186-2672186 on Build GRCh38
rs769377626 Chr.4: 2672301-2672301 on Build GRCh38
rs776951724 Chr.4: 2672399-2672399 on Build GRCh38
rs768419448 Chr.4: 2672148-2672148 on Build GRCh38
rs17164082 Chr.4: 2672165-2672165 on Build GRCh38
rs765026092 Chr.4: 2672088-2672088 on Build GRCh38
rs747531860 Chr.4: 2672355-2672355 on Build GRCh38
rs201813795 Chr.4: 2672069-2672069 on Build GRCh38
rs573989247 Chr.4: 2672016-2672016 on Build GRCh38
rs754014117 Chr.4: 2672318-2672318 on Build GRCh38
rs1005262776 Chr.4: 2672422-2672422 on Build GRCh38
rs980257383 Chr.4: 2672064-2672064 on Build GRCh38
rs374910870 Chr.4: 2672322-2672322 on Build GRCh38
rs763214074 Chr.4: 2672100-2672100 on Build GRCh38
rs199576427 Chr.4: 2672109-2672109 on Build GRCh38
rs745845222 Chr.4: 2672237-2672237 on Build GRCh38
rs367800296 Chr.4: 2672073-2672073 on Build GRCh38
rs749360825 Chr.4: 2672075-2672075 on Build GRCh38
rs983102271 Chr.4: 2671991-2671991 on Build GRCh38
rs1001346598 Chr.4: 2671999-2671999 on Build GRCh38
rs750584757 Chr.4: 2672101-2672101 on Build GRCh38
rs1056971830 Chr.4: 2672291-2672291 on Build GRCh38
rs757257266 Chr.4: 2672227-2672227 on Build GRCh38
rs778761068 Chr.4: 2672095-2672095 on Build GRCh38
rs762093236 Chr.4: 2672401-2672401 on Build GRCh38
rs1015868264 Chr.4: 2671995-2671995 on Build GRCh38
rs927151696 Chr.4: 2672096-2672096 on Build GRCh38
rs761438497 Chr.4: 2672288-2672288 on Build GRCh38
rs942800314 Chr.4: 2672218-2672218 on Build GRCh38
rs776357650 Chr.4: 2672285-2672285 on Build GRCh38
rs775995497 Chr.4: 2672362-2672362 on Build GRCh38
rs768437554 Chr.4: 2672282-2672282 on Build GRCh38
rs372896676 Chr.4: 2672320-2672320 on Build GRCh38

View Full Product Details