Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 81749502-81750047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46A
Entrez Gene ID
55603
Gene Name
family with sequence similarity 46 member A
Gene Aliases
C6orf37, XTP11
UniGene
Hs.10784
Gene Chromosome Location
Chr.6: 81745730-81752711 on Build GRCh38
Transcripts
NM_017633.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
197190 c.1287G>A p.T429T Chr.6: 81749737-81749737 on Build GRCh38
6175199 c.1115A>T p.H372L Chr.6: 81749909-81749909 on Build GRCh38
3176264 c.1155T>A p.A385A Chr.6: 81749869-81749869 on Build GRCh38
4868556 c.1502A>G p.Y501C Chr.6: 81749765-81749765 on Build GRCh38
4812538 c.1414G>A p.D472N Chr.6: 81749853-81749853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112538963 Chr.6: 81749605-81749605 on Build GRCh38
rs746161231 Chr.6: 81749934-81749934 on Build GRCh38
rs760735219 Chr.6: 81749835-81749835 on Build GRCh38
rs145308758 Chr.6: 81749740-81749740 on Build GRCh38
rs774536189 Chr.6: 81749812-81749812 on Build GRCh38
rs376895458 Chr.6: 81749794-81749794 on Build GRCh38
rs747593288 Chr.6: 81749769-81749769 on Build GRCh38
rs758339269 Chr.6: 81749651-81749651 on Build GRCh38
rs770655416 Chr.6: 81749928-81749928 on Build GRCh38
rs759649898 Chr.6: 81749717-81749717 on Build GRCh38
rs780521689 Chr.6: 81749965-81749965 on Build GRCh38
rs148802326 Chr.6: 81749788-81749788 on Build GRCh38
rs773616942 Chr.6: 81749731-81749731 on Build GRCh38
rs774897515 Chr.6: 81749711-81749711 on Build GRCh38
rs879010540 Chr.6: 81749628-81749628 on Build GRCh38
rs1030913001 Chr.6: 81749545-81749545 on Build GRCh38
rs376631603 Chr.6: 81749732-81749732 on Build GRCh38
rs767612943 Chr.6: 81749719-81749719 on Build GRCh38
rs548850600 Chr.6: 81749763-81749763 on Build GRCh38
rs749658715 Chr.6: 81749699-81749699 on Build GRCh38
rs1040650566 Chr.6: 81749557-81749557 on Build GRCh38
rs773451915 Chr.6: 81749697-81749697 on Build GRCh38
rs1020177515 Chr.6: 81749585-81749585 on Build GRCh38
rs906390149 Chr.6: 81749826-81749826 on Build GRCh38
rs747825481 Chr.6: 81749681-81749681 on Build GRCh38
rs780029980 Chr.6: 81749669-81749669 on Build GRCh38
rs376057313 Chr.6: 81749628-81749628 on Build GRCh38
rs199706042 Chr.6: 81749617-81749617 on Build GRCh38
rs779345430 Chr.6: 81749930-81749930 on Build GRCh38
rs560203341 Chr.6: 81749625-81749625 on Build GRCh38
rs771292824 Chr.6: 81749800-81749800 on Build GRCh38
rs1046610 Chr.6: 81749525-81749525 on Build GRCh38
rs903987448 Chr.6: 81749623-81749623 on Build GRCh38
rs546736217 Chr.6: 81749599-81749599 on Build GRCh38
rs916676917 Chr.6: 81749747-81749747 on Build GRCh38
rs965670205 Chr.6: 81749601-81749601 on Build GRCh38
rs200190773 Chr.6: 81749860-81749860 on Build GRCh38
rs766270220 Chr.6: 81749758-81749758 on Build GRCh38
rs957716501 Chr.6: 81749548-81749548 on Build GRCh38
rs751342220 Chr.6: 81749868-81749868 on Build GRCh38
rs1010575659 Chr.6: 81749549-81749549 on Build GRCh38
rs762108942 Chr.6: 81750006-81750006 on Build GRCh38
rs1024914418 Chr.6: 81749757-81749757 on Build GRCh38
rs552106518 Chr.6: 81749641-81749641 on Build GRCh38
rs776883813 Chr.6: 81749855-81749855 on Build GRCh38
rs1026328504 Chr.6: 81749614-81749614 on Build GRCh38
rs1020988328 Chr.6: 81749616-81749616 on Build GRCh38
rs534392403 Chr.6: 81749821-81749821 on Build GRCh38
rs575374370 Chr.6: 81749572-81749572 on Build GRCh38
rs754508310 Chr.6: 81749674-81749674 on Build GRCh38
rs541226280 Chr.6: 81749985-81749985 on Build GRCh38
rs750169192 Chr.6: 81749650-81749650 on Build GRCh38
rs544660565 Chr.6: 81749714-81749714 on Build GRCh38
rs755897612 Chr.6: 81749782-81749782 on Build GRCh38
rs777329980 Chr.6: 81749896-81749896 on Build GRCh38
rs935135508 Chr.6: 81749828-81749828 on Build GRCh38
rs760896955 Chr.6: 81749737-81749737 on Build GRCh38
rs1009721003 Chr.6: 81749597-81749597 on Build GRCh38
rs375136104 Chr.6: 81749675-81749675 on Build GRCh38
rs765035543 Chr.6: 81749648-81749648 on Build GRCh38
rs879128167 Chr.6: 81749618-81749618 on Build GRCh38
rs772506719 Chr.6: 81749823-81749823 on Build GRCh38
rs771398269 Chr.6: 81749705-81749705 on Build GRCh38
rs902003557 Chr.6: 81749571-81749571 on Build GRCh38
rs767558102 Chr.6: 81749877-81749877 on Build GRCh38
rs768814618 Chr.6: 81749976-81749976 on Build GRCh38
rs772629416 Chr.6: 81749959-81749959 on Build GRCh38
rs769514314 Chr.6: 81749692-81749692 on Build GRCh38
rs1028777015 Chr.6: 81749569-81749569 on Build GRCh38
rs972988760 Chr.6: 81749616-81749626 on Build GRCh38
rs553952728 Chr.6: 81749833-81749833 on Build GRCh38
rs985649196 Chr.6: 81749575-81749575 on Build GRCh38
rs769841953 Chr.6: 81750013-81750013 on Build GRCh38
rs746552366 Chr.6: 81749673-81749673 on Build GRCh38
rs750247269 Chr.6: 81749748-81749748 on Build GRCh38
rs768812635 Chr.6: 81749841-81749841 on Build GRCh38
rs868292886 Chr.6: 81749537-81749537 on Build GRCh38
rs532380636 Chr.6: 81749635-81749635 on Build GRCh38
rs747135448 Chr.6: 81749975-81749975 on Build GRCh38
rs774354455 Chr.6: 81750019-81750019 on Build GRCh38
rs758192773 Chr.6: 81749749-81749749 on Build GRCh38
rs952117888 Chr.6: 81749643-81749643 on Build GRCh38
rs371926731 Chr.6: 81749765-81749765 on Build GRCh38
rs138858550 Chr.6: 81749887-81749887 on Build GRCh38
rs761901784 Chr.6: 81749857-81749857 on Build GRCh38
rs753555884 Chr.6: 81749900-81749900 on Build GRCh38
rs142530175 Chr.6: 81749761-81749761 on Build GRCh38
rs759552344 Chr.6: 81749874-81749874 on Build GRCh38
rs373242187 Chr.6: 81749986-81749986 on Build GRCh38
rs943590979 Chr.6: 81749527-81749527 on Build GRCh38
rs545732284 Chr.6: 81749629-81749629 on Build GRCh38
rs953215653 Chr.6: 81749574-81749574 on Build GRCh38

View Full Product Details