Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 97858444-97858987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
UniGene
Hs.489207
Gene Chromosome Location
Chr.7: 97852117-97872542 on Build GRCh38
Transcripts
NM_001178075.1 NM_001178076.1 NM_001178077.1 NM_001673.4 NM_133436.3 NM_183356.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5358509 c.761G>A p.G254D Chr.7: 97858868-97858868 on Build GRCh38
3642633 c.703C>T p.L235F Chr.7: 97858926-97858926 on Build GRCh38
1578368 c.718_719delAA p.N240fs*1 Chr.7: 97858910-97858911 on Build GRCh38
5158272 c.674-9T>C p.? Chr.7: 97858964-97858964 on Build GRCh38
3642634 c.703C>T p.L235F Chr.7: 97858926-97858926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764380360 Chr.7: 97858883-97858883 on Build GRCh38
rs181921458 Chr.7: 97858682-97858682 on Build GRCh38
rs767849373 Chr.7: 97858824-97858824 on Build GRCh38
rs756536266 Chr.7: 97858520-97858520 on Build GRCh38
rs11976728 Chr.7: 97858519-97858519 on Build GRCh38
rs958872669 Chr.7: 97858806-97858806 on Build GRCh38
rs757830451 Chr.7: 97858941-97858941 on Build GRCh38
rs754378047 Chr.7: 97858887-97858887 on Build GRCh38
rs1050744833 Chr.7: 97858544-97858544 on Build GRCh38
rs370169709 Chr.7: 97858641-97858641 on Build GRCh38
rs1008699526 Chr.7: 97858579-97858579 on Build GRCh38
rs537943182 Chr.7: 97858963-97858963 on Build GRCh38
rs765369330 Chr.7: 97858842-97858842 on Build GRCh38
rs763900453 Chr.7: 97858564-97858564 on Build GRCh38
rs779252740 Chr.7: 97858898-97858898 on Build GRCh38
rs377223004 Chr.7: 97858774-97858774 on Build GRCh38
rs538866229 Chr.7: 97858819-97858819 on Build GRCh38
rs74707892 Chr.7: 97858834-97858834 on Build GRCh38
rs148111963 Chr.7: 97858901-97858901 on Build GRCh38
rs773724311 Chr.7: 97858834-97858836 on Build GRCh38
rs761044095 Chr.7: 97858871-97858871 on Build GRCh38
rs397734729 Chr.7: 97858841-97858842 on Build GRCh38
rs36021744 Chr.7: 97858834-97858835 on Build GRCh38
rs377402852 Chr.7: 97858491-97858491 on Build GRCh38
rs1020272918 Chr.7: 97858782-97858782 on Build GRCh38
rs779863917 Chr.7: 97858959-97858959 on Build GRCh38
rs866610593 Chr.7: 97858499-97858499 on Build GRCh38
rs566786916 Chr.7: 97858793-97858793 on Build GRCh38
rs774539538 Chr.7: 97858809-97858809 on Build GRCh38
rs185314350 Chr.7: 97858786-97858786 on Build GRCh38
rs1010277519 Chr.7: 97858714-97858714 on Build GRCh38
rs757828751 Chr.7: 97858893-97858893 on Build GRCh38
rs767368474 Chr.7: 97858937-97858938 on Build GRCh38
rs770938720 Chr.7: 97858808-97858808 on Build GRCh38
rs552328986 Chr.7: 97858841-97858841 on Build GRCh38
rs562593949 Chr.7: 97858571-97858571 on Build GRCh38
rs776550941 Chr.7: 97858835-97858835 on Build GRCh38
rs752897486 Chr.7: 97858874-97858876 on Build GRCh38
rs927508844 Chr.7: 97858798-97858798 on Build GRCh38
rs762661692 Chr.7: 97858533-97858533 on Build GRCh38
rs746591365 Chr.7: 97858961-97858961 on Build GRCh38
rs1019715637 Chr.7: 97858656-97858656 on Build GRCh38
rs530474003 Chr.7: 97858767-97858767 on Build GRCh38
rs757128920 Chr.7: 97858897-97858897 on Build GRCh38
rs7792744 Chr.7: 97858659-97858659 on Build GRCh38
rs755408859 Chr.7: 97858812-97858813 on Build GRCh38
rs979055818 Chr.7: 97858787-97858787 on Build GRCh38

View Full Product Details