Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 16992543-16993139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF20
Entrez Gene ID
26281
Gene Name
fibroblast growth factor 20
Gene Aliases
FGF-20, RHDA2
UniGene
Hs.199905
Gene Chromosome Location
Chr.8: 16992825-17002165 on Build GRCh38
Transcripts
NM_019851.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874277 c.590C>T p.P197L Chr.8: 16993118-16993118 on Build GRCh38
486239 c.609G>A p.L203L Chr.8: 16993099-16993099 on Build GRCh38
3834343 c.586G>A p.D196N Chr.8: 16993122-16993122 on Build GRCh38
5874278 c.575C>T p.P192L Chr.8: 16993133-16993133 on Build GRCh38
4587943 c.630C>T p.Y210Y Chr.8: 16993078-16993078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113374862 Chr.8: 16992918-16992918 on Build GRCh38
rs116309944 Chr.8: 16992918-16992918 on Build GRCh38
rs1024432862 Chr.8: 16992582-16992582 on Build GRCh38
rs1005934099 Chr.8: 16992696-16992696 on Build GRCh38
rs774693591 Chr.8: 16993062-16993062 on Build GRCh38
rs77259875 Chr.8: 16992750-16992750 on Build GRCh38
rs772551583 Chr.8: 16992660-16992660 on Build GRCh38
rs999003744 Chr.8: 16992787-16992787 on Build GRCh38
rs1018452811 Chr.8: 16992605-16992605 on Build GRCh38
rs369979222 Chr.8: 16993064-16993064 on Build GRCh38
rs572374220 Chr.8: 16992926-16992926 on Build GRCh38
rs1040204264 Chr.8: 16993108-16993108 on Build GRCh38
rs1041831813 Chr.8: 16992785-16992785 on Build GRCh38
rs1721100 Chr.8: 16992989-16992989 on Build GRCh38
rs575888899 Chr.8: 16992662-16992662 on Build GRCh38
rs780961429 Chr.8: 16993080-16993080 on Build GRCh38
rs1017087794 Chr.8: 16993035-16993035 on Build GRCh38
rs746294164 Chr.8: 16992640-16992640 on Build GRCh38
rs569516194 Chr.8: 16992959-16992959 on Build GRCh38
rs1045775155 Chr.8: 16992946-16992946 on Build GRCh38
rs779217367 Chr.8: 16992638-16992638 on Build GRCh38
rs769791216 Chr.8: 16993051-16993051 on Build GRCh38
rs975825506 Chr.8: 16992882-16992882 on Build GRCh38
rs751701947 Chr.8: 16992849-16992849 on Build GRCh38
rs780521897 Chr.8: 16993069-16993069 on Build GRCh38
rs372796901 Chr.8: 16993098-16993098 on Build GRCh38
rs182573765 Chr.8: 16992976-16992976 on Build GRCh38
rs951641387 Chr.8: 16992885-16992885 on Build GRCh38
rs984994210 Chr.8: 16993042-16993042 on Build GRCh38
rs774471972 Chr.8: 16993062-16993062 on Build GRCh38
rs754685284 Chr.8: 16993078-16993078 on Build GRCh38
rs1024379139 Chr.8: 16992777-16992777 on Build GRCh38
rs564045462 Chr.8: 16992911-16992911 on Build GRCh38
rs375846209 Chr.8: 16993076-16993076 on Build GRCh38
rs1029805434 Chr.8: 16992858-16992858 on Build GRCh38
rs538577421 Chr.8: 16992665-16992665 on Build GRCh38
rs147590629 Chr.8: 16992917-16992917 on Build GRCh38
rs747943357 Chr.8: 16993083-16993083 on Build GRCh38
rs777595585 Chr.8: 16993084-16993084 on Build GRCh38
rs17550360 Chr.8: 16993092-16993092 on Build GRCh38
rs1020404581 Chr.8: 16992577-16992577 on Build GRCh38
rs574070314 Chr.8: 16992887-16992887 on Build GRCh38
rs750613452 Chr.8: 16992603-16992604 on Build GRCh38
rs769148271 Chr.8: 16992886-16992886 on Build GRCh38
rs1014302079 Chr.8: 16992755-16992755 on Build GRCh38
rs764711524 Chr.8: 16993042-16993043 on Build GRCh38
rs1037370828 Chr.8: 16992710-16992710 on Build GRCh38
rs112602726 Chr.8: 16992952-16992952 on Build GRCh38
rs530611016 Chr.8: 16992903-16992903 on Build GRCh38
rs566436899 Chr.8: 16992922-16992922 on Build GRCh38
rs773213151 Chr.8: 16993055-16993055 on Build GRCh38
rs17515351 Chr.8: 16992829-16992829 on Build GRCh38
rs768410520 Chr.8: 16993034-16993034 on Build GRCh38
rs927226719 Chr.8: 16992727-16992727 on Build GRCh38
rs887727487 Chr.8: 16992687-16992687 on Build GRCh38
rs977040898 Chr.8: 16992626-16992626 on Build GRCh38
rs192962979 Chr.8: 16992661-16992661 on Build GRCh38
rs758002395 Chr.8: 16993022-16993022 on Build GRCh38
rs998865931 Chr.8: 16992850-16992850 on Build GRCh38
rs1000193361 Chr.8: 16992825-16992825 on Build GRCh38
rs904484059 Chr.8: 16992804-16992804 on Build GRCh38
rs865776582 Chr.8: 16993075-16993075 on Build GRCh38
rs113126203 Chr.8: 16993038-16993038 on Build GRCh38
rs967570638 Chr.8: 16992595-16992595 on Build GRCh38
rs771137602 Chr.8: 16993059-16993059 on Build GRCh38
rs900067949 Chr.8: 16992845-16992845 on Build GRCh38
rs946124140 Chr.8: 16992781-16992781 on Build GRCh38
rs746261060 Chr.8: 16992908-16992908 on Build GRCh38
rs373460171 Chr.8: 16992931-16992931 on Build GRCh38
rs142106513 Chr.8: 16992924-16992924 on Build GRCh38
rs573963889 Chr.8: 16993077-16993077 on Build GRCh38
rs975773082 Chr.8: 16993020-16993020 on Build GRCh38
rs369523536 Chr.8: 16993025-16993025 on Build GRCh38
rs937947933 Chr.8: 16992918-16992919 on Build GRCh38
rs1033200361 Chr.8: 16992975-16992975 on Build GRCh38
rs575400716 Chr.8: 16993057-16993057 on Build GRCh38
rs774535729 Chr.8: 16993095-16993095 on Build GRCh38
rs937285731 Chr.8: 16992735-16992735 on Build GRCh38
rs749890928 Chr.8: 16993070-16993070 on Build GRCh38
rs539061752 Chr.8: 16992598-16992598 on Build GRCh38
rs1049431393 Chr.8: 16992678-16992678 on Build GRCh38
rs12720208 Chr.8: 16992890-16992890 on Build GRCh38
rs2640901 Chr.8: 16992907-16992907 on Build GRCh38
rs895657448 Chr.8: 16992752-16992752 on Build GRCh38
rs746061786 Chr.8: 16993096-16993096 on Build GRCh38
rs746767850 Chr.8: 16993030-16993030 on Build GRCh38
rs965682514 Chr.8: 16993019-16993019 on Build GRCh38
rs10089759 Chr.8: 16992647-16992647 on Build GRCh38
rs779979768 Chr.8: 16992906-16992907 on Build GRCh38
rs377266598 Chr.8: 16993046-16993046 on Build GRCh38
rs957243047 Chr.8: 16992604-16992604 on Build GRCh38
rs6982917 Chr.8: 16992961-16992961 on Build GRCh38
rs936045906 Chr.8: 16993107-16993107 on Build GRCh38
rs1000399884 Chr.8: 16992602-16992602 on Build GRCh38
rs560503402 Chr.8: 16992854-16992854 on Build GRCh38
rs535500971 Chr.8: 16992656-16992656 on Build GRCh38
rs922818507 Chr.8: 16992633-16992633 on Build GRCh38
rs534476986 Chr.8: 16992986-16992986 on Build GRCh38
rs965734689 Chr.8: 16992873-16992873 on Build GRCh38
rs578238179 Chr.8: 16992806-16992806 on Build GRCh38
rs1004331933 Chr.8: 16993024-16993024 on Build GRCh38
rs767969554 Chr.8: 16993066-16993066 on Build GRCh38
rs1721082 Chr.8: 16992623-16992623 on Build GRCh38
rs904598532 Chr.8: 16992925-16992925 on Build GRCh38
rs533487160 Chr.8: 16992889-16992889 on Build GRCh38
rs372623598 Chr.8: 16993067-16993067 on Build GRCh38
rs140595599 Chr.8: 16992984-16992984 on Build GRCh38
rs998417008 Chr.8: 16992977-16992977 on Build GRCh38

View Full Product Details