Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 126554347-126554934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM84B
Entrez Gene ID
157638
Gene Name
family with sequence similarity 84 member B
Gene Aliases
BCMP101, NSE2
UniGene
Hs.741352
Gene Chromosome Location
Chr.8: 126552438-126558499 on Build GRCh38
Transcripts
NM_174911.4 XM_017013107.1 XM_017013108.1

SNP Detail

SNP ID Chromosome Location
rs77693027 Chr.8: 126554418-126554418 on Build GRCh38
rs943986939 Chr.8: 126554706-126554706 on Build GRCh38
rs1055787766 Chr.8: 126554527-126554527 on Build GRCh38
rs929340960 Chr.8: 126554746-126554746 on Build GRCh38
rs189055983 Chr.8: 126554456-126554456 on Build GRCh38
rs575822932 Chr.8: 126554482-126554482 on Build GRCh38
rs927947640 Chr.8: 126554876-126554876 on Build GRCh38
rs985314961 Chr.8: 126554672-126554672 on Build GRCh38
rs747066732 Chr.8: 126554671-126554671 on Build GRCh38
rs149700734 Chr.8: 126554738-126554738 on Build GRCh38
rs1029362170 Chr.8: 126554692-126554692 on Build GRCh38
rs781501444 Chr.8: 126554765-126554765 on Build GRCh38
rs78338944 Chr.8: 126554445-126554445 on Build GRCh38
rs1047717235 Chr.8: 126554747-126554747 on Build GRCh38
rs73705689 Chr.8: 126554689-126554689 on Build GRCh38
rs772978722 Chr.8: 126554792-126554793 on Build GRCh38
rs1055353255 Chr.8: 126554887-126554887 on Build GRCh38
rs577841840 Chr.8: 126554386-126554386 on Build GRCh38
rs1005735235 Chr.8: 126554592-126554592 on Build GRCh38
rs939366625 Chr.8: 126554877-126554877 on Build GRCh38
rs549038110 Chr.8: 126554886-126554886 on Build GRCh38
rs74519505 Chr.8: 126554446-126554446 on Build GRCh38
rs191946946 Chr.8: 126554587-126554587 on Build GRCh38
rs1003130649 Chr.8: 126554862-126554862 on Build GRCh38
rs796501195 Chr.8: 126554566-126554566 on Build GRCh38
rs147791258 Chr.8: 126554859-126554859 on Build GRCh38
rs780050667 Chr.8: 126554571-126554571 on Build GRCh38
rs906004702 Chr.8: 126554846-126554846 on Build GRCh38
rs562292183 Chr.8: 126554465-126554465 on Build GRCh38
rs945520647 Chr.8: 126554856-126554856 on Build GRCh38
rs563903300 Chr.8: 126554633-126554633 on Build GRCh38
rs571511289 Chr.8: 126554804-126554804 on Build GRCh38
rs773137803 Chr.8: 126554902-126554902 on Build GRCh38
rs933960659 Chr.8: 126554411-126554411 on Build GRCh38
rs989971836 Chr.8: 126554810-126554810 on Build GRCh38
rs982074240 Chr.8: 126554582-126554582 on Build GRCh38
rs943935204 Chr.8: 126554423-126554423 on Build GRCh38
rs889749263 Chr.8: 126554420-126554420 on Build GRCh38
rs1042415824 Chr.8: 126554483-126554483 on Build GRCh38
rs1003833608 Chr.8: 126554519-126554519 on Build GRCh38
rs1006382738 Chr.8: 126554554-126554554 on Build GRCh38
rs528251767 Chr.8: 126554399-126554399 on Build GRCh38
rs111885486 Chr.8: 126554653-126554653 on Build GRCh38
rs887853792 Chr.8: 126554534-126554534 on Build GRCh38
rs945367822 Chr.8: 126554479-126554479 on Build GRCh38
rs796081134 Chr.8: 126554855-126554855 on Build GRCh38
rs58174190 Chr.8: 126554843-126554843 on Build GRCh38
rs1026321485 Chr.8: 126554508-126554508 on Build GRCh38
rs546590694 Chr.8: 126554701-126554701 on Build GRCh38
rs947170410 Chr.8: 126554403-126554403 on Build GRCh38
rs911033431 Chr.8: 126554697-126554697 on Build GRCh38
rs1044584153 Chr.8: 126554812-126554812 on Build GRCh38
rs1041320280 Chr.8: 126554732-126554732 on Build GRCh38
rs988569136 Chr.8: 126554611-126554611 on Build GRCh38
rs906776412 Chr.8: 126554518-126554518 on Build GRCh38
rs955860129 Chr.8: 126554601-126554601 on Build GRCh38
rs935841212 Chr.8: 126554440-126554440 on Build GRCh38

View Full Product Details