Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34159764-34160347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
UniGene
Hs.23361
Gene Chromosome Location
Chr.11: 34150987-34358008 on Build GRCh38
Transcripts
NM_145804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
141336 c.1921C>T p.R641W Chr.11: 34160272-34160272 on Build GRCh38
3446949 c.2000G>A p.G667E Chr.11: 34159954-34159954 on Build GRCh38
6591340 c.2572G>A p.A858T Chr.11: 34159940-34159940 on Build GRCh38
3446951 c.1966A>G p.K656E Chr.11: 34159988-34159988 on Build GRCh38
6591341 c.2014G>A p.A672T Chr.11: 34159940-34159940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371663376 Chr.11: 34160037-34160037 on Build GRCh38
rs550366636 Chr.11: 34160238-34160238 on Build GRCh38
rs754138701 Chr.11: 34160038-34160038 on Build GRCh38
rs754189449 Chr.11: 34159881-34159881 on Build GRCh38
rs200142986 Chr.11: 34159991-34159991 on Build GRCh38
rs778485371 Chr.11: 34160170-34160172 on Build GRCh38
rs891846754 Chr.11: 34160148-34160148 on Build GRCh38
rs745772913 Chr.11: 34160287-34160287 on Build GRCh38
rs542427404 Chr.11: 34159853-34159853 on Build GRCh38
rs752771357 Chr.11: 34159876-34159876 on Build GRCh38
rs752037682 Chr.11: 34160249-34160249 on Build GRCh38
rs757660076 Chr.11: 34160046-34160046 on Build GRCh38
rs753341667 Chr.11: 34160029-34160029 on Build GRCh38
rs766286581 Chr.11: 34160026-34160026 on Build GRCh38
rs758937099 Chr.11: 34160055-34160055 on Build GRCh38
rs766855827 Chr.11: 34160242-34160242 on Build GRCh38
rs868740404 Chr.11: 34159823-34159823 on Build GRCh38
rs374658635 Chr.11: 34159880-34159880 on Build GRCh38
rs764305150 Chr.11: 34160229-34160229 on Build GRCh38
rs776895074 Chr.11: 34160236-34160236 on Build GRCh38
rs750809554 Chr.11: 34160054-34160054 on Build GRCh38
rs143738329 Chr.11: 34159858-34159859 on Build GRCh38
rs527301061 Chr.11: 34160149-34160149 on Build GRCh38
rs761985842 Chr.11: 34160310-34160310 on Build GRCh38
rs561183589 Chr.11: 34159841-34159841 on Build GRCh38
rs763188531 Chr.11: 34160028-34160029 on Build GRCh38
rs779517626 Chr.11: 34159886-34159886 on Build GRCh38
rs780426738 Chr.11: 34160060-34160060 on Build GRCh38
rs746465398 Chr.11: 34160202-34160202 on Build GRCh38
rs930192475 Chr.11: 34159919-34159919 on Build GRCh38
rs762923164 Chr.11: 34160020-34160020 on Build GRCh38
rs201805676 Chr.11: 34160001-34160001 on Build GRCh38
rs371085454 Chr.11: 34160240-34160240 on Build GRCh38
rs376052637 Chr.11: 34160228-34160228 on Build GRCh38
rs1024284452 Chr.11: 34160223-34160223 on Build GRCh38
rs770654532 Chr.11: 34160271-34160271 on Build GRCh38
rs750378604 Chr.11: 34159863-34159863 on Build GRCh38
rs776405623 Chr.11: 34159856-34159856 on Build GRCh38
rs777297363 Chr.11: 34160265-34160265 on Build GRCh38
rs973166473 Chr.11: 34159869-34159869 on Build GRCh38
rs79718145 Chr.11: 34159836-34159836 on Build GRCh38
rs937611866 Chr.11: 34159934-34159934 on Build GRCh38
rs150092705 Chr.11: 34159818-34159818 on Build GRCh38
rs761600185 Chr.11: 34159858-34159858 on Build GRCh38
rs1000266013 Chr.11: 34159784-34159784 on Build GRCh38
rs267602866 Chr.11: 34159912-34159912 on Build GRCh38
rs116444349 Chr.11: 34160306-34160306 on Build GRCh38
rs1033123152 Chr.11: 34159794-34159794 on Build GRCh38
rs765493571 Chr.11: 34160321-34160321 on Build GRCh38
rs1053621775 Chr.11: 34160147-34160147 on Build GRCh38
rs746564433 Chr.11: 34159987-34159987 on Build GRCh38
rs984227996 Chr.11: 34160234-34160234 on Build GRCh38
rs759490402 Chr.11: 34159868-34159868 on Build GRCh38
rs139787884 Chr.11: 34160258-34160258 on Build GRCh38
rs780656840 Chr.11: 34160259-34160259 on Build GRCh38
rs929057639 Chr.11: 34159887-34159887 on Build GRCh38
rs746420534 Chr.11: 34159902-34159902 on Build GRCh38
rs984259126 Chr.11: 34159923-34159923 on Build GRCh38
rs927462390 Chr.11: 34159859-34159859 on Build GRCh38
rs78889266 Chr.11: 34159851-34159851 on Build GRCh38
rs562357557 Chr.11: 34159860-34159860 on Build GRCh38
rs902367691 Chr.11: 34159816-34159816 on Build GRCh38
rs999332742 Chr.11: 34160082-34160082 on Build GRCh38
rs779600086 Chr.11: 34159947-34159947 on Build GRCh38
rs564882263 Chr.11: 34160112-34160112 on Build GRCh38
rs778208632 Chr.11: 34159879-34159879 on Build GRCh38
rs570250062 Chr.11: 34160239-34160239 on Build GRCh38
rs775725508 Chr.11: 34159990-34159992 on Build GRCh38
rs767599928 Chr.11: 34159872-34159872 on Build GRCh38
rs1006826947 Chr.11: 34160165-34160165 on Build GRCh38
rs1054542159 Chr.11: 34160120-34160120 on Build GRCh38
rs371347160 Chr.11: 34159863-34159864 on Build GRCh38
rs755761010 Chr.11: 34160263-34160263 on Build GRCh38
rs1013350219 Chr.11: 34159795-34159795 on Build GRCh38
rs769607828 Chr.11: 34160016-34160016 on Build GRCh38
rs749055024 Chr.11: 34160267-34160267 on Build GRCh38
rs770836734 Chr.11: 34160225-34160225 on Build GRCh38
rs772188481 Chr.11: 34160302-34160302 on Build GRCh38
rs371809150 Chr.11: 34160214-34160214 on Build GRCh38
rs757713565 Chr.11: 34159883-34159883 on Build GRCh38
rs756347165 Chr.11: 34159877-34159877 on Build GRCh38
rs777238173 Chr.11: 34159821-34159821 on Build GRCh38
rs956016988 Chr.11: 34159808-34159808 on Build GRCh38
rs752819028 Chr.11: 34160028-34160028 on Build GRCh38
rs1025773755 Chr.11: 34160285-34160285 on Build GRCh38
rs145499198 Chr.11: 34160212-34160212 on Build GRCh38
rs779392238 Chr.11: 34160047-34160047 on Build GRCh38
rs376707571 Chr.11: 34160211-34160211 on Build GRCh38
rs1046186438 Chr.11: 34160025-34160025 on Build GRCh38
rs1018277341 Chr.11: 34160192-34160192 on Build GRCh38
rs761546937 Chr.11: 34160007-34160007 on Build GRCh38
rs764120985 Chr.11: 34160231-34160231 on Build GRCh38
rs911570188 Chr.11: 34159969-34159969 on Build GRCh38
rs747712308 Chr.11: 34160260-34160260 on Build GRCh38
rs773029237 Chr.11: 34160019-34160019 on Build GRCh38
rs918746854 Chr.11: 34159889-34159889 on Build GRCh38
rs536369887 Chr.11: 34160237-34160237 on Build GRCh38
rs965433472 Chr.11: 34160203-34160203 on Build GRCh38
rs547379772 Chr.11: 34160217-34160217 on Build GRCh38
rs145938805 Chr.11: 34159926-34159926 on Build GRCh38
rs770059886 Chr.11: 34159983-34159985 on Build GRCh38
rs559282338 Chr.11: 34160272-34160272 on Build GRCh38

View Full Product Details