Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119097110-119097704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPAGT1
Entrez Gene ID
1798
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
Gene Aliases
ALG7, CDG-Ij, CDG1J, CMS13, CMSTA2, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT
UniGene
Hs.524081
Gene Chromosome Location
Chr.11: 119096503-119102414 on Build GRCh38
Transcripts
NM_001382.3 XM_005271422.2 XM_011542648.1 XM_017017293.1 XM_017017294.1 XM_017017295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7119857 c.930G>A p.K310K Chr.11: 119097539-119097539 on Build GRCh38
5573430 c.685-2A>T p.? Chr.11: 119097299-119097299 on Build GRCh38
6500829 c.697C>T p.L233F Chr.11: 119097285-119097285 on Build GRCh38
7080319 c.984C>T p.F328F Chr.11: 119097485-119097485 on Build GRCh38
5201157 c.798delC p.I267fs*15 Chr.11: 119097184-119097184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780550367 Chr.11: 119097313-119097313 on Build GRCh38
rs1003269168 Chr.11: 119097530-119097530 on Build GRCh38
rs200747570 Chr.11: 119097175-119097175 on Build GRCh38
rs941907857 Chr.11: 119097477-119097477 on Build GRCh38
rs371389351 Chr.11: 119097544-119097544 on Build GRCh38
rs778250734 Chr.11: 119097304-119097304 on Build GRCh38
rs141849365 Chr.11: 119097374-119097374 on Build GRCh38
rs987583485 Chr.11: 119097661-119097661 on Build GRCh38
rs767497123 Chr.11: 119097418-119097418 on Build GRCh38
rs955174507 Chr.11: 119097635-119097635 on Build GRCh38
rs759542218 Chr.11: 119097180-119097180 on Build GRCh38
rs768805817 Chr.11: 119097162-119097162 on Build GRCh38
rs1045143648 Chr.11: 119097440-119097440 on Build GRCh38
rs547224768 Chr.11: 119097626-119097626 on Build GRCh38
rs549465462 Chr.11: 119097566-119097566 on Build GRCh38
rs756851791 Chr.11: 119097453-119097453 on Build GRCh38
rs748295799 Chr.11: 119097332-119097332 on Build GRCh38
rs1029520555 Chr.11: 119097632-119097632 on Build GRCh38
rs370544608 Chr.11: 119097568-119097568 on Build GRCh38
rs763515856 Chr.11: 119097560-119097560 on Build GRCh38
rs543421224 Chr.11: 119097260-119097260 on Build GRCh38
rs757905259 Chr.11: 119097479-119097479 on Build GRCh38
rs764995440 Chr.11: 119097474-119097474 on Build GRCh38
rs917922996 Chr.11: 119097355-119097355 on Build GRCh38
rs145206990 Chr.11: 119097569-119097569 on Build GRCh38
rs760501264 Chr.11: 119097437-119097437 on Build GRCh38
rs776720609 Chr.11: 119097164-119097164 on Build GRCh38
rs770121489 Chr.11: 119097338-119097338 on Build GRCh38
rs775602137 Chr.11: 119097427-119097427 on Build GRCh38
rs531346717 Chr.11: 119097559-119097559 on Build GRCh38
rs747534998 Chr.11: 119097491-119097491 on Build GRCh38
rs555841797 Chr.11: 119097279-119097279 on Build GRCh38
rs781380261 Chr.11: 119097498-119097498 on Build GRCh38
rs191573465 Chr.11: 119097556-119097556 on Build GRCh38
rs755527720 Chr.11: 119097493-119097493 on Build GRCh38
rs752484662 Chr.11: 119097254-119097254 on Build GRCh38
rs139295058 Chr.11: 119097192-119097192 on Build GRCh38
rs751109697 Chr.11: 119097597-119097597 on Build GRCh38
rs376996734 Chr.11: 119097285-119097285 on Build GRCh38
rs147104217 Chr.11: 119097394-119097394 on Build GRCh38
rs763140128 Chr.11: 119097415-119097415 on Build GRCh38
rs754036071 Chr.11: 119097488-119097489 on Build GRCh38
rs750564616 Chr.11: 119097179-119097179 on Build GRCh38
rs780497256 Chr.11: 119097480-119097480 on Build GRCh38
rs1042544648 Chr.11: 119097482-119097482 on Build GRCh38
rs745447691 Chr.11: 119097311-119097311 on Build GRCh38
rs764154648 Chr.11: 119097561-119097561 on Build GRCh38
rs910633422 Chr.11: 119097662-119097662 on Build GRCh38
rs1016844949 Chr.11: 119097149-119097149 on Build GRCh38
rs761911331 Chr.11: 119097169-119097169 on Build GRCh38
rs777530944 Chr.11: 119097277-119097277 on Build GRCh38
rs773152872 Chr.11: 119097340-119097340 on Build GRCh38
rs750104776 Chr.11: 119097575-119097575 on Build GRCh38
rs201656540 Chr.11: 119097555-119097555 on Build GRCh38
rs545736353 Chr.11: 119097526-119097526 on Build GRCh38
rs765851764 Chr.11: 119097589-119097589 on Build GRCh38
rs980077282 Chr.11: 119097204-119097204 on Build GRCh38
rs574176195 Chr.11: 119097323-119097323 on Build GRCh38
rs77836371 Chr.11: 119097300-119097300 on Build GRCh38
rs758885855 Chr.11: 119097312-119097312 on Build GRCh38
rs748605109 Chr.11: 119097523-119097523 on Build GRCh38
rs943371372 Chr.11: 119097339-119097339 on Build GRCh38
rs984471924 Chr.11: 119097136-119097136 on Build GRCh38
rs763778104 Chr.11: 119097438-119097438 on Build GRCh38
rs138544311 Chr.11: 119097475-119097475 on Build GRCh38
rs776863215 Chr.11: 119097324-119097324 on Build GRCh38
rs765254519 Chr.11: 119097173-119097173 on Build GRCh38
rs753712106 Chr.11: 119097444-119097444 on Build GRCh38
rs972565409 Chr.11: 119097170-119097170 on Build GRCh38
rs550478594 Chr.11: 119097497-119097497 on Build GRCh38
rs528948285 Chr.11: 119097586-119097586 on Build GRCh38
rs752042526 Chr.11: 119097616-119097616 on Build GRCh38

View Full Product Details