Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117625644-117626229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46C
Entrez Gene ID
54855
Gene Name
family with sequence similarity 46 member C
Gene Aliases
-
UniGene
Hs.356216
Gene Chromosome Location
Chr.1: 117605982-117628389 on Build GRCh38
Transcripts
NM_017709.3

SNP Detail

SNP ID Chromosome Location
rs150395948 Chr.1: 117626049-117626049 on Build GRCh38
rs929897877 Chr.1: 117626102-117626102 on Build GRCh38
rs960300241 Chr.1: 117625999-117625999 on Build GRCh38
rs940344161 Chr.1: 117625899-117625899 on Build GRCh38
rs903444572 Chr.1: 117625738-117625738 on Build GRCh38
rs896087501 Chr.1: 117626018-117626018 on Build GRCh38
rs371508363 Chr.1: 117626038-117626038 on Build GRCh38
rs1006347947 Chr.1: 117626065-117626065 on Build GRCh38
rs552710545 Chr.1: 117625864-117625864 on Build GRCh38
rs564558111 Chr.1: 117625944-117625944 on Build GRCh38
rs183925178 Chr.1: 117625993-117625993 on Build GRCh38
rs1057294481 Chr.1: 117626015-117626015 on Build GRCh38
rs1019939095 Chr.1: 117626033-117626033 on Build GRCh38
rs886552809 Chr.1: 117625962-117625962 on Build GRCh38
rs779754433 Chr.1: 117626078-117626078 on Build GRCh38
rs1048226235 Chr.1: 117626119-117626119 on Build GRCh38
rs1011562681 Chr.1: 117625666-117625666 on Build GRCh38
rs371924885 Chr.1: 117625984-117625984 on Build GRCh38
rs901062353 Chr.1: 117625943-117625943 on Build GRCh38
rs11581122 Chr.1: 117626204-117626204 on Build GRCh38
rs978596807 Chr.1: 117625887-117625887 on Build GRCh38
rs777142022 Chr.1: 117626105-117626105 on Build GRCh38
rs569447918 Chr.1: 117625975-117625975 on Build GRCh38
rs754719846 Chr.1: 117625798-117625798 on Build GRCh38
rs749061722 Chr.1: 117626153-117626153 on Build GRCh38
rs866150091 Chr.1: 117626005-117626005 on Build GRCh38
rs867130469 Chr.1: 117625783-117625783 on Build GRCh38
rs756945379 Chr.1: 117626200-117626200 on Build GRCh38
rs1040950682 Chr.1: 117625912-117625912 on Build GRCh38
rs969155560 Chr.1: 117626054-117626054 on Build GRCh38
rs201592555 Chr.1: 117626182-117626182 on Build GRCh38
rs988471075 Chr.1: 117625780-117625780 on Build GRCh38
rs1050714836 Chr.1: 117625971-117625971 on Build GRCh38
rs867400149 Chr.1: 117625987-117625987 on Build GRCh38
rs1047707524 Chr.1: 117625955-117625955 on Build GRCh38
rs922537040 Chr.1: 117626094-117626094 on Build GRCh38
rs397963907 Chr.1: 117626183-117626184 on Build GRCh38
rs903158425 Chr.1: 117626001-117626001 on Build GRCh38
rs751334841 Chr.1: 117625719-117625719 on Build GRCh38
rs531050479 Chr.1: 117625813-117625813 on Build GRCh38
rs992088245 Chr.1: 117625789-117625789 on Build GRCh38
rs140501183 Chr.1: 117626037-117626037 on Build GRCh38
rs1037914673 Chr.1: 117625969-117625969 on Build GRCh38
rs911050824 Chr.1: 117625890-117625890 on Build GRCh38
rs963860858 Chr.1: 117626057-117626057 on Build GRCh38
rs192682484 Chr.1: 117625746-117625746 on Build GRCh38
rs138846269 Chr.1: 117625704-117625704 on Build GRCh38
rs550993589 Chr.1: 117625769-117625769 on Build GRCh38
rs563859392 Chr.1: 117625778-117625778 on Build GRCh38
rs57603364 Chr.1: 117626182-117626183 on Build GRCh38
rs1037801345 Chr.1: 117626196-117626196 on Build GRCh38
rs910411986 Chr.1: 117626186-117626186 on Build GRCh38
rs984469565 Chr.1: 117625861-117625861 on Build GRCh38
rs546286689 Chr.1: 117625884-117625884 on Build GRCh38
rs548084333 Chr.1: 117626185-117626185 on Build GRCh38
rs1006303016 Chr.1: 117625963-117625963 on Build GRCh38
rs534147233 Chr.1: 117626069-117626069 on Build GRCh38
rs969573854 Chr.1: 117625827-117625827 on Build GRCh38
rs748985144 Chr.1: 117626160-117626160 on Build GRCh38
rs999097250 Chr.1: 117625755-117625755 on Build GRCh38
rs1010344147 Chr.1: 117626009-117626009 on Build GRCh38
rs150708230 Chr.1: 117625990-117625991 on Build GRCh38
rs942495642 Chr.1: 117625895-117625895 on Build GRCh38

View Full Product Details