Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112779320-112779829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1A
Entrez Gene ID
3552
Gene Name
interleukin 1 alpha
Gene Aliases
IL-1A, IL1, IL1-ALPHA, IL1F1
UniGene
Hs.1722
Gene Chromosome Location
Chr.2: 112773915-112785398 on Build GRCh38
Transcripts
NM_000575.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3757583 c.340G>T p.A114S Chr.2: 112779646-112779646 on Build GRCh38
5024420 c.366G>A p.V122V Chr.2: 112779620-112779620 on Build GRCh38
3743964 c.432A>G p.I144M Chr.2: 112779554-112779554 on Build GRCh38
6542941 c.436C>A p.R146R Chr.2: 112779550-112779550 on Build GRCh38
5719661 c.436C>T p.R146* Chr.2: 112779550-112779550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033232786 Chr.2: 112779646-112779647 on Build GRCh38
rs957699983 Chr.2: 112779655-112779655 on Build GRCh38
rs2071374 Chr.2: 112779775-112779775 on Build GRCh38
rs774342219 Chr.2: 112779679-112779679 on Build GRCh38
rs1004191767 Chr.2: 112779395-112779395 on Build GRCh38
rs780412122 Chr.2: 112779580-112779580 on Build GRCh38
rs17561 Chr.2: 112779646-112779646 on Build GRCh38
rs116381488 Chr.2: 112779379-112779379 on Build GRCh38
rs553259442 Chr.2: 112779589-112779589 on Build GRCh38
rs887204297 Chr.2: 112779382-112779382 on Build GRCh38
rs547310223 Chr.2: 112779367-112779367 on Build GRCh38
rs111867912 Chr.2: 112779623-112779623 on Build GRCh38
rs559669505 Chr.2: 112779406-112779407 on Build GRCh38
rs990581908 Chr.2: 112779509-112779509 on Build GRCh38
rs777277329 Chr.2: 112779495-112779495 on Build GRCh38
rs1048278740 Chr.2: 112779557-112779557 on Build GRCh38
rs529562970 Chr.2: 112779789-112779789 on Build GRCh38
rs138168355 Chr.2: 112779527-112779527 on Build GRCh38
rs56128969 Chr.2: 112779722-112779722 on Build GRCh38
rs949243284 Chr.2: 112779522-112779522 on Build GRCh38
rs775178763 Chr.2: 112779536-112779536 on Build GRCh38
rs549138446 Chr.2: 112779447-112779447 on Build GRCh38
rs770329499 Chr.2: 112779526-112779526 on Build GRCh38
rs17562 Chr.2: 112779613-112779613 on Build GRCh38
rs535680960 Chr.2: 112779391-112779391 on Build GRCh38
rs931474617 Chr.2: 112779377-112779377 on Build GRCh38
rs753694807 Chr.2: 112779560-112779560 on Build GRCh38
rs766602023 Chr.2: 112779571-112779571 on Build GRCh38
rs376957964 Chr.2: 112779572-112779572 on Build GRCh38
rs200281020 Chr.2: 112779676-112779676 on Build GRCh38
rs916228892 Chr.2: 112779747-112779747 on Build GRCh38
rs201678238 Chr.2: 112779549-112779549 on Build GRCh38
rs544340593 Chr.2: 112779735-112779735 on Build GRCh38
rs767094001 Chr.2: 112779534-112779534 on Build GRCh38
rs936299117 Chr.2: 112779492-112779492 on Build GRCh38
rs564072850 Chr.2: 112779421-112779421 on Build GRCh38
rs750387987 Chr.2: 112779702-112779702 on Build GRCh38
rs976909580 Chr.2: 112779349-112779349 on Build GRCh38
rs190193834 Chr.2: 112779564-112779564 on Build GRCh38
rs987079326 Chr.2: 112779577-112779577 on Build GRCh38
rs3783581 Chr.2: 112779574-112779574 on Build GRCh38
rs147192852 Chr.2: 112779578-112779578 on Build GRCh38
rs761519169 Chr.2: 112779673-112779673 on Build GRCh38
rs1020480904 Chr.2: 112779624-112779624 on Build GRCh38
rs147376170 Chr.2: 112779348-112779349 on Build GRCh38
rs370144723 Chr.2: 112779599-112779599 on Build GRCh38
rs542058674 Chr.2: 112779643-112779643 on Build GRCh38
rs376874217 Chr.2: 112779528-112779528 on Build GRCh38
rs2856841 Chr.2: 112779762-112779762 on Build GRCh38
rs777952076 Chr.2: 112779469-112779470 on Build GRCh38
rs1023215957 Chr.2: 112779546-112779546 on Build GRCh38
rs900033608 Chr.2: 112779342-112779342 on Build GRCh38
rs761675423 Chr.2: 112779729-112779729 on Build GRCh38
rs1035714873 Chr.2: 112779417-112779417 on Build GRCh38
rs763915713 Chr.2: 112779550-112779550 on Build GRCh38
rs113173604 Chr.2: 112779386-112779386 on Build GRCh38
rs748732116 Chr.2: 112779519-112779519 on Build GRCh38
rs748487868 Chr.2: 112779590-112779590 on Build GRCh38
rs768318856 Chr.2: 112779651-112779651 on Build GRCh38
rs769203512 Chr.2: 112779493-112779493 on Build GRCh38
rs181898774 Chr.2: 112779792-112779792 on Build GRCh38
rs747321400 Chr.2: 112779473-112779473 on Build GRCh38
rs776446133 Chr.2: 112779667-112779667 on Build GRCh38
rs372496657 Chr.2: 112779611-112779611 on Build GRCh38
rs924964291 Chr.2: 112779431-112779431 on Build GRCh38
rs990460189 Chr.2: 112779715-112779715 on Build GRCh38

View Full Product Details