Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34256082-34256664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL2
Entrez Gene ID
6347
Gene Name
C-C motif chemokine ligand 2
Gene Aliases
GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, SCYA2, SMC-CF
UniGene
Hs.303649
Gene Chromosome Location
Chr.17: 34255277-34257203 on Build GRCh38
Transcripts
NM_002982.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3180778 c.158G>T p.R53I Chr.17: 34256303-34256303 on Build GRCh38
3362076 c.140G>C p.R47T Chr.17: 34256285-34256285 on Build GRCh38
4903772 c.123G>A p.R41R Chr.17: 34256268-34256268 on Build GRCh38
4931284 c.132A>G p.S44S Chr.17: 34256277-34256277 on Build GRCh38
3402771 c.194T>A p.I65N Chr.17: 34256339-34256339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552587136 Chr.17: 34256617-34256617 on Build GRCh38
rs775202538 Chr.17: 34256450-34256450 on Build GRCh38
rs138139914 Chr.17: 34256185-34256185 on Build GRCh38
rs758414868 Chr.17: 34256557-34256557 on Build GRCh38
rs780082637 Chr.17: 34256377-34256377 on Build GRCh38
rs750326048 Chr.17: 34256411-34256411 on Build GRCh38
rs573451566 Chr.17: 34256603-34256603 on Build GRCh38
rs965087353 Chr.17: 34256584-34256584 on Build GRCh38
rs371187910 Chr.17: 34256265-34256265 on Build GRCh38
rs1014647063 Chr.17: 34256567-34256567 on Build GRCh38
rs1045059704 Chr.17: 34256412-34256412 on Build GRCh38
rs757324070 Chr.17: 34256210-34256210 on Build GRCh38
rs886567880 Chr.17: 34256505-34256505 on Build GRCh38
rs371894050 Chr.17: 34256372-34256372 on Build GRCh38
rs181103169 Chr.17: 34256443-34256443 on Build GRCh38
rs752116674 Chr.17: 34256197-34256197 on Build GRCh38
rs145056785 Chr.17: 34256267-34256267 on Build GRCh38
rs375842964 Chr.17: 34256171-34256171 on Build GRCh38
rs1027807658 Chr.17: 34256319-34256319 on Build GRCh38
rs977958322 Chr.17: 34256616-34256616 on Build GRCh38
rs368054591 Chr.17: 34256366-34256366 on Build GRCh38
rs759111149 Chr.17: 34256298-34256298 on Build GRCh38
rs866745401 Chr.17: 34256268-34256268 on Build GRCh38
rs375267079 Chr.17: 34256390-34256390 on Build GRCh38
rs141727366 Chr.17: 34256611-34256611 on Build GRCh38
rs1055603156 Chr.17: 34256524-34256524 on Build GRCh38
rs916218908 Chr.17: 34256402-34256402 on Build GRCh38
rs367586228 Chr.17: 34256354-34256354 on Build GRCh38
rs1053478963 Chr.17: 34256398-34256398 on Build GRCh38
rs4586 Chr.17: 34256250-34256250 on Build GRCh38
rs945848533 Chr.17: 34256167-34256167 on Build GRCh38
rs891547429 Chr.17: 34256149-34256149 on Build GRCh38
rs765613603 Chr.17: 34256174-34256174 on Build GRCh38
rs764759504 Chr.17: 34256305-34256305 on Build GRCh38
rs146604396 Chr.17: 34256227-34256227 on Build GRCh38
rs779990429 Chr.17: 34256558-34256558 on Build GRCh38
rs536707537 Chr.17: 34256451-34256451 on Build GRCh38
rs945801490 Chr.17: 34256442-34256442 on Build GRCh38
rs746327384 Chr.17: 34256172-34256172 on Build GRCh38
rs372033538 Chr.17: 34256602-34256602 on Build GRCh38
rs2857657 Chr.17: 34256113-34256113 on Build GRCh38
rs760531591 Chr.17: 34256633-34256633 on Build GRCh38
rs772588061 Chr.17: 34256268-34256269 on Build GRCh38
rs758795760 Chr.17: 34256188-34256188 on Build GRCh38
rs1047879741 Chr.17: 34256459-34256459 on Build GRCh38
rs143644230 Chr.17: 34256310-34256310 on Build GRCh38
rs3917888 Chr.17: 34256364-34256364 on Build GRCh38
rs750428936 Chr.17: 34256365-34256365 on Build GRCh38
rs912946382 Chr.17: 34256431-34256431 on Build GRCh38
rs904361088 Chr.17: 34256492-34256492 on Build GRCh38
rs186383230 Chr.17: 34256533-34256533 on Build GRCh38
rs745556919 Chr.17: 34256416-34256416 on Build GRCh38
rs780351792 Chr.17: 34256260-34256260 on Build GRCh38
rs752406037 Chr.17: 34256308-34256308 on Build GRCh38
rs937209119 Chr.17: 34256517-34256517 on Build GRCh38
rs1041161720 Chr.17: 34256140-34256140 on Build GRCh38
rs746076530 Chr.17: 34256238-34256238 on Build GRCh38
rs1006700403 Chr.17: 34256536-34256536 on Build GRCh38
rs148937652 Chr.17: 34256291-34256291 on Build GRCh38
rs1014914592 Chr.17: 34256582-34256582 on Build GRCh38
rs771891758 Chr.17: 34256449-34256449 on Build GRCh38
rs748759159 Chr.17: 34256194-34256194 on Build GRCh38
rs565820582 Chr.17: 34256195-34256195 on Build GRCh38
rs764505290 Chr.17: 34256342-34256342 on Build GRCh38
rs760761240 Chr.17: 34256290-34256290 on Build GRCh38
rs138768575 Chr.17: 34256289-34256289 on Build GRCh38

View Full Product Details