Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42781312-42781840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM216B
Entrez Gene ID
144809
Gene Name
family with sequence similarity 216 member B
Gene Aliases
C13orf30
UniGene
Hs.16970
Gene Chromosome Location
Chr.13: 42781550-42791549 on Build GRCh38
Transcripts
NM_001318932.1 NM_182508.2

SNP Detail

SNP ID Chromosome Location
rs954861079 Chr.13: 42781639-42781639 on Build GRCh38
rs995752566 Chr.13: 42781376-42781376 on Build GRCh38
rs529747881 Chr.13: 42781379-42781379 on Build GRCh38
rs765447026 Chr.13: 42781378-42781378 on Build GRCh38
rs910403597 Chr.13: 42781685-42781685 on Build GRCh38
rs913778046 Chr.13: 42781604-42781604 on Build GRCh38
rs1031964869 Chr.13: 42781535-42781535 on Build GRCh38
rs1039847278 Chr.13: 42781696-42781696 on Build GRCh38
rs1005347896 Chr.13: 42781601-42781601 on Build GRCh38
rs781114968 Chr.13: 42781687-42781687 on Build GRCh38
rs145381008 Chr.13: 42781637-42781637 on Build GRCh38
rs982755682 Chr.13: 42781784-42781784 on Build GRCh38
rs763035521 Chr.13: 42781396-42781396 on Build GRCh38
rs116194421 Chr.13: 42781713-42781713 on Build GRCh38
rs763420305 Chr.13: 42781696-42781696 on Build GRCh38
rs931264310 Chr.13: 42781722-42781722 on Build GRCh38
rs547508499 Chr.13: 42781706-42781706 on Build GRCh38
rs954804690 Chr.13: 42781631-42781631 on Build GRCh38
rs927683155 Chr.13: 42781803-42781803 on Build GRCh38
rs117479184 Chr.13: 42781602-42781602 on Build GRCh38
rs888296821 Chr.13: 42781549-42781549 on Build GRCh38
rs989710339 Chr.13: 42781377-42781377 on Build GRCh38
rs546277776 Chr.13: 42781498-42781498 on Build GRCh38
rs927620022 Chr.13: 42781463-42781463 on Build GRCh38
rs184881432 Chr.13: 42781660-42781660 on Build GRCh38
rs998506229 Chr.13: 42781613-42781613 on Build GRCh38
rs117066061 Chr.13: 42781358-42781358 on Build GRCh38
rs192791125 Chr.13: 42781603-42781603 on Build GRCh38
rs945045805 Chr.13: 42781444-42781444 on Build GRCh38
rs1047525913 Chr.13: 42781740-42781740 on Build GRCh38
rs980511859 Chr.13: 42781504-42781504 on Build GRCh38
rs1013732254 Chr.13: 42781384-42781384 on Build GRCh38
rs764142571 Chr.13: 42781457-42781457 on Build GRCh38
rs187715510 Chr.13: 42781362-42781362 on Build GRCh38
rs967027404 Chr.13: 42781768-42781768 on Build GRCh38
rs750195248 Chr.13: 42781762-42781762 on Build GRCh38
rs764626196 Chr.13: 42781704-42781705 on Build GRCh38
rs937802097 Chr.13: 42781495-42781495 on Build GRCh38
rs41288317 Chr.13: 42781672-42781672 on Build GRCh38
rs137976253 Chr.13: 42781665-42781665 on Build GRCh38
rs776639837 Chr.13: 42781560-42781560 on Build GRCh38
rs1017620205 Chr.13: 42781681-42781681 on Build GRCh38
rs371631165 Chr.13: 42781725-42781725 on Build GRCh38
rs36038851 Chr.13: 42781737-42781738 on Build GRCh38
rs1025576997 Chr.13: 42781393-42781393 on Build GRCh38
rs887338982 Chr.13: 42781812-42781812 on Build GRCh38

View Full Product Details