Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42526887-42527423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM1
Entrez Gene ID
634
Gene Name
carcinoembryonic antigen related cell adhesion molecule 1
Gene Aliases
BGP, BGP1, BGPI
UniGene
Hs.512682
Gene Chromosome Location
Chr.19: 42507306-42528509 on Build GRCh38
Transcripts
NM_001024912.2 NM_001184813.1 NM_001184815.1 NM_001184816.1 NM_001205344.1 NM_001712.4 XM_011527206.1 NR_073179.1 NR_073180.1

Gene Symbol
LIPE-AS1
Entrez Gene ID
100996307
Gene Name
LIPE antisense RNA 1
Gene Aliases
-
UniGene
Hs.466840
Gene Chromosome Location
Chr.19: 42397148-42652355 on Build GRCh38
Transcripts
NM_001024912.2 NM_001184813.1 NM_001184815.1 NM_001184816.1 NM_001205344.1 NM_001712.4 XM_011527206.1 NR_073179.1 NR_073180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4600593 c.350C>T p.T117I Chr.19: 42527115-42527115 on Build GRCh38
6337450 c.338C>T p.T113I Chr.19: 42527127-42527127 on Build GRCh38
4671307 c.385C>A p.L129I Chr.19: 42527080-42527080 on Build GRCh38
4483564 c.270C>T p.T90T Chr.19: 42527195-42527195 on Build GRCh38
7200222 c.229C>A p.R77S Chr.19: 42527236-42527236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769948505 Chr.19: 42527312-42527312 on Build GRCh38
rs780866468 Chr.19: 42527388-42527388 on Build GRCh38
rs931297348 Chr.19: 42526958-42526958 on Build GRCh38
rs41368746 Chr.19: 42527246-42527246 on Build GRCh38
rs371432630 Chr.19: 42527291-42527291 on Build GRCh38
rs8111171 Chr.19: 42527362-42527362 on Build GRCh38
rs746267737 Chr.19: 42527045-42527045 on Build GRCh38
rs745803770 Chr.19: 42527186-42527186 on Build GRCh38
rs537091721 Chr.19: 42527003-42527003 on Build GRCh38
rs754014608 Chr.19: 42527154-42527154 on Build GRCh38
rs911026263 Chr.19: 42526918-42526918 on Build GRCh38
rs36032678 Chr.19: 42527145-42527146 on Build GRCh38
rs775481585 Chr.19: 42527191-42527191 on Build GRCh38
rs756887837 Chr.19: 42527156-42527156 on Build GRCh38
rs140166708 Chr.19: 42527203-42527203 on Build GRCh38
rs772023155 Chr.19: 42527127-42527127 on Build GRCh38
rs759399284 Chr.19: 42527005-42527005 on Build GRCh38
rs776442230 Chr.19: 42527294-42527294 on Build GRCh38
rs764240232 Chr.19: 42527027-42527027 on Build GRCh38
rs772076119 Chr.19: 42527188-42527188 on Build GRCh38
rs8110904 Chr.19: 42527217-42527217 on Build GRCh38
rs774699735 Chr.19: 42527330-42527330 on Build GRCh38
rs201059984 Chr.19: 42527131-42527131 on Build GRCh38
rs917883278 Chr.19: 42527297-42527297 on Build GRCh38
rs574464572 Chr.19: 42527219-42527219 on Build GRCh38
rs764207981 Chr.19: 42527150-42527150 on Build GRCh38
rs763466920 Chr.19: 42527223-42527223 on Build GRCh38
rs756902598 Chr.19: 42527254-42527254 on Build GRCh38
rs143477834 Chr.19: 42527177-42527177 on Build GRCh38
rs756251280 Chr.19: 42527017-42527017 on Build GRCh38
rs754874897 Chr.19: 42526973-42526973 on Build GRCh38
rs1045273595 Chr.19: 42527077-42527077 on Build GRCh38
rs766946386 Chr.19: 42527258-42527258 on Build GRCh38
rs758736431 Chr.19: 42527050-42527050 on Build GRCh38
rs751204805 Chr.19: 42527035-42527038 on Build GRCh38
rs946653493 Chr.19: 42526931-42526931 on Build GRCh38
rs368731299 Chr.19: 42527012-42527012 on Build GRCh38
rs757027296 Chr.19: 42527339-42527339 on Build GRCh38
rs145087706 Chr.19: 42527176-42527176 on Build GRCh38
rs142053263 Chr.19: 42527182-42527182 on Build GRCh38
rs748326525 Chr.19: 42527295-42527295 on Build GRCh38
rs777289066 Chr.19: 42527180-42527180 on Build GRCh38
rs753047939 Chr.19: 42527334-42527334 on Build GRCh38
rs148302261 Chr.19: 42527331-42527331 on Build GRCh38
rs200708090 Chr.19: 42527232-42527232 on Build GRCh38
rs762042180 Chr.19: 42527208-42527208 on Build GRCh38
rs199627333 Chr.19: 42527166-42527166 on Build GRCh38
rs755269102 Chr.19: 42527238-42527238 on Build GRCh38
rs201542185 Chr.19: 42527001-42527001 on Build GRCh38
rs139751234 Chr.19: 42527041-42527041 on Build GRCh38
rs780343852 Chr.19: 42527060-42527060 on Build GRCh38
rs200819679 Chr.19: 42527352-42527352 on Build GRCh38
rs151150997 Chr.19: 42527183-42527183 on Build GRCh38
rs765778184 Chr.19: 42527158-42527158 on Build GRCh38
rs765609135 Chr.19: 42527005-42527005 on Build GRCh38
rs770969753 Chr.19: 42527097-42527097 on Build GRCh38
rs77501817 Chr.19: 42527115-42527115 on Build GRCh38
rs769266784 Chr.19: 42527088-42527088 on Build GRCh38
rs923879481 Chr.19: 42527242-42527242 on Build GRCh38
rs749866933 Chr.19: 42527307-42527309 on Build GRCh38
rs751912957 Chr.19: 42527235-42527235 on Build GRCh38
rs770325557 Chr.19: 42527185-42527185 on Build GRCh38
rs749934399 Chr.19: 42527376-42527376 on Build GRCh38
rs961371875 Chr.19: 42527168-42527168 on Build GRCh38
rs760963879 Chr.19: 42527132-42527132 on Build GRCh38
rs753803949 Chr.19: 42527033-42527033 on Build GRCh38
rs542149555 Chr.19: 42527212-42527212 on Build GRCh38
rs142826356 Chr.19: 42527067-42527067 on Build GRCh38
rs766913973 Chr.19: 42527323-42527323 on Build GRCh38
rs773342137 Chr.19: 42527314-42527314 on Build GRCh38
rs760381268 Chr.19: 42527195-42527195 on Build GRCh38
rs144124872 Chr.19: 42527156-42527156 on Build GRCh38
rs887923942 Chr.19: 42527079-42527079 on Build GRCh38
rs985267148 Chr.19: 42526912-42526912 on Build GRCh38
rs140316654 Chr.19: 42527282-42527282 on Build GRCh38
rs772200865 Chr.19: 42527260-42527260 on Build GRCh38
rs8111468 Chr.19: 42527096-42527096 on Build GRCh38
rs112464894 Chr.19: 42526930-42526930 on Build GRCh38
rs79326931 Chr.19: 42527255-42527255 on Build GRCh38
rs375987435 Chr.19: 42527205-42527205 on Build GRCh38
rs774401390 Chr.19: 42527105-42527105 on Build GRCh38
rs377492728 Chr.19: 42527375-42527375 on Build GRCh38
rs766860794 Chr.19: 42527225-42527225 on Build GRCh38
rs920018659 Chr.19: 42526957-42526957 on Build GRCh38
rs41345145 Chr.19: 42527004-42527005 on Build GRCh38
rs902551857 Chr.19: 42526992-42526992 on Build GRCh38
rs143222169 Chr.19: 42527317-42527317 on Build GRCh38
rs764975765 Chr.19: 42527343-42527343 on Build GRCh38
rs772794650 Chr.19: 42527090-42527090 on Build GRCh38
rs758867188 Chr.19: 42527172-42527172 on Build GRCh38
rs1046613330 Chr.19: 42527320-42527320 on Build GRCh38
rs779171413 Chr.19: 42526966-42526966 on Build GRCh38
rs767891864 Chr.19: 42527006-42527006 on Build GRCh38

View Full Product Details