Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 108939679-108940219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM206A
Entrez Gene ID
54942
Gene Name
family with sequence similarity 206 member A
Gene Aliases
C9orf6, CG-8, Simiate
UniGene
Hs.29276
Gene Chromosome Location
Chr.9: 108934393-108950745 on Build GRCh38
Transcripts
NM_017832.3 XM_011518816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659314 c.539C>T p.T180M Chr.9: 108939679-108939679 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs138410652 Chr.9: 108940188-108940188 on Build GRCh38
rs967862351 Chr.9: 108939745-108939745 on Build GRCh38
rs567510331 Chr.9: 108940166-108940166 on Build GRCh38
rs748497990 Chr.9: 108939802-108939802 on Build GRCh38
rs772538756 Chr.9: 108939870-108939870 on Build GRCh38
rs776599910 Chr.9: 108940040-108940040 on Build GRCh38
rs551799944 Chr.9: 108939910-108939910 on Build GRCh38
rs919810700 Chr.9: 108939896-108939896 on Build GRCh38
rs773326607 Chr.9: 108939720-108939720 on Build GRCh38
rs1046099209 Chr.9: 108940113-108940113 on Build GRCh38
rs111581446 Chr.9: 108939967-108939967 on Build GRCh38
rs771223544 Chr.9: 108939997-108939997 on Build GRCh38
rs1001628063 Chr.9: 108940118-108940118 on Build GRCh38
rs201618355 Chr.9: 108940149-108940149 on Build GRCh38
rs182108702 Chr.9: 108939944-108939944 on Build GRCh38
rs986914558 Chr.9: 108939746-108939746 on Build GRCh38
rs755461653 Chr.9: 108939708-108939708 on Build GRCh38
rs543774629 Chr.9: 108940150-108940151 on Build GRCh38
rs376866063 Chr.9: 108939883-108939883 on Build GRCh38
rs904954457 Chr.9: 108940116-108940116 on Build GRCh38
rs942469714 Chr.9: 108939831-108939831 on Build GRCh38
rs562263888 Chr.9: 108940185-108940185 on Build GRCh38
rs35501409 Chr.9: 108940154-108940155 on Build GRCh38
rs759624626 Chr.9: 108940086-108940086 on Build GRCh38
rs563981119 Chr.9: 108940095-108940095 on Build GRCh38
rs576392394 Chr.9: 108940137-108940137 on Build GRCh38
rs768772964 Chr.9: 108940181-108940181 on Build GRCh38
rs143613903 Chr.9: 108939787-108939787 on Build GRCh38
rs886818597 Chr.9: 108940014-108940014 on Build GRCh38

View Full Product Details