Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18588005-18588590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL5
Entrez Gene ID
6792
Gene Name
cyclin dependent kinase like 5
Gene Aliases
CFAP247, EIEE2, ISSX, STK9
UniGene
Hs.659851
Gene Chromosome Location
Chr.X: 18425605-18653629 on Build GRCh38
Transcripts
NM_001037343.1 NM_001323289.1 NM_003159.2

SNP Detail

SNP ID Chromosome Location
rs984314889 Chr.X: 18588444-18588444 on Build GRCh38
rs587783088 Chr.X: 18588025-18588025 on Build GRCh38
rs1025782197 Chr.X: 18588283-18588283 on Build GRCh38
rs972248532 Chr.X: 18588432-18588432 on Build GRCh38
rs868502587 Chr.X: 18588126-18588126 on Build GRCh38
rs199850115 Chr.X: 18588192-18588198 on Build GRCh38
rs998525792 Chr.X: 18588178-18588178 on Build GRCh38
rs748464940 Chr.X: 18588395-18588396 on Build GRCh38
rs267608511 Chr.X: 18588058-18588058 on Build GRCh38
rs910963859 Chr.X: 18588494-18588494 on Build GRCh38
rs983754693 Chr.X: 18588317-18588317 on Build GRCh38
rs756917829 Chr.X: 18588487-18588487 on Build GRCh38
rs786204963 Chr.X: 18588055-18588055 on Build GRCh38
rs886041764 Chr.X: 18588092-18588092 on Build GRCh38
rs781623603 Chr.X: 18588512-18588512 on Build GRCh38
rs961113154 Chr.X: 18588361-18588361 on Build GRCh38
rs150769601 Chr.X: 18588255-18588255 on Build GRCh38
rs752118445 Chr.X: 18588073-18588073 on Build GRCh38
rs587783089 Chr.X: 18588099-18588099 on Build GRCh38
rs752062491 Chr.X: 18588242-18588245 on Build GRCh38
rs374585296 Chr.X: 18588141-18588141 on Build GRCh38
rs786204990 Chr.X: 18588063-18588064 on Build GRCh38
rs781568006 Chr.X: 18588292-18588292 on Build GRCh38
rs901121074 Chr.X: 18588064-18588064 on Build GRCh38
rs970943968 Chr.X: 18588038-18588038 on Build GRCh38
rs1016727536 Chr.X: 18588346-18588346 on Build GRCh38
rs755330309 Chr.X: 18588086-18588086 on Build GRCh38
rs766475539 Chr.X: 18588118-18588118 on Build GRCh38
rs267608515 Chr.X: 18588079-18588079 on Build GRCh38
rs994349946 Chr.X: 18588261-18588261 on Build GRCh38
rs370073267 Chr.X: 18588059-18588059 on Build GRCh38
rs976423808 Chr.X: 18588521-18588521 on Build GRCh38
rs869312851 Chr.X: 18588078-18588090 on Build GRCh38
rs751883002 Chr.X: 18588035-18588035 on Build GRCh38
rs752286259 Chr.X: 18588140-18588140 on Build GRCh38
rs755342565 Chr.X: 18588314-18588314 on Build GRCh38

View Full Product Details