Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 70958031-70958591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP1
Entrez Gene ID
27086
Gene Name
forkhead box P1
Gene Aliases
12CC4, HSPC215, MFH, QRF1, hFKH1B
UniGene
Hs.59368
Gene Chromosome Location
Chr.3: 70954714-71583989 on Build GRCh38
Transcripts
NM_001244808.1 NM_001244810.1 NM_001244812.1 NM_001244813.1 NM_001244814.1 NM_001244815.1 NM_001244816.1 NM_032682.5 XM_006713102.2 XM_011533584.2 XM_011533585.2 XM_017006165.1

SNP Detail

SNP ID Chromosome Location
rs886058847 Chr.3: 70958143-70958143 on Build GRCh38
rs199796506 Chr.3: 70958196-70958196 on Build GRCh38
rs189975378 Chr.3: 70958404-70958404 on Build GRCh38
rs879794923 Chr.3: 70958323-70958324 on Build GRCh38
rs199626546 Chr.3: 70958122-70958123 on Build GRCh38
rs756449056 Chr.3: 70958312-70958312 on Build GRCh38
rs867338460 Chr.3: 70958116-70958116 on Build GRCh38
rs932923321 Chr.3: 70958064-70958064 on Build GRCh38
rs750931869 Chr.3: 70958212-70958215 on Build GRCh38
rs559246026 Chr.3: 70958211-70958211 on Build GRCh38
rs796832662 Chr.3: 70958122-70958122 on Build GRCh38
rs60236784 Chr.3: 70958431-70958431 on Build GRCh38
rs771272366 Chr.3: 70958312-70958312 on Build GRCh38
rs185411934 Chr.3: 70958251-70958251 on Build GRCh38
rs555165294 Chr.3: 70958058-70958058 on Build GRCh38
rs1050024390 Chr.3: 70958350-70958350 on Build GRCh38
rs549654259 Chr.3: 70958206-70958210 on Build GRCh38
rs912786562 Chr.3: 70958364-70958364 on Build GRCh38
rs755536663 Chr.3: 70958225-70958228 on Build GRCh38
rs148080961 Chr.3: 70958286-70958286 on Build GRCh38
rs925528524 Chr.3: 70958057-70958057 on Build GRCh38
rs886058848 Chr.3: 70958147-70958147 on Build GRCh38
rs372613770 Chr.3: 70958105-70958105 on Build GRCh38
rs886058852 Chr.3: 70958503-70958504 on Build GRCh38
rs376254944 Chr.3: 70958438-70958438 on Build GRCh38
rs200557663 Chr.3: 70958138-70958138 on Build GRCh38
rs564312925 Chr.3: 70958201-70958201 on Build GRCh38
rs548648975 Chr.3: 70958215-70958215 on Build GRCh38
rs1001433405 Chr.3: 70958262-70958263 on Build GRCh38
rs995580595 Chr.3: 70958466-70958466 on Build GRCh38
rs577554096 Chr.3: 70958239-70958239 on Build GRCh38
rs757890424 Chr.3: 70958553-70958553 on Build GRCh38
rs777481567 Chr.3: 70958273-70958273 on Build GRCh38
rs769123792 Chr.3: 70958338-70958338 on Build GRCh38
rs542290375 Chr.3: 70958422-70958422 on Build GRCh38
rs1001465890 Chr.3: 70958538-70958538 on Build GRCh38
rs544766693 Chr.3: 70958312-70958314 on Build GRCh38
rs962619624 Chr.3: 70958281-70958281 on Build GRCh38
rs1036028283 Chr.3: 70958564-70958564 on Build GRCh38
rs886058850 Chr.3: 70958207-70958210 on Build GRCh38
rs1026985056 Chr.3: 70958231-70958231 on Build GRCh38
rs868014063 Chr.3: 70958311-70958311 on Build GRCh38
rs1006325220 Chr.3: 70958256-70958256 on Build GRCh38
rs1037537383 Chr.3: 70958210-70958210 on Build GRCh38
rs568661192 Chr.3: 70958420-70958420 on Build GRCh38
rs886058849 Chr.3: 70958196-70958197 on Build GRCh38
rs988631959 Chr.3: 70958285-70958285 on Build GRCh38
rs928668402 Chr.3: 70958495-70958495 on Build GRCh38
rs944213365 Chr.3: 70958417-70958417 on Build GRCh38
rs752181758 Chr.3: 70958252-70958252 on Build GRCh38
rs57633791 Chr.3: 70958457-70958457 on Build GRCh38
rs747818299 Chr.3: 70958197-70958197 on Build GRCh38
rs995471068 Chr.3: 70958216-70958217 on Build GRCh38
rs577920948 Chr.3: 70958204-70958204 on Build GRCh38
rs181284076 Chr.3: 70958053-70958053 on Build GRCh38
rs905894649 Chr.3: 70958505-70958505 on Build GRCh38
rs912979818 Chr.3: 70958306-70958309 on Build GRCh38
rs867069977 Chr.3: 70958305-70958305 on Build GRCh38
rs1048447904 Chr.3: 70958468-70958468 on Build GRCh38
rs897315599 Chr.3: 70958421-70958421 on Build GRCh38
rs199789083 Chr.3: 70958515-70958515 on Build GRCh38
rs895699086 Chr.3: 70958571-70958571 on Build GRCh38
rs563597456 Chr.3: 70958368-70958368 on Build GRCh38
rs891313529 Chr.3: 70958202-70958202 on Build GRCh38
rs542332238 Chr.3: 70958386-70958386 on Build GRCh38
rs781578551 Chr.3: 70958332-70958332 on Build GRCh38
rs1049992297 Chr.3: 70958133-70958133 on Build GRCh38
rs752980081 Chr.3: 70958311-70958312 on Build GRCh38
rs758107528 Chr.3: 70958255-70958255 on Build GRCh38
rs1053797 Chr.3: 70958210-70958210 on Build GRCh38
rs145455295 Chr.3: 70958318-70958319 on Build GRCh38
rs1042710360 Chr.3: 70958052-70958055 on Build GRCh38
rs571782265 Chr.3: 70958349-70958349 on Build GRCh38
rs754515627 Chr.3: 70958235-70958235 on Build GRCh38
rs886058851 Chr.3: 70958216-70958217 on Build GRCh38
rs768176286 Chr.3: 70958303-70958304 on Build GRCh38
rs3194391 Chr.3: 70958220-70958220 on Build GRCh38

View Full Product Details