Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 107716374-107716943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
UniGene
Hs.124366
Gene Chromosome Location
Chr.3: 107522936-107811329 on Build GRCh38
Transcripts
NM_001142568.2 NM_001276286.1 NM_020235.6 XM_005247642.3 XM_005247643.3 XM_005247644.3 XM_011512999.2 XM_011513000.1 XM_011513001.1 XM_011513002.2 XM_011513003.2 XM_011513004.2 XM_011513005.2 XM_011513006.2 XM_011513007.2 XM_011513010.2 XM_017006881.1 XM_017006882.1 XM_017006883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7319123 c.326G>T p.G109V Chr.3: 107716770-107716770 on Build GRCh38
3767153 c.277C>T p.R93C Chr.3: 107716721-107716721 on Build GRCh38
3673798 c.242G>T p.R81L Chr.3: 107716686-107716686 on Build GRCh38
1647514 c.210T>G p.T70T Chr.3: 107716654-107716654 on Build GRCh38
6192729 c.172C>T p.L58F Chr.3: 107716616-107716616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188300391 Chr.3: 107716857-107716857 on Build GRCh38
rs766935975 Chr.3: 107716661-107716661 on Build GRCh38
rs757116282 Chr.3: 107716686-107716686 on Build GRCh38
rs750437806 Chr.3: 107716766-107716766 on Build GRCh38
rs767644500 Chr.3: 107716727-107716728 on Build GRCh38
rs368121079 Chr.3: 107716863-107716863 on Build GRCh38
rs759476047 Chr.3: 107716591-107716591 on Build GRCh38
rs755272385 Chr.3: 107716792-107716792 on Build GRCh38
rs373633459 Chr.3: 107716812-107716812 on Build GRCh38
rs748575488 Chr.3: 107716727-107716727 on Build GRCh38
rs766157331 Chr.3: 107716677-107716677 on Build GRCh38
rs1042391143 Chr.3: 107716886-107716886 on Build GRCh38
rs754738815 Chr.3: 107716679-107716679 on Build GRCh38
rs764724679 Chr.3: 107716404-107716404 on Build GRCh38
rs376164348 Chr.3: 107716731-107716731 on Build GRCh38
rs141068105 Chr.3: 107716758-107716758 on Build GRCh38
rs762812771 Chr.3: 107716895-107716895 on Build GRCh38
rs776320042 Chr.3: 107716874-107716874 on Build GRCh38
rs902223005 Chr.3: 107716610-107716610 on Build GRCh38
rs142400819 Chr.3: 107716685-107716685 on Build GRCh38
rs1044460424 Chr.3: 107716518-107716518 on Build GRCh38
rs372598327 Chr.3: 107716626-107716626 on Build GRCh38
rs922079554 Chr.3: 107716470-107716470 on Build GRCh38
rs775784528 Chr.3: 107716629-107716629 on Build GRCh38
rs758218676 Chr.3: 107716759-107716759 on Build GRCh38
rs1052379245 Chr.3: 107716498-107716498 on Build GRCh38
rs756206962 Chr.3: 107716650-107716650 on Build GRCh38
rs961493546 Chr.3: 107716737-107716737 on Build GRCh38
rs755899872 Chr.3: 107716585-107716585 on Build GRCh38
rs151035965 Chr.3: 107716428-107716428 on Build GRCh38
rs770557356 Chr.3: 107716867-107716867 on Build GRCh38
rs764050907 Chr.3: 107716765-107716765 on Build GRCh38
rs749597573 Chr.3: 107716722-107716722 on Build GRCh38
rs745968551 Chr.3: 107716694-107716694 on Build GRCh38
rs150018279 Chr.3: 107716624-107716624 on Build GRCh38
rs752367391 Chr.3: 107716453-107716453 on Build GRCh38
rs377216695 Chr.3: 107716636-107716636 on Build GRCh38
rs948693475 Chr.3: 107716478-107716478 on Build GRCh38
rs780167081 Chr.3: 107716786-107716786 on Build GRCh38
rs199641554 Chr.3: 107716668-107716668 on Build GRCh38
rs532073155 Chr.3: 107716747-107716747 on Build GRCh38
rs16853783 Chr.3: 107716881-107716881 on Build GRCh38
rs749687434 Chr.3: 107716634-107716634 on Build GRCh38
rs747572698 Chr.3: 107716866-107716866 on Build GRCh38
rs967749088 Chr.3: 107716649-107716649 on Build GRCh38
rs772806132 Chr.3: 107716730-107716730 on Build GRCh38
rs763964738 Chr.3: 107716896-107716896 on Build GRCh38
rs200978545 Chr.3: 107716616-107716616 on Build GRCh38
rs151079480 Chr.3: 107716740-107716740 on Build GRCh38
rs946295345 Chr.3: 107716558-107716558 on Build GRCh38
rs35906921 Chr.3: 107716623-107716623 on Build GRCh38
rs758399930 Chr.3: 107716681-107716681 on Build GRCh38
rs942089095 Chr.3: 107716444-107716444 on Build GRCh38
rs1022287476 Chr.3: 107716644-107716644 on Build GRCh38
rs938794341 Chr.3: 107716844-107716844 on Build GRCh38
rs186182645 Chr.3: 107716822-107716822 on Build GRCh38
rs375449960 Chr.3: 107716621-107716621 on Build GRCh38
rs573621357 Chr.3: 107716500-107716500 on Build GRCh38
rs779967848 Chr.3: 107716595-107716595 on Build GRCh38
rs912087215 Chr.3: 107716767-107716767 on Build GRCh38
rs774850210 Chr.3: 107716724-107716724 on Build GRCh38
rs766067246 Chr.3: 107716736-107716736 on Build GRCh38
rs199498147 Chr.3: 107716625-107716625 on Build GRCh38
rs749470151 Chr.3: 107716791-107716791 on Build GRCh38
rs767143926 Chr.3: 107716578-107716578 on Build GRCh38
rs750090973 Chr.3: 107716584-107716584 on Build GRCh38
rs902170405 Chr.3: 107716907-107716907 on Build GRCh38
rs376419281 Chr.3: 107716750-107716750 on Build GRCh38
rs932159335 Chr.3: 107716472-107716472 on Build GRCh38
rs772688060 Chr.3: 107716728-107716728 on Build GRCh38
rs144423719 Chr.3: 107716721-107716721 on Build GRCh38
rs568228467 Chr.3: 107716898-107716898 on Build GRCh38
rs753705535 Chr.3: 107716599-107716599 on Build GRCh38
rs774734427 Chr.3: 107716641-107716641 on Build GRCh38
rs370567196 Chr.3: 107716797-107716797 on Build GRCh38
rs761233387 Chr.3: 107716571-107716571 on Build GRCh38
rs890681422 Chr.3: 107716535-107716535 on Build GRCh38
rs754901525 Chr.3: 107716613-107716613 on Build GRCh38
rs756290745 Chr.3: 107716695-107716695 on Build GRCh38
rs762444200 Chr.3: 107716658-107716658 on Build GRCh38
rs973516328 Chr.3: 107716450-107716450 on Build GRCh38
rs141444090 Chr.3: 107716780-107716780 on Build GRCh38
rs768890393 Chr.3: 107716723-107716723 on Build GRCh38
rs760384793 Chr.3: 107716675-107716675 on Build GRCh38
rs775106386 Chr.3: 107716892-107716892 on Build GRCh38
rs76764171 Chr.3: 107716680-107716680 on Build GRCh38
rs769507273 Chr.3: 107716889-107716889 on Build GRCh38

View Full Product Details