Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113784091-113784427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1A
Entrez Gene ID
523
Gene Name
ATPase H+ transporting V1 subunit A
Gene Aliases
ATP6A1, ATP6V1A1, HO68, VA68, VPP2, Vma1
UniGene
Hs.477155
Gene Chromosome Location
Chr.3: 113747019-113812058 on Build GRCh38
Transcripts
NM_001690.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3767179 c.280A>T p.I94F Chr.3: 113784292-113784292 on Build GRCh38
7114769 c.263T>C p.V88A Chr.3: 113784275-113784275 on Build GRCh38
4489527 c.348C>T p.I116I Chr.3: 113784360-113784360 on Build GRCh38
3845874 c.260C>T p.S87F Chr.3: 113784272-113784272 on Build GRCh38
6608094 c.367A>G p.N123D Chr.3: 113784379-113784379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946926088 Chr.3: 113784397-113784397 on Build GRCh38
rs773297197 Chr.3: 113784272-113784272 on Build GRCh38
rs751048812 Chr.3: 113784335-113784335 on Build GRCh38
rs756891811 Chr.3: 113784209-113784209 on Build GRCh38
rs199933874 Chr.3: 113784211-113784211 on Build GRCh38
rs199687122 Chr.3: 113784308-113784308 on Build GRCh38
rs956137267 Chr.3: 113784121-113784121 on Build GRCh38
rs769912385 Chr.3: 113784228-113784228 on Build GRCh38
rs762970699 Chr.3: 113784358-113784358 on Build GRCh38
rs372411018 Chr.3: 113784384-113784384 on Build GRCh38
rs138089165 Chr.3: 113784381-113784381 on Build GRCh38
rs760842074 Chr.3: 113784275-113784275 on Build GRCh38
rs191211820 Chr.3: 113784174-113784174 on Build GRCh38
rs753243524 Chr.3: 113784351-113784351 on Build GRCh38
rs1015493953 Chr.3: 113784136-113784136 on Build GRCh38
rs374773637 Chr.3: 113784181-113784181 on Build GRCh38
rs777035513 Chr.3: 113784296-113784296 on Build GRCh38
rs747834495 Chr.3: 113784286-113784286 on Build GRCh38
rs775563385 Chr.3: 113784235-113784235 on Build GRCh38
rs753216033 Chr.3: 113784330-113784330 on Build GRCh38
rs1004247260 Chr.3: 113784122-113784122 on Build GRCh38
rs898385553 Chr.3: 113784140-113784140 on Build GRCh38
rs758889776 Chr.3: 113784332-113784332 on Build GRCh38
rs757846933 Chr.3: 113784197-113784197 on Build GRCh38
rs755979343 Chr.3: 113784353-113784353 on Build GRCh38
rs756911040 Chr.3: 113784339-113784339 on Build GRCh38
rs777278204 Chr.3: 113784198-113784198 on Build GRCh38
rs955687537 Chr.3: 113784342-113784342 on Build GRCh38
rs995441612 Chr.3: 113784177-113784177 on Build GRCh38
rs1023179728 Chr.3: 113784178-113784178 on Build GRCh38
rs145564764 Chr.3: 113784333-113784333 on Build GRCh38
rs780980407 Chr.3: 113784348-113784348 on Build GRCh38
rs749171534 Chr.3: 113784257-113784257 on Build GRCh38
rs754781460 Chr.3: 113784133-113784133 on Build GRCh38
rs765516232 Chr.3: 113784319-113784319 on Build GRCh38
rs981146211 Chr.3: 113784229-113784229 on Build GRCh38
rs778615930 Chr.3: 113784259-113784259 on Build GRCh38
rs372507494 Chr.3: 113784206-113784206 on Build GRCh38
rs749209409 Chr.3: 113784382-113784382 on Build GRCh38
rs367946158 Chr.3: 113784191-113784191 on Build GRCh38
rs992691558 Chr.3: 113784388-113784388 on Build GRCh38
rs1021578845 Chr.3: 113784148-113784148 on Build GRCh38
rs766598300 Chr.3: 113784292-113784292 on Build GRCh38
rs773207970 Chr.3: 113784396-113784396 on Build GRCh38
rs570912378 Chr.3: 113784401-113784401 on Build GRCh38
rs745637760 Chr.3: 113784219-113784219 on Build GRCh38

View Full Product Details