Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122386907-122387421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADPS2
Entrez Gene ID
93664
Gene Name
calcium dependent secretion activator 2
Gene Aliases
CAPS2
UniGene
Hs.649459
Gene Chromosome Location
Chr.7: 122318424-122886759 on Build GRCh38
Transcripts
NM_001009571.3 NM_001167940.1 NM_017954.10 XM_005250695.4 XM_005250696.4 XM_005250697.4 XM_005250698.4 XM_005250699.4 XM_005250700.4 XM_005250701.4 XM_005250702.4 XM_005250703.4 XM_005250704.4 XM_005250705.4 XM_005250706.4 XM_005250707.4 XM_005250708.3 XM_011516695.2 XM_011516696.2 XM_011516697.2 XM_017012794.1 XM_017012795.1 XM_017012796.1 XM_017012797.1 XM_017012798.1 XM_017012799.1 XM_017012800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4782670 c.3078C>A p.D1026E Chr.7: 122387122-122387122 on Build GRCh38
4488290 c.3293C>T p.A1098V Chr.7: 122387045-122387045 on Build GRCh38
5406339 c.3234C>T p.S1078S Chr.7: 122387104-122387104 on Build GRCh38
1447648 c.3216C>A p.D1072E Chr.7: 122387122-122387122 on Build GRCh38
744821 c.3181G>C p.E1061Q Chr.7: 122387157-122387157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374329379 Chr.7: 122387038-122387038 on Build GRCh38
rs907862191 Chr.7: 122387097-122387097 on Build GRCh38
rs1004623955 Chr.7: 122387240-122387240 on Build GRCh38
rs530861221 Chr.7: 122387317-122387317 on Build GRCh38
rs1028847078 Chr.7: 122387387-122387387 on Build GRCh38
rs894671681 Chr.7: 122387174-122387174 on Build GRCh38
rs751664927 Chr.7: 122387033-122387033 on Build GRCh38
rs796737267 Chr.7: 122387197-122387198 on Build GRCh38
rs759460629 Chr.7: 122386970-122386970 on Build GRCh38
rs534927407 Chr.7: 122387044-122387044 on Build GRCh38
rs775241690 Chr.7: 122387392-122387392 on Build GRCh38
rs1014409527 Chr.7: 122387199-122387199 on Build GRCh38
rs555852045 Chr.7: 122386961-122386961 on Build GRCh38
rs756515384 Chr.7: 122387077-122387077 on Build GRCh38
rs185750941 Chr.7: 122387385-122387385 on Build GRCh38
rs774144676 Chr.7: 122387204-122387204 on Build GRCh38
rs866503738 Chr.7: 122387034-122387034 on Build GRCh38
rs796816501 Chr.7: 122387327-122387331 on Build GRCh38
rs201650811 Chr.7: 122387222-122387222 on Build GRCh38
rs775608238 Chr.7: 122386978-122386978 on Build GRCh38
rs746540673 Chr.7: 122387103-122387103 on Build GRCh38
rs866584553 Chr.7: 122387064-122387064 on Build GRCh38
rs542308886 Chr.7: 122387205-122387205 on Build GRCh38
rs1001741377 Chr.7: 122386975-122386975 on Build GRCh38
rs1016975281 Chr.7: 122387248-122387248 on Build GRCh38
rs569922864 Chr.7: 122387348-122387348 on Build GRCh38
rs183942971 Chr.7: 122386969-122386969 on Build GRCh38
rs920633848 Chr.7: 122387122-122387122 on Build GRCh38
rs752801002 Chr.7: 122386948-122386948 on Build GRCh38
rs556647244 Chr.7: 122387104-122387104 on Build GRCh38
rs545271770 Chr.7: 122387254-122387254 on Build GRCh38
rs542997444 Chr.7: 122387094-122387094 on Build GRCh38
rs745485476 Chr.7: 122387118-122387118 on Build GRCh38
rs945726831 Chr.7: 122386999-122386999 on Build GRCh38
rs144898246 Chr.7: 122387332-122387332 on Build GRCh38
rs189070876 Chr.7: 122387057-122387057 on Build GRCh38
rs769522547 Chr.7: 122387127-122387127 on Build GRCh38
rs769707126 Chr.7: 122386974-122386974 on Build GRCh38
rs76528953 Chr.7: 122387076-122387076 on Build GRCh38
rs748970206 Chr.7: 122387165-122387165 on Build GRCh38
rs181462771 Chr.7: 122387117-122387117 on Build GRCh38
rs528288641 Chr.7: 122387224-122387224 on Build GRCh38
rs887158061 Chr.7: 122387217-122387217 on Build GRCh38
rs768653068 Chr.7: 122387190-122387190 on Build GRCh38
rs767841241 Chr.7: 122387060-122387060 on Build GRCh38
rs181444145 Chr.7: 122386930-122386930 on Build GRCh38
rs912171351 Chr.7: 122387303-122387303 on Build GRCh38
rs754149149 Chr.7: 122387087-122387087 on Build GRCh38
rs780378892 Chr.7: 122387083-122387083 on Build GRCh38
rs371753070 Chr.7: 122386991-122386991 on Build GRCh38
rs963025080 Chr.7: 122387285-122387285 on Build GRCh38
rs763899111 Chr.7: 122387003-122387003 on Build GRCh38
rs565157773 Chr.7: 122387311-122387311 on Build GRCh38
rs550298942 Chr.7: 122387327-122387327 on Build GRCh38
rs775457034 Chr.7: 122387146-122387146 on Build GRCh38
rs755537025 Chr.7: 122387093-122387093 on Build GRCh38
rs1051023641 Chr.7: 122387235-122387235 on Build GRCh38
rs749194312 Chr.7: 122387052-122387052 on Build GRCh38

View Full Product Details