Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6935647-6936239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFA4
Entrez Gene ID
1669
Gene Name
defensin alpha 4
Gene Aliases
DEF4, HNP-4, HP-4, HP4
UniGene
Hs.591391
Gene Chromosome Location
Chr.8: 6935820-6938338 on Build GRCh38
Transcripts
NM_001925.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1101166 c.294A>G p.*98* Chr.8: 6936020-6936020 on Build GRCh38
1101167 c.289G>A p.D97N Chr.8: 6936025-6936025 on Build GRCh38
3925483 c.182G>A p.R61K Chr.8: 6936132-6936132 on Build GRCh38
1101168 c.226G>T p.E76* Chr.8: 6936088-6936088 on Build GRCh38
6113561 c.209T>A p.V70E Chr.8: 6936105-6936105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778665674 Chr.8: 6936067-6936067 on Build GRCh38
rs948523273 Chr.8: 6935859-6935859 on Build GRCh38
rs976876751 Chr.8: 6935685-6935685 on Build GRCh38
rs764473726 Chr.8: 6936050-6936050 on Build GRCh38
rs367726165 Chr.8: 6936019-6936019 on Build GRCh38
rs749190718 Chr.8: 6935985-6935985 on Build GRCh38
rs754867893 Chr.8: 6936030-6936030 on Build GRCh38
rs779843803 Chr.8: 6935681-6935681 on Build GRCh38
rs975171030 Chr.8: 6935806-6935806 on Build GRCh38
rs1040351833 Chr.8: 6936207-6936207 on Build GRCh38
rs779529290 Chr.8: 6936149-6936149 on Build GRCh38
rs531562489 Chr.8: 6935894-6935894 on Build GRCh38
rs954692723 Chr.8: 6935915-6935915 on Build GRCh38
rs796589153 Chr.8: 6935989-6935989 on Build GRCh38
rs886685265 Chr.8: 6935764-6935764 on Build GRCh38
rs746317759 Chr.8: 6935821-6935821 on Build GRCh38
rs764715890 Chr.8: 6936010-6936010 on Build GRCh38
rs556475956 Chr.8: 6935768-6935768 on Build GRCh38
rs780890509 Chr.8: 6935734-6935734 on Build GRCh38
rs116014310 Chr.8: 6935983-6935983 on Build GRCh38
rs899796798 Chr.8: 6936065-6936065 on Build GRCh38
rs745327485 Chr.8: 6936206-6936206 on Build GRCh38
rs200392892 Chr.8: 6936096-6936096 on Build GRCh38
rs913185101 Chr.8: 6935760-6935760 on Build GRCh38
rs938732403 Chr.8: 6935907-6935907 on Build GRCh38
rs756317418 Chr.8: 6936060-6936060 on Build GRCh38
rs900677712 Chr.8: 6935947-6935947 on Build GRCh38
rs761767369 Chr.8: 6935991-6935991 on Build GRCh38
rs988681906 Chr.8: 6935944-6935944 on Build GRCh38
rs934549081 Chr.8: 6935686-6935686 on Build GRCh38
rs752622006 Chr.8: 6936129-6936129 on Build GRCh38
rs564566496 Chr.8: 6935837-6935837 on Build GRCh38
rs760899482 Chr.8: 6936162-6936162 on Build GRCh38
rs771624264 Chr.8: 6935785-6935785 on Build GRCh38
rs751339382 Chr.8: 6936156-6936156 on Build GRCh38
rs781455661 Chr.8: 6935980-6935980 on Build GRCh38
rs964505901 Chr.8: 6935682-6935682 on Build GRCh38
rs541044947 Chr.8: 6935827-6935827 on Build GRCh38
rs371954556 Chr.8: 6936122-6936122 on Build GRCh38
rs760213234 Chr.8: 6935996-6935996 on Build GRCh38
rs368110185 Chr.8: 6936089-6936089 on Build GRCh38
rs776383257 Chr.8: 6936085-6936085 on Build GRCh38
rs1037927767 Chr.8: 6935935-6935935 on Build GRCh38
rs748865471 Chr.8: 6936152-6936152 on Build GRCh38
rs758199661 Chr.8: 6936071-6936071 on Build GRCh38
rs752817815 Chr.8: 6936176-6936176 on Build GRCh38
rs557438926 Chr.8: 6936007-6936007 on Build GRCh38
rs780005204 Chr.8: 6935984-6935984 on Build GRCh38
rs1003249204 Chr.8: 6935746-6935746 on Build GRCh38
rs746662971 Chr.8: 6936035-6936035 on Build GRCh38
rs139389771 Chr.8: 6936186-6936186 on Build GRCh38
rs757454632 Chr.8: 6935979-6935979 on Build GRCh38
rs1028039850 Chr.8: 6935814-6935814 on Build GRCh38
rs736227 Chr.8: 6936018-6936018 on Build GRCh38
rs763380458 Chr.8: 6936006-6936006 on Build GRCh38
rs370729964 Chr.8: 6935836-6935836 on Build GRCh38
rs772958227 Chr.8: 6936033-6936033 on Build GRCh38
rs766016367 Chr.8: 6935998-6935998 on Build GRCh38
rs568940728 Chr.8: 6935885-6935885 on Build GRCh38
rs559546450 Chr.8: 6936031-6936031 on Build GRCh38
rs765817203 Chr.8: 6935974-6935974 on Build GRCh38
rs770482785 Chr.8: 6936082-6936082 on Build GRCh38
rs749444927 Chr.8: 6936028-6936028 on Build GRCh38
rs768898104 Chr.8: 6935973-6935973 on Build GRCh38
rs527650560 Chr.8: 6935811-6935811 on Build GRCh38
rs770415634 Chr.8: 6936036-6936036 on Build GRCh38
rs766489304 Chr.8: 6936103-6936103 on Build GRCh38
rs572566325 Chr.8: 6936009-6936009 on Build GRCh38
rs576619034 Chr.8: 6935796-6935796 on Build GRCh38
rs372333304 Chr.8: 6936141-6936141 on Build GRCh38
rs751861970 Chr.8: 6936011-6936011 on Build GRCh38
rs146362624 Chr.8: 6936032-6936032 on Build GRCh38
rs773048043 Chr.8: 6935995-6935995 on Build GRCh38
rs568379406 Chr.8: 6935911-6935911 on Build GRCh38
rs781058831 Chr.8: 6936188-6936188 on Build GRCh38
rs1044432007 Chr.8: 6935720-6935720 on Build GRCh38
rs773757509 Chr.8: 6936160-6936160 on Build GRCh38
rs754112985 Chr.8: 6936113-6936113 on Build GRCh38
rs202025875 Chr.8: 6936068-6936068 on Build GRCh38
rs150510521 Chr.8: 6936181-6936181 on Build GRCh38
rs927269225 Chr.8: 6936106-6936106 on Build GRCh38
rs533728297 Chr.8: 6935917-6935917 on Build GRCh38
rs747640060 Chr.8: 6935757-6935757 on Build GRCh38
rs2738100 Chr.8: 6936059-6936059 on Build GRCh38
rs369532654 Chr.8: 6935969-6935969 on Build GRCh38
rs540981590 Chr.8: 6936026-6936026 on Build GRCh38
rs1054208649 Chr.8: 6935694-6935694 on Build GRCh38
rs189429870 Chr.8: 6935666-6935666 on Build GRCh38
rs371076652 Chr.8: 6935960-6935960 on Build GRCh38
rs201568737 Chr.8: 6936000-6936000 on Build GRCh38
rs913132732 Chr.8: 6935957-6935957 on Build GRCh38
rs372897482 Chr.8: 6935748-6935748 on Build GRCh38
rs907877986 Chr.8: 6936086-6936086 on Build GRCh38
rs377182690 Chr.8: 6936069-6936069 on Build GRCh38
rs777721342 Chr.8: 6936074-6936074 on Build GRCh38
rs74947437 Chr.8: 6935882-6935882 on Build GRCh38
rs774005512 Chr.8: 6935990-6935990 on Build GRCh38
rs546330547 Chr.8: 6935744-6935744 on Build GRCh38
rs55977310 Chr.8: 6936193-6936193 on Build GRCh38
rs189271139 Chr.8: 6936094-6936094 on Build GRCh38
rs41396548 Chr.8: 6935853-6935853 on Build GRCh38
rs765363428 Chr.8: 6936124-6936124 on Build GRCh38
rs762433303 Chr.8: 6936093-6936093 on Build GRCh38
rs138031064 Chr.8: 6936097-6936097 on Build GRCh38
rs542102643 Chr.8: 6935797-6935797 on Build GRCh38
rs539448726 Chr.8: 6936203-6936203 on Build GRCh38
rs758394710 Chr.8: 6936130-6936130 on Build GRCh38
rs368348117 Chr.8: 6936119-6936119 on Build GRCh38
rs775987941 Chr.8: 6936043-6936043 on Build GRCh38
rs768442361 Chr.8: 6935988-6935988 on Build GRCh38
rs996304290 Chr.8: 6935956-6935956 on Build GRCh38
rs868137471 Chr.8: 6936039-6936039 on Build GRCh38
rs759422902 Chr.8: 6935974-6935974 on Build GRCh38
rs561916588 Chr.8: 6936174-6936174 on Build GRCh38
rs751364788 Chr.8: 6936135-6936135 on Build GRCh38
rs2702867 Chr.8: 6935800-6935800 on Build GRCh38
rs142741700 Chr.8: 6935717-6935717 on Build GRCh38
rs375331533 Chr.8: 6935838-6935838 on Build GRCh38
rs765263399 Chr.8: 6936030-6936030 on Build GRCh38
rs373531935 Chr.8: 6935976-6935976 on Build GRCh38
rs978634013 Chr.8: 6936105-6936105 on Build GRCh38
rs1016421562 Chr.8: 6935771-6935771 on Build GRCh38
rs952989433 Chr.8: 6935856-6935856 on Build GRCh38
rs893246430 Chr.8: 6935713-6935713 on Build GRCh38
rs769382429 Chr.8: 6936090-6936090 on Build GRCh38
rs377107156 Chr.8: 6936196-6936196 on Build GRCh38
rs1026608896 Chr.8: 6935758-6935758 on Build GRCh38
rs146967029 Chr.8: 6936063-6936063 on Build GRCh38
rs866059785 Chr.8: 6935765-6935765 on Build GRCh38
rs964096444 Chr.8: 6935884-6935884 on Build GRCh38
rs373619174 Chr.8: 6936057-6936057 on Build GRCh38
rs769131579 Chr.8: 6936146-6936146 on Build GRCh38
rs528468114 Chr.8: 6936051-6936051 on Build GRCh38
rs200506506 Chr.8: 6936081-6936081 on Build GRCh38
rs1044462261 Chr.8: 6935868-6935868 on Build GRCh38
rs757120933 Chr.8: 6936136-6936136 on Build GRCh38
rs570472812 Chr.8: 6935965-6935965 on Build GRCh38
rs201104526 Chr.8: 6936025-6936025 on Build GRCh38
rs759149591 Chr.8: 6936044-6936044 on Build GRCh38
rs56066813 Chr.8: 6936194-6936194 on Build GRCh38
rs55699509 Chr.8: 6936054-6936054 on Build GRCh38
rs1056898329 Chr.8: 6935835-6935835 on Build GRCh38
rs914420901 Chr.8: 6935858-6935858 on Build GRCh38
rs568553179 Chr.8: 6935784-6935784 on Build GRCh38
rs771358026 Chr.8: 6936165-6936165 on Build GRCh38
rs758747774 Chr.8: 6935975-6935975 on Build GRCh38
rs765611001 Chr.8: 6936175-6936175 on Build GRCh38
rs745930702 Chr.8: 6935981-6935981 on Build GRCh38
rs111242797 Chr.8: 6936180-6936180 on Build GRCh38
rs755135282 Chr.8: 6936064-6936064 on Build GRCh38
rs761142862 Chr.8: 6936100-6936100 on Build GRCh38
rs548472770 Chr.8: 6935910-6935910 on Build GRCh38
rs771820391 Chr.8: 6935994-6935994 on Build GRCh38
rs368531362 Chr.8: 6935908-6935908 on Build GRCh38

View Full Product Details