Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68742089-68742664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCAR1
Entrez Gene ID
55749
Gene Name
cell division cycle and apoptosis regulator 1
Gene Aliases
-
UniGene
Hs.49853
Gene Chromosome Location
Chr.10: 68721144-68792377 on Build GRCh38
Transcripts
NM_001282959.1 NM_001282960.1 NM_018237.3 NR_104262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4144795 c.480G>A p.T160T Chr.10: 68742531-68742531 on Build GRCh38
7325093 c.325C>A p.P109T Chr.10: 68742376-68742376 on Build GRCh38
465882 c.331G>T p.V111F Chr.10: 68742382-68742382 on Build GRCh38
6621145 c.507C>A p.F169L Chr.10: 68742558-68742558 on Build GRCh38
2159539 c.450C>T p.F150F Chr.10: 68742501-68742501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149791976 Chr.10: 68742508-68742508 on Build GRCh38
rs769641736 Chr.10: 68742329-68742329 on Build GRCh38
rs945136291 Chr.10: 68742183-68742183 on Build GRCh38
rs3740593 Chr.10: 68742153-68742153 on Build GRCh38
rs778590121 Chr.10: 68742520-68742520 on Build GRCh38
rs543161845 Chr.10: 68742481-68742481 on Build GRCh38
rs376008579 Chr.10: 68742369-68742369 on Build GRCh38
rs756107245 Chr.10: 68742482-68742482 on Build GRCh38
rs758462824 Chr.10: 68742454-68742454 on Build GRCh38
rs770537826 Chr.10: 68742516-68742516 on Build GRCh38
rs777578014 Chr.10: 68742336-68742336 on Build GRCh38
rs371513606 Chr.10: 68742460-68742460 on Build GRCh38
rs556061059 Chr.10: 68742164-68742164 on Build GRCh38
rs998361856 Chr.10: 68742310-68742310 on Build GRCh38
rs1007608264 Chr.10: 68742278-68742278 on Build GRCh38
rs914907407 Chr.10: 68742617-68742617 on Build GRCh38
rs529963612 Chr.10: 68742259-68742259 on Build GRCh38
rs770612150 Chr.10: 68742341-68742341 on Build GRCh38
rs888569526 Chr.10: 68742367-68742367 on Build GRCh38
rs73264536 Chr.10: 68742170-68742170 on Build GRCh38
rs775911028 Chr.10: 68742394-68742394 on Build GRCh38
rs921150630 Chr.10: 68742125-68742125 on Build GRCh38
rs1057408829 Chr.10: 68742290-68742290 on Build GRCh38
rs139158551 Chr.10: 68742452-68742452 on Build GRCh38
rs572923192 Chr.10: 68742579-68742579 on Build GRCh38
rs764501065 Chr.10: 68742405-68742405 on Build GRCh38
rs376765384 Chr.10: 68742605-68742605 on Build GRCh38
rs772488840 Chr.10: 68742372-68742372 on Build GRCh38
rs144648961 Chr.10: 68742522-68742522 on Build GRCh38
rs769110001 Chr.10: 68742343-68742347 on Build GRCh38
rs557147054 Chr.10: 68742296-68742296 on Build GRCh38
rs773772414 Chr.10: 68742351-68742351 on Build GRCh38
rs962757338 Chr.10: 68742609-68742609 on Build GRCh38
rs1018545578 Chr.10: 68742282-68742282 on Build GRCh38
rs989583933 Chr.10: 68742606-68742606 on Build GRCh38
rs749077072 Chr.10: 68742509-68742509 on Build GRCh38
rs754202191 Chr.10: 68742408-68742408 on Build GRCh38
rs747985378 Chr.10: 68742326-68742326 on Build GRCh38
rs997950943 Chr.10: 68742458-68742458 on Build GRCh38
rs139860659 Chr.10: 68742564-68742564 on Build GRCh38
rs1012953160 Chr.10: 68742324-68742324 on Build GRCh38
rs566201173 Chr.10: 68742144-68742144 on Build GRCh38
rs770990361 Chr.10: 68742642-68742642 on Build GRCh38
rs1016059536 Chr.10: 68742398-68742398 on Build GRCh38
rs761983547 Chr.10: 68742416-68742416 on Build GRCh38
rs368132401 Chr.10: 68742187-68742187 on Build GRCh38
rs750488056 Chr.10: 68742450-68742450 on Build GRCh38
rs866271098 Chr.10: 68742376-68742376 on Build GRCh38
rs763046876 Chr.10: 68742373-68742373 on Build GRCh38
rs1300253 Chr.10: 68742531-68742531 on Build GRCh38
rs894714219 Chr.10: 68742319-68742319 on Build GRCh38
rs769124552 Chr.10: 68742593-68742593 on Build GRCh38
rs1029826468 Chr.10: 68742328-68742328 on Build GRCh38
rs924804730 Chr.10: 68742190-68742190 on Build GRCh38
rs200363269 Chr.10: 68742435-68742435 on Build GRCh38
rs775662355 Chr.10: 68742358-68742358 on Build GRCh38
rs749005518 Chr.10: 68742339-68742339 on Build GRCh38
rs761041904 Chr.10: 68742400-68742400 on Build GRCh38
rs1028220290 Chr.10: 68742529-68742529 on Build GRCh38

View Full Product Details