Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13710596-13711182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAR1
Entrez Gene ID
84188
Gene Name
fatty acyl-CoA reductase 1
Gene Aliases
MLSTD2, PFCRD, SDR10E1
UniGene
Hs.501991
Gene Chromosome Location
Chr.11: 13668659-13732346 on Build GRCh38
Transcripts
NM_032228.5 XM_011520400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3445579 c.697G>A p.G233S Chr.11: 13710844-13710844 on Build GRCh38
1206527 c.661A>C p.K221Q Chr.11: 13710808-13710808 on Build GRCh38
7301088 c.546G>A p.E182E Chr.11: 13710693-13710693 on Build GRCh38
1289518 c.723+2T>C p.? Chr.11: 13710872-13710872 on Build GRCh38
3791357 c.599G>A p.R200K Chr.11: 13710746-13710746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112294567 Chr.11: 13710952-13710952 on Build GRCh38
rs750654745 Chr.11: 13710789-13710789 on Build GRCh38
rs773907660 Chr.11: 13710707-13710707 on Build GRCh38
rs748761809 Chr.11: 13710688-13710688 on Build GRCh38
rs758755383 Chr.11: 13710796-13710796 on Build GRCh38
rs111747096 Chr.11: 13710705-13710705 on Build GRCh38
rs1054334931 Chr.11: 13711057-13711057 on Build GRCh38
rs745515720 Chr.11: 13710921-13710921 on Build GRCh38
rs149868108 Chr.11: 13710711-13710711 on Build GRCh38
rs1033496002 Chr.11: 13710963-13710963 on Build GRCh38
rs867397990 Chr.11: 13710848-13710848 on Build GRCh38
rs767854733 Chr.11: 13710710-13710710 on Build GRCh38
rs756021546 Chr.11: 13710840-13710840 on Build GRCh38
rs971628903 Chr.11: 13711042-13711042 on Build GRCh38
rs950919647 Chr.11: 13711105-13711105 on Build GRCh38
rs777595479 Chr.11: 13710871-13710871 on Build GRCh38
rs546734964 Chr.11: 13710987-13710987 on Build GRCh38
rs761099444 Chr.11: 13710721-13710721 on Build GRCh38
rs370576330 Chr.11: 13710671-13710671 on Build GRCh38
rs765400630 Chr.11: 13710759-13710759 on Build GRCh38
rs186951895 Chr.11: 13711114-13711114 on Build GRCh38
rs912230897 Chr.11: 13711011-13711011 on Build GRCh38
rs929074516 Chr.11: 13710712-13710712 on Build GRCh38
rs1026502587 Chr.11: 13711087-13711087 on Build GRCh38
rs757310319 Chr.11: 13710756-13710756 on Build GRCh38
rs189664117 Chr.11: 13710625-13710625 on Build GRCh38
rs772712760 Chr.11: 13710684-13710684 on Build GRCh38
rs772819785 Chr.11: 13711138-13711138 on Build GRCh38
rs914017656 Chr.11: 13711064-13711064 on Build GRCh38
rs771776356 Chr.11: 13711051-13711051 on Build GRCh38
rs759234971 Chr.11: 13710708-13710708 on Build GRCh38
rs933175761 Chr.11: 13710902-13710902 on Build GRCh38
rs556172857 Chr.11: 13710940-13710940 on Build GRCh38
rs368878955 Chr.11: 13710726-13710726 on Build GRCh38
rs375397112 Chr.11: 13710675-13710675 on Build GRCh38
rs182336044 Chr.11: 13710702-13710702 on Build GRCh38
rs995707095 Chr.11: 13710980-13710980 on Build GRCh38
rs371856810 Chr.11: 13710644-13710644 on Build GRCh38
rs371311197 Chr.11: 13710813-13710813 on Build GRCh38
rs1012867266 Chr.11: 13710998-13710998 on Build GRCh38
rs945444859 Chr.11: 13711067-13711067 on Build GRCh38
rs748793320 Chr.11: 13710900-13710900 on Build GRCh38
rs921477200 Chr.11: 13711048-13711048 on Build GRCh38
rs773967374 Chr.11: 13710919-13710919 on Build GRCh38
rs746757255 Chr.11: 13710645-13710645 on Build GRCh38
rs764518716 Chr.11: 13710725-13710725 on Build GRCh38
rs1002789464 Chr.11: 13710760-13710760 on Build GRCh38
rs747962795 Chr.11: 13710828-13710828 on Build GRCh38
rs964416593 Chr.11: 13711116-13711116 on Build GRCh38
rs922900322 Chr.11: 13711151-13711151 on Build GRCh38
rs748089623 Chr.11: 13710668-13710668 on Build GRCh38
rs529502099 Chr.11: 13710659-13710659 on Build GRCh38
rs1038994389 Chr.11: 13710880-13710880 on Build GRCh38
rs984585897 Chr.11: 13711113-13711113 on Build GRCh38
rs9787838 Chr.11: 13711082-13711082 on Build GRCh38
rs768543557 Chr.11: 13710646-13710646 on Build GRCh38
rs747643001 Chr.11: 13710901-13710901 on Build GRCh38
rs192506655 Chr.11: 13711140-13711140 on Build GRCh38
rs936802060 Chr.11: 13710665-13710665 on Build GRCh38
rs936874476 Chr.11: 13711052-13711052 on Build GRCh38
rs528318391 Chr.11: 13710953-13710953 on Build GRCh38
rs965654774 Chr.11: 13711040-13711040 on Build GRCh38
rs1034629442 Chr.11: 13710915-13710915 on Build GRCh38
rs143433267 Chr.11: 13711010-13711010 on Build GRCh38
rs148703026 Chr.11: 13710971-13710971 on Build GRCh38
rs893159647 Chr.11: 13710989-13710989 on Build GRCh38

View Full Product Details