Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64230144-64230666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC4
Entrez Gene ID
3338
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
Gene Aliases
DANJC4, HSPF2, MCG18
UniGene
Hs.172847
Gene Chromosome Location
Chr.11: 64230281-64234281 on Build GRCh38
Transcripts
NM_001307980.1 NM_001307981.1 NM_005528.3

SNP Detail

SNP ID Chromosome Location
rs374783887 Chr.11: 64230533-64230533 on Build GRCh38
rs908889598 Chr.11: 64230266-64230266 on Build GRCh38
rs778694477 Chr.11: 64230410-64230410 on Build GRCh38
rs1051222620 Chr.11: 64230581-64230581 on Build GRCh38
rs971803524 Chr.11: 64230547-64230547 on Build GRCh38
rs952586280 Chr.11: 64230208-64230208 on Build GRCh38
rs763127418 Chr.11: 64230632-64230632 on Build GRCh38
rs1013076008 Chr.11: 64230481-64230481 on Build GRCh38
rs58040631 Chr.11: 64230502-64230502 on Build GRCh38
rs890006443 Chr.11: 64230319-64230319 on Build GRCh38
rs544872892 Chr.11: 64230358-64230358 on Build GRCh38
rs934184827 Chr.11: 64230308-64230308 on Build GRCh38
rs946874199 Chr.11: 64230391-64230391 on Build GRCh38
rs573626753 Chr.11: 64230636-64230636 on Build GRCh38
rs1015442588 Chr.11: 64230530-64230530 on Build GRCh38
rs766532534 Chr.11: 64230194-64230194 on Build GRCh38
rs575903876 Chr.11: 64230351-64230351 on Build GRCh38
rs557832609 Chr.11: 64230367-64230367 on Build GRCh38
rs984434697 Chr.11: 64230264-64230264 on Build GRCh38
rs927248716 Chr.11: 64230232-64230232 on Build GRCh38
rs542826138 Chr.11: 64230640-64230640 on Build GRCh38
rs940423485 Chr.11: 64230274-64230274 on Build GRCh38
rs183362657 Chr.11: 64230296-64230296 on Build GRCh38
rs76497858 Chr.11: 64230277-64230277 on Build GRCh38
rs892840487 Chr.11: 64230406-64230406 on Build GRCh38
rs1010304240 Chr.11: 64230356-64230356 on Build GRCh38
rs750928306 Chr.11: 64230432-64230432 on Build GRCh38
rs116750851 Chr.11: 64230298-64230298 on Build GRCh38
rs777388549 Chr.11: 64230410-64230411 on Build GRCh38
rs775214653 Chr.11: 64230592-64230592 on Build GRCh38
rs746568975 Chr.11: 64230513-64230513 on Build GRCh38
rs922653731 Chr.11: 64230268-64230268 on Build GRCh38
rs745804637 Chr.11: 64230565-64230565 on Build GRCh38
rs981322671 Chr.11: 64230230-64230230 on Build GRCh38
rs770588282 Chr.11: 64230539-64230539 on Build GRCh38
rs747790642 Chr.11: 64230411-64230412 on Build GRCh38
rs902783022 Chr.11: 64230456-64230456 on Build GRCh38
rs1034149230 Chr.11: 64230493-64230493 on Build GRCh38
rs961330957 Chr.11: 64230257-64230257 on Build GRCh38
rs398016372 Chr.11: 64230423-64230423 on Build GRCh38
rs924135394 Chr.11: 64230646-64230646 on Build GRCh38
rs560171950 Chr.11: 64230561-64230561 on Build GRCh38
rs762281280 Chr.11: 64230392-64230392 on Build GRCh38
rs1020062957 Chr.11: 64230176-64230176 on Build GRCh38
rs1020366673 Chr.11: 64230372-64230372 on Build GRCh38
rs772919063 Chr.11: 64230213-64230213 on Build GRCh38
rs769617741 Chr.11: 64230566-64230566 on Build GRCh38
rs752385613 Chr.11: 64230485-64230485 on Build GRCh38
rs763216613 Chr.11: 64230324-64230324 on Build GRCh38
rs746940886 Chr.11: 64230217-64230217 on Build GRCh38
rs933062794 Chr.11: 64230568-64230568 on Build GRCh38
rs755511267 Chr.11: 64230340-64230340 on Build GRCh38
rs1002629045 Chr.11: 64230473-64230473 on Build GRCh38
rs1055140494 Chr.11: 64230600-64230600 on Build GRCh38
rs1022673490 Chr.11: 64230497-64230497 on Build GRCh38
rs1007554227 Chr.11: 64230335-64230335 on Build GRCh38
rs11311686 Chr.11: 64230411-64230411 on Build GRCh38
rs1054435243 Chr.11: 64230337-64230337 on Build GRCh38
rs771033854 Chr.11: 64230234-64230234 on Build GRCh38
rs780300125 Chr.11: 64230457-64230457 on Build GRCh38
rs781116215 Chr.11: 64230546-64230546 on Build GRCh38
rs540365500 Chr.11: 64230440-64230440 on Build GRCh38
rs776268371 Chr.11: 64230286-64230286 on Build GRCh38
rs971229704 Chr.11: 64230191-64230191 on Build GRCh38
rs532741967 Chr.11: 64230618-64230618 on Build GRCh38
rs759328057 Chr.11: 64230295-64230295 on Build GRCh38
rs1003822218 Chr.11: 64230471-64230471 on Build GRCh38
rs774268184 Chr.11: 64230626-64230626 on Build GRCh38
rs767959102 Chr.11: 64230401-64230401 on Build GRCh38
rs1051338336 Chr.11: 64230318-64230318 on Build GRCh38
rs768728981 Chr.11: 64230529-64230529 on Build GRCh38
rs577705130 Chr.11: 64230373-64230373 on Build GRCh38
rs751676296 Chr.11: 64230382-64230382 on Build GRCh38
rs577457546 Chr.11: 64230172-64230172 on Build GRCh38
rs796658523 Chr.11: 64230256-64230256 on Build GRCh38
rs1005355150 Chr.11: 64230525-64230525 on Build GRCh38
rs777386609 Chr.11: 64230505-64230505 on Build GRCh38
rs764153571 Chr.11: 64230371-64230371 on Build GRCh38
rs908863933 Chr.11: 64230544-64230544 on Build GRCh38
rs758191893 Chr.11: 64230492-64230492 on Build GRCh38
rs1042559733 Chr.11: 64230616-64230616 on Build GRCh38
rs1010687890 Chr.11: 64230621-64230621 on Build GRCh38
rs985913036 Chr.11: 64230582-64230582 on Build GRCh38
rs367799311 Chr.11: 64230506-64230506 on Build GRCh38
rs911408754 Chr.11: 64230586-64230586 on Build GRCh38
rs952514064 Chr.11: 64230501-64230501 on Build GRCh38
rs983964609 Chr.11: 64230236-64230236 on Build GRCh38
rs1023211053 Chr.11: 64230205-64230205 on Build GRCh38
rs958625382 Chr.11: 64230520-64230520 on Build GRCh38

View Full Product Details