Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40514327-40514867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXO5
Entrez Gene ID
64789
Gene Name
exonuclease 5
Gene Aliases
C1orf176, DEM1, Exo V, hExo5
UniGene
Hs.59584
Gene Chromosome Location
Chr.1: 40508719-40516556 on Build GRCh38
Transcripts
NM_022774.1 XM_005271125.3 XM_005271126.3 XM_005271128.3 XM_011541970.2 XM_017002099.1 XM_017002100.1 XM_017002101.1 XM_017002102.1 XM_017002103.1 XM_017002104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
464618 c.43T>G p.S15A Chr.1: 40514587-40514587 on Build GRCh38
146484 c.301G>A p.E101K Chr.1: 40514845-40514845 on Build GRCh38
1687517 c.151C>T p.L51F Chr.1: 40514695-40514695 on Build GRCh38
2169259 c.189G>A p.W63* Chr.1: 40514733-40514733 on Build GRCh38
909464 c.89A>G p.D30G Chr.1: 40514633-40514633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745683025 Chr.1: 40514658-40514658 on Build GRCh38
rs368964061 Chr.1: 40514599-40514599 on Build GRCh38
rs965565347 Chr.1: 40514770-40514770 on Build GRCh38
rs953417198 Chr.1: 40514691-40514691 on Build GRCh38
rs755436405 Chr.1: 40514570-40514570 on Build GRCh38
rs1048166246 Chr.1: 40514482-40514482 on Build GRCh38
rs745740081 Chr.1: 40514525-40514525 on Build GRCh38
rs764901873 Chr.1: 40514760-40514760 on Build GRCh38
rs774647599 Chr.1: 40514761-40514761 on Build GRCh38
rs778780840 Chr.1: 40514656-40514656 on Build GRCh38
rs991498483 Chr.1: 40514538-40514538 on Build GRCh38
rs74330144 Chr.1: 40514637-40514637 on Build GRCh38
rs143920251 Chr.1: 40514636-40514637 on Build GRCh38
rs764019469 Chr.1: 40514788-40514788 on Build GRCh38
rs373218757 Chr.1: 40514659-40514659 on Build GRCh38
rs757641363 Chr.1: 40514556-40514560 on Build GRCh38
rs929628099 Chr.1: 40514427-40514427 on Build GRCh38
rs757564216 Chr.1: 40514612-40514612 on Build GRCh38
rs746416501 Chr.1: 40514786-40514787 on Build GRCh38
rs566618397 Chr.1: 40514415-40514415 on Build GRCh38
rs775019851 Chr.1: 40514535-40514535 on Build GRCh38
rs746528535 Chr.1: 40514678-40514678 on Build GRCh38
rs114551256 Chr.1: 40514554-40514554 on Build GRCh38
rs1000165673 Chr.1: 40514646-40514646 on Build GRCh38
rs888161463 Chr.1: 40514486-40514486 on Build GRCh38
rs1001659184 Chr.1: 40514489-40514489 on Build GRCh38
rs749060437 Chr.1: 40514505-40514505 on Build GRCh38
rs781624040 Chr.1: 40514576-40514576 on Build GRCh38
rs763776173 Chr.1: 40514539-40514539 on Build GRCh38
rs1011746404 Chr.1: 40514416-40514418 on Build GRCh38
rs780031828 Chr.1: 40514838-40514838 on Build GRCh38
rs749574908 Chr.1: 40514606-40514606 on Build GRCh38
rs942087109 Chr.1: 40514565-40514565 on Build GRCh38
rs779328840 Chr.1: 40514815-40514815 on Build GRCh38
rs749580296 Chr.1: 40514556-40514557 on Build GRCh38
rs772137317 Chr.1: 40514530-40514530 on Build GRCh38
rs768109747 Chr.1: 40514679-40514679 on Build GRCh38
rs895959259 Chr.1: 40514512-40514512 on Build GRCh38
rs559869905 Chr.1: 40514409-40514409 on Build GRCh38
rs376563843 Chr.1: 40514772-40514772 on Build GRCh38
rs757512390 Chr.1: 40514806-40514806 on Build GRCh38
rs918491703 Chr.1: 40514787-40514787 on Build GRCh38
rs775333739 Chr.1: 40514666-40514666 on Build GRCh38
rs201053995 Chr.1: 40514706-40514706 on Build GRCh38
rs750215357 Chr.1: 40514819-40514819 on Build GRCh38
rs746864844 Chr.1: 40514844-40514844 on Build GRCh38
rs200406212 Chr.1: 40514762-40514762 on Build GRCh38
rs374827341 Chr.1: 40514581-40514581 on Build GRCh38
rs760791949 Chr.1: 40514777-40514777 on Build GRCh38
rs778048591 Chr.1: 40514600-40514600 on Build GRCh38
rs562627911 Chr.1: 40514701-40514701 on Build GRCh38
rs751927703 Chr.1: 40514566-40514566 on Build GRCh38
rs766674662 Chr.1: 40514555-40514555 on Build GRCh38
rs58904273 Chr.1: 40514637-40514638 on Build GRCh38
rs967195381 Chr.1: 40514473-40514473 on Build GRCh38
rs911084576 Chr.1: 40514742-40514742 on Build GRCh38
rs753968355 Chr.1: 40514793-40514793 on Build GRCh38
rs202019457 Chr.1: 40514537-40514537 on Build GRCh38
rs867517703 Chr.1: 40514681-40514681 on Build GRCh38
rs911146079 Chr.1: 40514394-40514394 on Build GRCh38
rs199704407 Chr.1: 40514741-40514741 on Build GRCh38
rs866377423 Chr.1: 40514694-40514694 on Build GRCh38
rs758198157 Chr.1: 40514830-40514830 on Build GRCh38
rs764764789 Chr.1: 40514514-40514514 on Build GRCh38
rs896840440 Chr.1: 40514426-40514426 on Build GRCh38
rs761391346 Chr.1: 40514714-40514714 on Build GRCh38
rs763290175 Chr.1: 40514547-40514547 on Build GRCh38
rs752798781 Chr.1: 40514590-40514590 on Build GRCh38
rs770913050 Chr.1: 40514510-40514510 on Build GRCh38
rs776213369 Chr.1: 40514543-40514543 on Build GRCh38

View Full Product Details