Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3562096-3562573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRN
Entrez Gene ID
8455
Gene Name
attractin
Gene Aliases
DPPT-L, MGCA
UniGene
Hs.276252
Gene Chromosome Location
Chr.20: 3471018-3651122 on Build GRCh38
Transcripts
NM_001207047.2 NM_001323332.1 NM_139321.2 NM_139322.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6608513 c.1606C>T p.R536* Chr.20: 3562434-3562434 on Build GRCh38
6608512 c.1606C>T p.R536* Chr.20: 3562434-3562434 on Build GRCh38
2759953 c.1522G>A p.D508N Chr.20: 3562350-3562350 on Build GRCh38
7168247 c.1607G>A p.R536Q Chr.20: 3562435-3562435 on Build GRCh38
6608515 c.1585C>T p.R529W Chr.20: 3562413-3562413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766121603 Chr.20: 3562349-3562349 on Build GRCh38
rs963474947 Chr.20: 3562427-3562427 on Build GRCh38
rs780078684 Chr.20: 3562254-3562254 on Build GRCh38
rs748641685 Chr.20: 3562452-3562452 on Build GRCh38
rs375088927 Chr.20: 3562476-3562476 on Build GRCh38
rs116013056 Chr.20: 3562512-3562512 on Build GRCh38
rs6115943 Chr.20: 3562130-3562130 on Build GRCh38
rs894198201 Chr.20: 3562164-3562164 on Build GRCh38
rs759140019 Chr.20: 3562474-3562474 on Build GRCh38
rs986623804 Chr.20: 3562518-3562518 on Build GRCh38
rs528145458 Chr.20: 3562467-3562467 on Build GRCh38
rs753896417 Chr.20: 3562254-3562255 on Build GRCh38
rs955229791 Chr.20: 3562515-3562515 on Build GRCh38
rs766403198 Chr.20: 3562492-3562492 on Build GRCh38
rs1048414871 Chr.20: 3562411-3562411 on Build GRCh38
rs774874714 Chr.20: 3562327-3562327 on Build GRCh38
rs749496294 Chr.20: 3562435-3562435 on Build GRCh38
rs185855121 Chr.20: 3562332-3562332 on Build GRCh38
rs752587387 Chr.20: 3562278-3562278 on Build GRCh38
rs370113491 Chr.20: 3562235-3562235 on Build GRCh38
rs569482622 Chr.20: 3562367-3562367 on Build GRCh38
rs376147226 Chr.20: 3562236-3562236 on Build GRCh38
rs755331623 Chr.20: 3562438-3562438 on Build GRCh38
rs543121309 Chr.20: 3562503-3562503 on Build GRCh38
rs763952829 Chr.20: 3562484-3562484 on Build GRCh38
rs769611586 Chr.20: 3562475-3562475 on Build GRCh38
rs760763447 Chr.20: 3562488-3562488 on Build GRCh38
rs759313901 Chr.20: 3562366-3562366 on Build GRCh38
rs1042923541 Chr.20: 3562168-3562168 on Build GRCh38
rs767574295 Chr.20: 3562414-3562414 on Build GRCh38
rs748177806 Chr.20: 3562534-3562534 on Build GRCh38
rs749600242 Chr.20: 3562315-3562315 on Build GRCh38
rs755991653 Chr.20: 3562248-3562253 on Build GRCh38
rs149091209 Chr.20: 3562350-3562350 on Build GRCh38
rs765151181 Chr.20: 3562248-3562248 on Build GRCh38
rs574277715 Chr.20: 3562216-3562216 on Build GRCh38
rs753175761 Chr.20: 3562505-3562505 on Build GRCh38
rs755201780 Chr.20: 3562501-3562501 on Build GRCh38
rs147007493 Chr.20: 3562453-3562453 on Build GRCh38
rs201686279 Chr.20: 3562293-3562293 on Build GRCh38
rs143113044 Chr.20: 3562373-3562373 on Build GRCh38
rs200211700 Chr.20: 3562485-3562485 on Build GRCh38
rs748698763 Chr.20: 3562328-3562328 on Build GRCh38
rs754061976 Chr.20: 3562496-3562496 on Build GRCh38
rs762873703 Chr.20: 3562479-3562479 on Build GRCh38
rs541750840 Chr.20: 3562286-3562286 on Build GRCh38
rs41279368 Chr.20: 3562432-3562432 on Build GRCh38
rs942377993 Chr.20: 3562335-3562335 on Build GRCh38
rs746448500 Chr.20: 3562466-3562466 on Build GRCh38
rs780281720 Chr.20: 3562312-3562312 on Build GRCh38
rs934471726 Chr.20: 3562507-3562507 on Build GRCh38
rs6115944 Chr.20: 3562174-3562174 on Build GRCh38
rs377718503 Chr.20: 3562301-3562301 on Build GRCh38
rs140193658 Chr.20: 3562211-3562211 on Build GRCh38
rs138182554 Chr.20: 3562307-3562307 on Build GRCh38
rs772896328 Chr.20: 3562334-3562334 on Build GRCh38
rs967996054 Chr.20: 3562299-3562299 on Build GRCh38
rs1022316220 Chr.20: 3562491-3562491 on Build GRCh38
rs142929276 Chr.20: 3562439-3562439 on Build GRCh38
rs974204378 Chr.20: 3562549-3562549 on Build GRCh38
rs963831090 Chr.20: 3562536-3562536 on Build GRCh38
rs758381844 Chr.20: 3562379-3562379 on Build GRCh38
rs750572237 Chr.20: 3562421-3562421 on Build GRCh38
rs534840880 Chr.20: 3562196-3562196 on Build GRCh38
rs780265846 Chr.20: 3562433-3562433 on Build GRCh38
rs752637372 Chr.20: 3562378-3562378 on Build GRCh38
rs781291839 Chr.20: 3562287-3562287 on Build GRCh38
rs1035051937 Chr.20: 3562177-3562177 on Build GRCh38
rs953576224 Chr.20: 3562364-3562364 on Build GRCh38
rs772843318 Chr.20: 3562462-3562462 on Build GRCh38
rs1030441549 Chr.20: 3562352-3562352 on Build GRCh38
rs776295756 Chr.20: 3562468-3562468 on Build GRCh38
rs895421915 Chr.20: 3562419-3562419 on Build GRCh38
rs200888866 Chr.20: 3562326-3562326 on Build GRCh38
rs986153241 Chr.20: 3562370-3562370 on Build GRCh38
rs758834796 Chr.20: 3562511-3562511 on Build GRCh38
rs562300171 Chr.20: 3562527-3562527 on Build GRCh38
rs566810821 Chr.20: 3562166-3562166 on Build GRCh38
rs904330962 Chr.20: 3562200-3562200 on Build GRCh38

View Full Product Details