Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36755487-36756055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSN1
Entrez Gene ID
79980
Gene Name
DSN1 homolog, MIS12 kinetochore complex component
Gene Aliases
C20orf172, KNL3, MIS13, dJ469A13.2, hKNL-3
UniGene
Hs.632268
Gene Chromosome Location
Chr.20: 36751791-36773827 on Build GRCh38
Transcripts
NM_001145315.1 NM_001145316.1 NM_001145317.1 NM_001145318.1 NM_024918.3 XM_006723876.2 XM_017028070.1 XM_017028071.1 XM_017028072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4823014 c.841C>A p.Q281K Chr.20: 36755714-36755714 on Build GRCh38
4164697 c.873G>T p.V291V Chr.20: 36755682-36755682 on Build GRCh38
6093250 c.823C>T p.Q275* Chr.20: 36755732-36755732 on Build GRCh38
4172257 c.838A>T p.N280Y Chr.20: 36755717-36755717 on Build GRCh38
5652581 c.783delG p.S262fs*30 Chr.20: 36755772-36755772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754677415 Chr.20: 36755944-36755944 on Build GRCh38
rs557839882 Chr.20: 36755576-36755576 on Build GRCh38
rs747349348 Chr.20: 36755879-36755879 on Build GRCh38
rs1028675799 Chr.20: 36755939-36755939 on Build GRCh38
rs113146610 Chr.20: 36755996-36755996 on Build GRCh38
rs757454963 Chr.20: 36755680-36755680 on Build GRCh38
rs944537514 Chr.20: 36755981-36755981 on Build GRCh38
rs562431667 Chr.20: 36755970-36755970 on Build GRCh38
rs748974994 Chr.20: 36755938-36755938 on Build GRCh38
rs772116545 Chr.20: 36756008-36756008 on Build GRCh38
rs751563183 Chr.20: 36755809-36755823 on Build GRCh38
rs376610938 Chr.20: 36755844-36755844 on Build GRCh38
rs756034281 Chr.20: 36755660-36755660 on Build GRCh38
rs920169062 Chr.20: 36755720-36755720 on Build GRCh38
rs1002433550 Chr.20: 36755756-36755756 on Build GRCh38
rs776981207 Chr.20: 36755695-36755698 on Build GRCh38
rs763993322 Chr.20: 36755804-36755804 on Build GRCh38
rs746199909 Chr.20: 36755695-36755695 on Build GRCh38
rs780400708 Chr.20: 36755854-36755854 on Build GRCh38
rs1027936719 Chr.20: 36755676-36755676 on Build GRCh38
rs191089619 Chr.20: 36755770-36755770 on Build GRCh38
rs779084103 Chr.20: 36755836-36755836 on Build GRCh38
rs888920962 Chr.20: 36755608-36755608 on Build GRCh38
rs756718324 Chr.20: 36755817-36755817 on Build GRCh38
rs368110726 Chr.20: 36755659-36755659 on Build GRCh38
rs141615184 Chr.20: 36755822-36755822 on Build GRCh38
rs2425273 Chr.20: 36755986-36755986 on Build GRCh38
rs974472767 Chr.20: 36755971-36755974 on Build GRCh38
rs372365736 Chr.20: 36755669-36755669 on Build GRCh38
rs1055792842 Chr.20: 36755760-36755760 on Build GRCh38
rs1008654010 Chr.20: 36755617-36755617 on Build GRCh38
rs551239398 Chr.20: 36755983-36755983 on Build GRCh38
rs762893193 Chr.20: 36755700-36755701 on Build GRCh38
rs1015668242 Chr.20: 36755964-36755965 on Build GRCh38
rs775740368 Chr.20: 36755716-36755716 on Build GRCh38
rs777839173 Chr.20: 36755663-36755663 on Build GRCh38
rs998460820 Chr.20: 36755824-36755824 on Build GRCh38
rs752436319 Chr.20: 36755795-36755795 on Build GRCh38
rs200710679 Chr.20: 36755777-36755777 on Build GRCh38
rs533161829 Chr.20: 36755974-36755974 on Build GRCh38
rs758650741 Chr.20: 36755849-36755849 on Build GRCh38
rs966709549 Chr.20: 36755933-36755933 on Build GRCh38
rs775448911 Chr.20: 36756004-36756004 on Build GRCh38
rs955194005 Chr.20: 36755520-36755523 on Build GRCh38
rs772723090 Chr.20: 36755739-36755739 on Build GRCh38
rs544146033 Chr.20: 36755932-36755932 on Build GRCh38
rs183801856 Chr.20: 36755931-36755931 on Build GRCh38
rs150788766 Chr.20: 36755521-36755524 on Build GRCh38
rs961120907 Chr.20: 36755772-36755772 on Build GRCh38
rs1048777640 Chr.20: 36755997-36755997 on Build GRCh38
rs1054159969 Chr.20: 36755908-36755908 on Build GRCh38
rs772474350 Chr.20: 36755714-36755714 on Build GRCh38
rs761597086 Chr.20: 36755878-36755878 on Build GRCh38
rs754890661 Chr.20: 36755649-36755649 on Build GRCh38
rs760600951 Chr.20: 36755800-36755800 on Build GRCh38
rs759280569 Chr.20: 36755782-36755782 on Build GRCh38
rs757366862 Chr.20: 36755831-36755831 on Build GRCh38
rs935862349 Chr.20: 36755542-36755542 on Build GRCh38
rs935930772 Chr.20: 36755947-36755947 on Build GRCh38
rs769049955 Chr.20: 36755737-36755737 on Build GRCh38
rs996735818 Chr.20: 36755929-36755929 on Build GRCh38
rs1050241847 Chr.20: 36755600-36755600 on Build GRCh38
rs201203099 Chr.20: 36755635-36755635 on Build GRCh38
rs763939518 Chr.20: 36755734-36755735 on Build GRCh38
rs750009303 Chr.20: 36755631-36755631 on Build GRCh38
rs767299325 Chr.20: 36755785-36755785 on Build GRCh38
rs955162507 Chr.20: 36755957-36755957 on Build GRCh38
rs892798857 Chr.20: 36755912-36755912 on Build GRCh38
rs866381419 Chr.20: 36756033-36756033 on Build GRCh38
rs145774066 Chr.20: 36755763-36755763 on Build GRCh38
rs993154619 Chr.20: 36755674-36755674 on Build GRCh38
rs779172449 Chr.20: 36755687-36755687 on Build GRCh38
rs868210221 Chr.20: 36755548-36755548 on Build GRCh38
rs767330643 Chr.20: 36755657-36755657 on Build GRCh38
rs747511912 Chr.20: 36755732-36755732 on Build GRCh38
rs1008092493 Chr.20: 36755963-36755964 on Build GRCh38

View Full Product Details