Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99173372-99173967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL11B
Entrez Gene ID
64919
Gene Name
B-cell CLL/lymphoma 11B
Gene Aliases
ATL1, ATL1-alpha, ATL1-beta, ATL1-delta, ATL1-gamma, CTIP-2, CTIP2, RIT1, ZNF856B, hRIT1-alpha
UniGene
Hs.709690
Gene Chromosome Location
Chr.14: 99169287-99271713 on Build GRCh38
Transcripts
NM_001282237.1 NM_001282238.1 NM_022898.2 NM_138576.3

SNP Detail

SNP ID Chromosome Location
rs1009832279 Chr.14: 99173409-99173409 on Build GRCh38
rs144498557 Chr.14: 99173805-99173806 on Build GRCh38
rs982738875 Chr.14: 99173517-99173517 on Build GRCh38
rs397959537 Chr.14: 99173587-99173588 on Build GRCh38
rs199876731 Chr.14: 99173567-99173567 on Build GRCh38
rs181198466 Chr.14: 99173507-99173507 on Build GRCh38
rs1004486411 Chr.14: 99173872-99173872 on Build GRCh38
rs1037926023 Chr.14: 99173864-99173865 on Build GRCh38
rs756925446 Chr.14: 99173729-99173729 on Build GRCh38
rs1027211827 Chr.14: 99173835-99173835 on Build GRCh38
rs927927561 Chr.14: 99173810-99173810 on Build GRCh38
rs890738032 Chr.14: 99173736-99173736 on Build GRCh38
rs990036456 Chr.14: 99173743-99173743 on Build GRCh38
rs59537098 Chr.14: 99173554-99173555 on Build GRCh38
rs1015506831 Chr.14: 99173893-99173893 on Build GRCh38
rs943524236 Chr.14: 99173467-99173467 on Build GRCh38
rs948690578 Chr.14: 99173847-99173847 on Build GRCh38
rs200960315 Chr.14: 99173554-99173554 on Build GRCh38
rs552057176 Chr.14: 99173593-99173593 on Build GRCh38
rs945504217 Chr.14: 99173539-99173539 on Build GRCh38
rs372531102 Chr.14: 99173613-99173624 on Build GRCh38
rs918953831 Chr.14: 99173422-99173422 on Build GRCh38
rs987757845 Chr.14: 99173454-99173454 on Build GRCh38
rs754614912 Chr.14: 99173592-99173592 on Build GRCh38
rs938103037 Chr.14: 99173636-99173636 on Build GRCh38
rs867036983 Chr.14: 99173578-99173578 on Build GRCh38
rs1042514848 Chr.14: 99173806-99173806 on Build GRCh38
rs954401390 Chr.14: 99173712-99173712 on Build GRCh38
rs915428823 Chr.14: 99173723-99173723 on Build GRCh38
rs1056980937 Chr.14: 99173601-99173604 on Build GRCh38
rs982147074 Chr.14: 99173586-99173586 on Build GRCh38
rs1053982213 Chr.14: 99173403-99173403 on Build GRCh38
rs200395137 Chr.14: 99173575-99173576 on Build GRCh38
rs796546180 Chr.14: 99173619-99173619 on Build GRCh38
rs939364613 Chr.14: 99173813-99173816 on Build GRCh38
rs201962171 Chr.14: 99173565-99173565 on Build GRCh38
rs1007900314 Chr.14: 99173452-99173452 on Build GRCh38
rs1019317463 Chr.14: 99173458-99173458 on Build GRCh38
rs753462891 Chr.14: 99173713-99173713 on Build GRCh38
rs894280889 Chr.14: 99173614-99173614 on Build GRCh38
rs996567460 Chr.14: 99173833-99173833 on Build GRCh38
rs575405928 Chr.14: 99173453-99173453 on Build GRCh38
rs200160903 Chr.14: 99173577-99173577 on Build GRCh38
rs141214295 Chr.14: 99173449-99173449 on Build GRCh38
rs533505255 Chr.14: 99173587-99173587 on Build GRCh38
rs1040713461 Chr.14: 99173776-99173776 on Build GRCh38
rs1007848112 Chr.14: 99173768-99173768 on Build GRCh38
rs1026804786 Chr.14: 99173504-99173504 on Build GRCh38
rs899581313 Chr.14: 99173777-99173777 on Build GRCh38
rs945442423 Chr.14: 99173806-99173806 on Build GRCh38
rs1008585010 Chr.14: 99173915-99173919 on Build GRCh38
rs371982162 Chr.14: 99173552-99173552 on Build GRCh38
rs1002741881 Chr.14: 99173599-99173601 on Build GRCh38
rs764788704 Chr.14: 99173906-99173906 on Build GRCh38
rs996616255 Chr.14: 99173488-99173488 on Build GRCh38
rs993290747 Chr.14: 99173641-99173641 on Build GRCh38
rs987687617 Chr.14: 99173719-99173720 on Build GRCh38
rs1041751274 Chr.14: 99173433-99173433 on Build GRCh38
rs34800317 Chr.14: 99173577-99173578 on Build GRCh38
rs970745071 Chr.14: 99173584-99173584 on Build GRCh38
rs563792353 Chr.14: 99173638-99173638 on Build GRCh38
rs775027012 Chr.14: 99173466-99173466 on Build GRCh38
rs934680914 Chr.14: 99173537-99173537 on Build GRCh38
rs945579306 Chr.14: 99173725-99173725 on Build GRCh38
rs1012719240 Chr.14: 99173617-99173617 on Build GRCh38
rs572582602 Chr.14: 99173536-99173536 on Build GRCh38
rs531153753 Chr.14: 99173789-99173789 on Build GRCh38
rs899031999 Chr.14: 99173869-99173871 on Build GRCh38
rs952580184 Chr.14: 99173870-99173870 on Build GRCh38
rs894033333 Chr.14: 99173406-99173406 on Build GRCh38
rs397949117 Chr.14: 99173564-99173565 on Build GRCh38
rs1015994739 Chr.14: 99173627-99173627 on Build GRCh38
rs569199941 Chr.14: 99173520-99173520 on Build GRCh38
rs767786252 Chr.14: 99173653-99173653 on Build GRCh38
rs560673191 Chr.14: 99173531-99173531 on Build GRCh38

View Full Product Details