Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99510762-99511323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC85C
Entrez Gene ID
317762
Gene Name
coiled-coil domain containing 85C
Gene Aliases
-
UniGene
Hs.709288
Gene Chromosome Location
Chr.14: 99511266-99604390 on Build GRCh38
Transcripts
NM_001144995.1 NM_001099402.1 XM_005268154.4 XM_011537275.2

Gene Symbol
CCNK
Entrez Gene ID
8812
Gene Name
cyclin K
Gene Aliases
CPR4
UniGene
Hs.510409
Gene Chromosome Location
Chr.14: 99481402-99511515 on Build GRCh38
Transcripts
NM_001144995.1 NM_001099402.1 XM_005268154.4 XM_011537275.2

SNP Detail

SNP ID Chromosome Location
rs1057500571 Chr.14: 99510794-99510794 on Build GRCh38
rs969107026 Chr.14: 99511232-99511232 on Build GRCh38
rs916087748 Chr.14: 99511298-99511298 on Build GRCh38
rs968779718 Chr.14: 99510962-99510962 on Build GRCh38
rs894761881 Chr.14: 99510801-99510801 on Build GRCh38
rs916140110 Chr.14: 99511017-99511017 on Build GRCh38
rs956299340 Chr.14: 99510958-99510958 on Build GRCh38
rs542787565 Chr.14: 99510783-99510783 on Build GRCh38
rs762125168 Chr.14: 99510798-99510799 on Build GRCh38
rs4905841 Chr.14: 99511059-99511059 on Build GRCh38
rs534472378 Chr.14: 99511149-99511149 on Build GRCh38
rs977479499 Chr.14: 99511240-99511240 on Build GRCh38
rs1022258502 Chr.14: 99511220-99511220 on Build GRCh38
rs1047921195 Chr.14: 99511117-99511117 on Build GRCh38
rs975369404 Chr.14: 99510928-99510928 on Build GRCh38
rs993199871 Chr.14: 99511075-99511075 on Build GRCh38
rs917116288 Chr.14: 99511191-99511191 on Build GRCh38
rs1033155637 Chr.14: 99510957-99510957 on Build GRCh38
rs910314656 Chr.14: 99511069-99511069 on Build GRCh38
rs182338033 Chr.14: 99511150-99511150 on Build GRCh38
rs750853717 Chr.14: 99510829-99510829 on Build GRCh38
rs201589535 Chr.14: 99511074-99511074 on Build GRCh38
rs779768159 Chr.14: 99510831-99510831 on Build GRCh38
rs745707450 Chr.14: 99511296-99511296 on Build GRCh38
rs3918127 Chr.14: 99511010-99511010 on Build GRCh38
rs548853061 Chr.14: 99511056-99511056 on Build GRCh38
rs758744749 Chr.14: 99510878-99510878 on Build GRCh38
rs190948481 Chr.14: 99510979-99510979 on Build GRCh38
rs964642036 Chr.14: 99510916-99510916 on Build GRCh38
rs3918126 Chr.14: 99510934-99510934 on Build GRCh38
rs924664209 Chr.14: 99511255-99511255 on Build GRCh38
rs1047636450 Chr.14: 99511161-99511161 on Build GRCh38
rs559873312 Chr.14: 99510799-99510799 on Build GRCh38
rs977904783 Chr.14: 99510970-99510970 on Build GRCh38
rs769954154 Chr.14: 99510802-99510803 on Build GRCh38
rs960172351 Chr.14: 99511266-99511266 on Build GRCh38
rs565861187 Chr.14: 99511097-99511097 on Build GRCh38
rs549716119 Chr.14: 99510999-99510999 on Build GRCh38
rs757165035 Chr.14: 99510807-99510807 on Build GRCh38
rs549815043 Chr.14: 99510883-99510883 on Build GRCh38
rs993662247 Chr.14: 99510851-99510851 on Build GRCh38
rs938837496 Chr.14: 99511006-99511006 on Build GRCh38
rs188264130 Chr.14: 99510901-99510901 on Build GRCh38
rs778926708 Chr.14: 99510800-99510800 on Build GRCh38
rs753953415 Chr.14: 99510784-99510784 on Build GRCh38
rs1017504356 Chr.14: 99510910-99510910 on Build GRCh38
rs3918125 Chr.14: 99510881-99510881 on Build GRCh38
rs571182098 Chr.14: 99511303-99511303 on Build GRCh38
rs1020668039 Chr.14: 99510997-99510997 on Build GRCh38
rs768534672 Chr.14: 99511208-99511208 on Build GRCh38
rs964586621 Chr.14: 99511174-99511174 on Build GRCh38
rs751539159 Chr.14: 99510803-99510803 on Build GRCh38
rs1017448985 Chr.14: 99511169-99511174 on Build GRCh38
rs776794029 Chr.14: 99510791-99510791 on Build GRCh38
rs539655114 Chr.14: 99510882-99510882 on Build GRCh38
rs867332738 Chr.14: 99511104-99511104 on Build GRCh38
rs3918128 Chr.14: 99511025-99511025 on Build GRCh38
rs574365914 Chr.14: 99511092-99511092 on Build GRCh38
rs1044490070 Chr.14: 99510897-99510897 on Build GRCh38
rs769407837 Chr.14: 99510830-99510830 on Build GRCh38
rs991454579 Chr.14: 99511070-99511070 on Build GRCh38
rs950089872 Chr.14: 99511192-99511192 on Build GRCh38
rs565536102 Chr.14: 99510955-99510955 on Build GRCh38

View Full Product Details