Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5745779-5746354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERD
Entrez Gene ID
257062
Gene Name
cation channel sperm associated auxiliary subunit delta
Gene Aliases
TMEM146
UniGene
Hs.631842
Gene Chromosome Location
Chr.19: 5720385-5778734 on Build GRCh38
Transcripts
NM_152784.3 XM_011527885.2 XM_011527886.2 XM_011527887.2 XM_011527888.2 XM_011527889.2 XM_011527890.2 XM_017026564.1 XM_017026565.1 XM_017026566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
179257 c.699C>T p.F233F Chr.19: 5745954-5745954 on Build GRCh38
6152684 c.692G>T p.R231L Chr.19: 5745947-5745947 on Build GRCh38
5909687 c.709T>A p.F237I Chr.19: 5745964-5745964 on Build GRCh38
370484 c.713A>T p.D238V Chr.19: 5745968-5745968 on Build GRCh38
4081968 c.702G>T p.G234G Chr.19: 5745957-5745957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140624041 Chr.19: 5745821-5745821 on Build GRCh38
rs1016262840 Chr.19: 5745849-5745849 on Build GRCh38
rs373573573 Chr.19: 5746096-5746096 on Build GRCh38
rs991856122 Chr.19: 5746317-5746317 on Build GRCh38
rs866062069 Chr.19: 5746305-5746305 on Build GRCh38
rs777047011 Chr.19: 5745919-5745919 on Build GRCh38
rs757001566 Chr.19: 5746075-5746075 on Build GRCh38
rs1014305372 Chr.19: 5746283-5746283 on Build GRCh38
rs753615039 Chr.19: 5745966-5745966 on Build GRCh38
rs773094567 Chr.19: 5746043-5746045 on Build GRCh38
rs563807776 Chr.19: 5745808-5745808 on Build GRCh38
rs541381158 Chr.19: 5746321-5746321 on Build GRCh38
rs200681020 Chr.19: 5745968-5745968 on Build GRCh38
rs143692878 Chr.19: 5746120-5746120 on Build GRCh38
rs777630392 Chr.19: 5745960-5745960 on Build GRCh38
rs773518485 Chr.19: 5746016-5746016 on Build GRCh38
rs555078118 Chr.19: 5746170-5746170 on Build GRCh38
rs777593773 Chr.19: 5745870-5745870 on Build GRCh38
rs373521530 Chr.19: 5746033-5746033 on Build GRCh38
rs1034314598 Chr.19: 5746201-5746201 on Build GRCh38
rs768054369 Chr.19: 5746110-5746110 on Build GRCh38
rs769080778 Chr.19: 5745917-5745917 on Build GRCh38
rs960006418 Chr.19: 5746253-5746253 on Build GRCh38
rs74336104 Chr.19: 5745996-5745996 on Build GRCh38
rs963437359 Chr.19: 5745852-5745852 on Build GRCh38
rs1002156943 Chr.19: 5746195-5746195 on Build GRCh38
rs781695133 Chr.19: 5746112-5746112 on Build GRCh38
rs756054945 Chr.19: 5745862-5745862 on Build GRCh38
rs1007576618 Chr.19: 5745837-5745837 on Build GRCh38
rs771944238 Chr.19: 5745998-5745998 on Build GRCh38
rs535522234 Chr.19: 5746139-5746139 on Build GRCh38
rs370051045 Chr.19: 5745991-5745991 on Build GRCh38
rs371533016 Chr.19: 5745879-5745879 on Build GRCh38
rs904764129 Chr.19: 5746188-5746188 on Build GRCh38
rs757052375 Chr.19: 5745967-5745967 on Build GRCh38
rs368212653 Chr.19: 5745942-5745942 on Build GRCh38
rs1051489708 Chr.19: 5746119-5746119 on Build GRCh38
rs745546371 Chr.19: 5745978-5745978 on Build GRCh38
rs762325095 Chr.19: 5745935-5745935 on Build GRCh38
rs778660807 Chr.19: 5746076-5746076 on Build GRCh38
rs886911161 Chr.19: 5746214-5746214 on Build GRCh38
rs527602493 Chr.19: 5745881-5745881 on Build GRCh38
rs1025670647 Chr.19: 5746304-5746304 on Build GRCh38
rs759549516 Chr.19: 5746038-5746038 on Build GRCh38
rs969144670 Chr.19: 5745854-5745854 on Build GRCh38
rs199735628 Chr.19: 5745872-5745872 on Build GRCh38
rs765495581 Chr.19: 5745937-5745937 on Build GRCh38
rs571960153 Chr.19: 5746300-5746300 on Build GRCh38
rs771606317 Chr.19: 5745886-5745886 on Build GRCh38
rs983302574 Chr.19: 5746320-5746320 on Build GRCh38
rs766577195 Chr.19: 5746026-5746026 on Build GRCh38
rs547957312 Chr.19: 5746155-5746155 on Build GRCh38
rs8109786 Chr.19: 5745814-5745814 on Build GRCh38
rs570335771 Chr.19: 5746002-5746002 on Build GRCh38
rs1034428925 Chr.19: 5745873-5745873 on Build GRCh38
rs1010383756 Chr.19: 5746130-5746130 on Build GRCh38
rs745319465 Chr.19: 5745884-5745884 on Build GRCh38
rs756076644 Chr.19: 5745957-5745957 on Build GRCh38
rs762200295 Chr.19: 5746003-5746003 on Build GRCh38
rs751648177 Chr.19: 5746037-5746037 on Build GRCh38
rs983802960 Chr.19: 5745956-5745956 on Build GRCh38
rs1002725347 Chr.19: 5746291-5746291 on Build GRCh38
rs993522895 Chr.19: 5746168-5746168 on Build GRCh38
rs766461198 Chr.19: 5745954-5745954 on Build GRCh38
rs771757701 Chr.19: 5745983-5745983 on Build GRCh38
rs1043739902 Chr.19: 5746160-5746160 on Build GRCh38
rs775815872 Chr.19: 5745901-5745901 on Build GRCh38
rs761218348 Chr.19: 5745914-5745914 on Build GRCh38
rs200180226 Chr.19: 5746013-5746013 on Build GRCh38
rs750255698 Chr.19: 5746093-5746093 on Build GRCh38
rs781163904 Chr.19: 5746279-5746279 on Build GRCh38
rs766377761 Chr.19: 5746109-5746110 on Build GRCh38
rs77656808 Chr.19: 5746110-5746111 on Build GRCh38
rs865994557 Chr.19: 5745897-5745897 on Build GRCh38
rs377544635 Chr.19: 5746045-5746045 on Build GRCh38
rs760768181 Chr.19: 5746052-5746052 on Build GRCh38
rs569694155 Chr.19: 5746131-5746131 on Build GRCh38
rs763086834 Chr.19: 5745946-5745946 on Build GRCh38
rs753806882 Chr.19: 5746062-5746062 on Build GRCh38
rs369406616 Chr.19: 5746094-5746094 on Build GRCh38
rs960121712 Chr.19: 5745924-5745924 on Build GRCh38
rs367882733 Chr.19: 5746148-5746148 on Build GRCh38
rs201497391 Chr.19: 5745955-5745955 on Build GRCh38
rs746469165 Chr.19: 5746097-5746097 on Build GRCh38
rs374016093 Chr.19: 5745938-5745938 on Build GRCh38
rs189729809 Chr.19: 5745833-5745833 on Build GRCh38
rs368808350 Chr.19: 5746023-5746023 on Build GRCh38

View Full Product Details