Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 76503471-76504033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT1
Entrez Gene ID
2650
Gene Name
glucosaminyl (N-acetyl) transferase 1, core 2
Gene Aliases
C2GNT, C2GNT-L, C2GNT1, G6NT, NACGT2, NAGCT2
UniGene
Hs.521568
Gene Chromosome Location
Chr.9: 76441666-76507416 on Build GRCh38
Transcripts
NM_001097633.1 NM_001097634.1 NM_001097635.1 NM_001097636.1 NM_001490.4 XM_017014620.1 XM_017014621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
373378 c.1140G>T p.V380V Chr.9: 76503521-76503521 on Build GRCh38
4521128 c.1096G>A p.G366S Chr.9: 76503477-76503477 on Build GRCh38
3716350 c.1178G>A p.R393H Chr.9: 76503559-76503559 on Build GRCh38
5698055 c.1219G>T p.D407Y Chr.9: 76503600-76503600 on Build GRCh38
1636223 c.1144A>G p.I382V Chr.9: 76503525-76503525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760020490 Chr.9: 76503496-76503496 on Build GRCh38
rs557684688 Chr.9: 76503914-76503914 on Build GRCh38
rs375561651 Chr.9: 76503580-76503580 on Build GRCh38
rs747452441 Chr.9: 76503702-76503702 on Build GRCh38
rs201887404 Chr.9: 76503559-76503559 on Build GRCh38
rs899969039 Chr.9: 76503938-76503938 on Build GRCh38
rs978893321 Chr.9: 76503520-76503520 on Build GRCh38
rs111932037 Chr.9: 76503675-76503675 on Build GRCh38
rs760645168 Chr.9: 76503499-76503499 on Build GRCh38
rs756435252 Chr.9: 76503582-76503582 on Build GRCh38
rs913304435 Chr.9: 76503821-76503821 on Build GRCh38
rs750711164 Chr.9: 76503630-76503630 on Build GRCh38
rs752530181 Chr.9: 76503655-76503655 on Build GRCh38
rs947396688 Chr.9: 76503840-76503840 on Build GRCh38
rs765367343 Chr.9: 76503626-76503626 on Build GRCh38
rs142834264 Chr.9: 76503943-76503943 on Build GRCh38
rs928790227 Chr.9: 76503785-76503785 on Build GRCh38
rs548515147 Chr.9: 76503717-76503717 on Build GRCh38
rs767377136 Chr.9: 76503651-76503651 on Build GRCh38
rs199518053 Chr.9: 76503534-76503534 on Build GRCh38
rs761199284 Chr.9: 76503591-76503591 on Build GRCh38
rs774591039 Chr.9: 76503494-76503494 on Build GRCh38
rs904086580 Chr.9: 76503978-76503978 on Build GRCh38
rs771375099 Chr.9: 76503491-76503491 on Build GRCh38
rs991187335 Chr.9: 76503780-76503780 on Build GRCh38
rs751311121 Chr.9: 76503528-76503528 on Build GRCh38
rs759806412 Chr.9: 76503568-76503568 on Build GRCh38
rs778403708 Chr.9: 76503676-76503676 on Build GRCh38
rs369252814 Chr.9: 76503678-76503678 on Build GRCh38
rs772694784 Chr.9: 76503964-76503964 on Build GRCh38
rs191384358 Chr.9: 76503962-76503962 on Build GRCh38
rs775826223 Chr.9: 76503590-76503590 on Build GRCh38
rs756136033 Chr.9: 76503659-76503659 on Build GRCh38
rs560994326 Chr.9: 76503955-76503955 on Build GRCh38
rs79645784 Chr.9: 76503681-76503681 on Build GRCh38
rs921879624 Chr.9: 76503894-76503894 on Build GRCh38
rs754698237 Chr.9: 76503649-76503649 on Build GRCh38
rs941221969 Chr.9: 76503898-76503898 on Build GRCh38
rs970334784 Chr.9: 76503776-76503776 on Build GRCh38
rs775942464 Chr.9: 76503698-76503698 on Build GRCh38
rs532810254 Chr.9: 76503612-76503612 on Build GRCh38
rs983082949 Chr.9: 76503896-76503896 on Build GRCh38
rs1004593957 Chr.9: 76503704-76503704 on Build GRCh38
rs182537966 Chr.9: 76503531-76503531 on Build GRCh38
rs200177188 Chr.9: 76503497-76503497 on Build GRCh38
rs988904959 Chr.9: 76503807-76503807 on Build GRCh38
rs199537430 Chr.9: 76503718-76503718 on Build GRCh38
rs996828793 Chr.9: 76503691-76503691 on Build GRCh38
rs898901554 Chr.9: 76504004-76504004 on Build GRCh38
rs748889157 Chr.9: 76503552-76503552 on Build GRCh38
rs370424129 Chr.9: 76503669-76503669 on Build GRCh38
rs764196313 Chr.9: 76503509-76503509 on Build GRCh38
rs769389794 Chr.9: 76503546-76503546 on Build GRCh38
rs754016515 Chr.9: 76503513-76503513 on Build GRCh38
rs3216171 Chr.9: 76503715-76503715 on Build GRCh38
rs369853279 Chr.9: 76503625-76503625 on Build GRCh38
rs1029618191 Chr.9: 76503700-76503700 on Build GRCh38
rs537323044 Chr.9: 76503769-76503769 on Build GRCh38
rs762389698 Chr.9: 76503965-76503965 on Build GRCh38
rs978780990 Chr.9: 76503865-76503865 on Build GRCh38
rs1057419 Chr.9: 76503745-76503745 on Build GRCh38
rs761718662 Chr.9: 76503832-76503832 on Build GRCh38
rs112742079 Chr.9: 76503712-76503712 on Build GRCh38
rs1038151009 Chr.9: 76503911-76503912 on Build GRCh38
rs540768389 Chr.9: 76503748-76503748 on Build GRCh38
rs960171935 Chr.9: 76503799-76503799 on Build GRCh38
rs761951470 Chr.9: 76503614-76503614 on Build GRCh38
rs377364894 Chr.9: 76503665-76503665 on Build GRCh38
rs550883200 Chr.9: 76503542-76503542 on Build GRCh38
rs753301345 Chr.9: 76503498-76503498 on Build GRCh38
rs777449301 Chr.9: 76503549-76503549 on Build GRCh38
rs979611357 Chr.9: 76503757-76503757 on Build GRCh38
rs113274175 Chr.9: 76503666-76503666 on Build GRCh38
rs757487322 Chr.9: 76503514-76503514 on Build GRCh38
rs111772588 Chr.9: 76503716-76503716 on Build GRCh38
rs577302091 Chr.9: 76503922-76503922 on Build GRCh38
rs777184329 Chr.9: 76503713-76503713 on Build GRCh38
rs908385692 Chr.9: 76503897-76503897 on Build GRCh38
rs746789076 Chr.9: 76503602-76503602 on Build GRCh38
rs1032784730 Chr.9: 76503759-76503759 on Build GRCh38
rs748801212 Chr.9: 76503671-76503671 on Build GRCh38
rs980726755 Chr.9: 76503777-76503777 on Build GRCh38
rs707739 Chr.9: 76503987-76503987 on Build GRCh38
rs754864501 Chr.9: 76503530-76503530 on Build GRCh38
rs779760950 Chr.9: 76503525-76503525 on Build GRCh38
rs774119437 Chr.9: 76503563-76503563 on Build GRCh38

View Full Product Details