Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132986340-132986901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFI1B
Entrez Gene ID
8328
Gene Name
growth factor independent 1B transcriptional repressor
Gene Aliases
BDPLT17, ZNF163B
UniGene
Hs.553160
Gene Chromosome Location
Chr.9: 132945545-132991697 on Build GRCh38
Transcripts
NM_001135031.1 NM_004188.6 XM_006717297.3 XM_011519068.2 XM_011519069.2 XM_011519070.2 XM_017015175.1 XM_017015176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6306653 c.67G>T p.D23Y Chr.9: 132986745-132986745 on Build GRCh38
3721923 c.40A>T p.T14S Chr.9: 132986718-132986718 on Build GRCh38
3655170 c.16C>T p.L6L Chr.9: 132986694-132986694 on Build GRCh38
4915469 c.74C>T p.P25L Chr.9: 132986752-132986752 on Build GRCh38
3926356 c.21G>A p.V7V Chr.9: 132986699-132986699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032380898 Chr.9: 132986427-132986427 on Build GRCh38
rs186517213 Chr.9: 132986398-132986398 on Build GRCh38
rs191271399 Chr.9: 132986665-132986665 on Build GRCh38
rs1044158296 Chr.9: 132986817-132986817 on Build GRCh38
rs765450265 Chr.9: 132986659-132986659 on Build GRCh38
rs776539618 Chr.9: 132986819-132986819 on Build GRCh38
rs963751005 Chr.9: 132986452-132986452 on Build GRCh38
rs778609270 Chr.9: 132986769-132986769 on Build GRCh38
rs898666868 Chr.9: 132986369-132986369 on Build GRCh38
rs543217257 Chr.9: 132986462-132986462 on Build GRCh38
rs559381069 Chr.9: 132986799-132986799 on Build GRCh38
rs763200851 Chr.9: 132986750-132986750 on Build GRCh38
rs2073577 Chr.9: 132986611-132986611 on Build GRCh38
rs879427164 Chr.9: 132986835-132986835 on Build GRCh38
rs201177977 Chr.9: 132986720-132986720 on Build GRCh38
rs568826386 Chr.9: 132986776-132986776 on Build GRCh38
rs578113282 Chr.9: 132986508-132986508 on Build GRCh38
rs866447063 Chr.9: 132986361-132986361 on Build GRCh38
rs938644590 Chr.9: 132986562-132986562 on Build GRCh38
rs751910167 Chr.9: 132986815-132986815 on Build GRCh38
rs540959188 Chr.9: 132986555-132986555 on Build GRCh38
rs764276098 Chr.9: 132986752-132986752 on Build GRCh38
rs753257069 Chr.9: 132986733-132986733 on Build GRCh38
rs998246328 Chr.9: 132986857-132986857 on Build GRCh38
rs757520760 Chr.9: 132986779-132986779 on Build GRCh38
rs1021775815 Chr.9: 132986726-132986726 on Build GRCh38
rs1038430183 Chr.9: 132986605-132986605 on Build GRCh38
rs544599977 Chr.9: 132986745-132986745 on Build GRCh38
rs758840675 Chr.9: 132986738-132986738 on Build GRCh38
rs759599251 Chr.9: 132986821-132986821 on Build GRCh38
rs954224732 Chr.9: 132986876-132986876 on Build GRCh38
rs369143285 Chr.9: 132986828-132986828 on Build GRCh38
rs939657415 Chr.9: 132986574-132986574 on Build GRCh38
rs746101008 Chr.9: 132986842-132986842 on Build GRCh38
rs776935786 Chr.9: 132986742-132986742 on Build GRCh38
rs35482159 Chr.9: 132986619-132986620 on Build GRCh38
rs775594146 Chr.9: 132986798-132986798 on Build GRCh38
rs1017018963 Chr.9: 132986367-132986367 on Build GRCh38
rs770253292 Chr.9: 132986793-132986793 on Build GRCh38
rs753415765 Chr.9: 132986702-132986702 on Build GRCh38
rs369970044 Chr.9: 132986635-132986635 on Build GRCh38
rs80161893 Chr.9: 132986569-132986569 on Build GRCh38
rs775541235 Chr.9: 132986749-132986749 on Build GRCh38
rs759494377 Chr.9: 132986656-132986656 on Build GRCh38
rs932938546 Chr.9: 132986547-132986547 on Build GRCh38
rs775137837 Chr.9: 132986779-132986782 on Build GRCh38
rs1054392022 Chr.9: 132986826-132986826 on Build GRCh38
rs749314167 Chr.9: 132986802-132986802 on Build GRCh38
rs781606245 Chr.9: 132986787-132986787 on Build GRCh38
rs958082723 Chr.9: 132986672-132986672 on Build GRCh38
rs764369477 Chr.9: 132986773-132986773 on Build GRCh38
rs776326772 Chr.9: 132986661-132986661 on Build GRCh38
rs528249677 Chr.9: 132986705-132986705 on Build GRCh38
rs373216343 Chr.9: 132986803-132986803 on Build GRCh38
rs976848683 Chr.9: 132986493-132986493 on Build GRCh38
rs758146973 Chr.9: 132986777-132986777 on Build GRCh38
rs530096075 Chr.9: 132986636-132986636 on Build GRCh38
rs61469632 Chr.9: 132986603-132986603 on Build GRCh38
rs763459499 Chr.9: 132986657-132986658 on Build GRCh38
rs556644999 Chr.9: 132986748-132986748 on Build GRCh38
rs577414650 Chr.9: 132986871-132986871 on Build GRCh38
rs114955344 Chr.9: 132986778-132986778 on Build GRCh38
rs774367148 Chr.9: 132986815-132986815 on Build GRCh38
rs369193177 Chr.9: 132986723-132986723 on Build GRCh38
rs377598720 Chr.9: 132986816-132986816 on Build GRCh38
rs776401954 Chr.9: 132986613-132986613 on Build GRCh38
rs977307406 Chr.9: 132986499-132986499 on Build GRCh38
rs764445985 Chr.9: 132986756-132986756 on Build GRCh38
rs756122349 Chr.9: 132986696-132986696 on Build GRCh38
rs1009956222 Chr.9: 132986715-132986715 on Build GRCh38
rs771820776 Chr.9: 132986729-132986729 on Build GRCh38
rs373220311 Chr.9: 132986829-132986829 on Build GRCh38
rs766157676 Chr.9: 132986818-132986818 on Build GRCh38
rs767365447 Chr.9: 132986667-132986667 on Build GRCh38
rs955678633 Chr.9: 132986378-132986378 on Build GRCh38
rs938560000 Chr.9: 132986751-132986751 on Build GRCh38
rs945890964 Chr.9: 132986464-132986464 on Build GRCh38
rs1143162 Chr.9: 132986764-132986764 on Build GRCh38
rs73554556 Chr.9: 132986852-132986852 on Build GRCh38
rs898639352 Chr.9: 132986610-132986610 on Build GRCh38
rs895151281 Chr.9: 132986586-132986586 on Build GRCh38
rs374383143 Chr.9: 132986424-132986424 on Build GRCh38
rs143455917 Chr.9: 132986734-132986734 on Build GRCh38
rs118082075 Chr.9: 132986615-132986615 on Build GRCh38
rs140302929 Chr.9: 132986732-132986732 on Build GRCh38
rs958265858 Chr.9: 132986441-132986441 on Build GRCh38
rs905689538 Chr.9: 132986641-132986641 on Build GRCh38
rs555244883 Chr.9: 132986473-132986473 on Build GRCh38
rs989909903 Chr.9: 132986460-132986460 on Build GRCh38
rs575203726 Chr.9: 132986408-132986408 on Build GRCh38
rs528120970 Chr.9: 132986854-132986854 on Build GRCh38
rs543669543 Chr.9: 132986539-132986539 on Build GRCh38
rs994401263 Chr.9: 132986371-132986371 on Build GRCh38
rs78483609 Chr.9: 132986362-132986362 on Build GRCh38
rs765218940 Chr.9: 132986824-132986824 on Build GRCh38
rs1026333296 Chr.9: 132986373-132986373 on Build GRCh38
rs534093250 Chr.9: 132986359-132986359 on Build GRCh38
rs1010150401 Chr.9: 132986834-132986834 on Build GRCh38
rs554684484 Chr.9: 132986791-132986791 on Build GRCh38
rs965532881 Chr.9: 132986470-132986470 on Build GRCh38
rs760342693 Chr.9: 132986753-132986753 on Build GRCh38
rs556276839 Chr.9: 132986853-132986853 on Build GRCh38
rs201670238 Chr.9: 132986663-132986663 on Build GRCh38
rs139980706 Chr.9: 132986693-132986693 on Build GRCh38

View Full Product Details