Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130713728-130714288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABL1
Entrez Gene ID
25
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
Gene Aliases
ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl
UniGene
Hs.431048
Gene Chromosome Location
Chr.9: 130713881-130887675 on Build GRCh38
Transcripts
NM_007313.2

SNP Detail

SNP ID Chromosome Location
rs962936691 Chr.9: 130713841-130713841 on Build GRCh38
rs34616588 Chr.9: 130713922-130713922 on Build GRCh38
rs993856999 Chr.9: 130714093-130714093 on Build GRCh38
rs959603480 Chr.9: 130714131-130714131 on Build GRCh38
rs896821935 Chr.9: 130714079-130714079 on Build GRCh38
rs992140160 Chr.9: 130713901-130713901 on Build GRCh38
rs530296443 Chr.9: 130713852-130713852 on Build GRCh38
rs901724977 Chr.9: 130713966-130713966 on Build GRCh38
rs75496288 Chr.9: 130714047-130714047 on Build GRCh38
rs114913770 Chr.9: 130713854-130713854 on Build GRCh38
rs945877231 Chr.9: 130713943-130713943 on Build GRCh38
rs893223824 Chr.9: 130713929-130713929 on Build GRCh38
rs758180942 Chr.9: 130713780-130713780 on Build GRCh38
rs545773633 Chr.9: 130713753-130713753 on Build GRCh38
rs1048157198 Chr.9: 130714098-130714098 on Build GRCh38
rs34519934 Chr.9: 130714249-130714249 on Build GRCh38
rs918810214 Chr.9: 130713869-130713869 on Build GRCh38
rs1006363743 Chr.9: 130714115-130714115 on Build GRCh38
rs917911391 Chr.9: 130713905-130713905 on Build GRCh38
rs978078361 Chr.9: 130714207-130714207 on Build GRCh38
rs879681684 Chr.9: 130714173-130714173 on Build GRCh38
rs35434141 Chr.9: 130714192-130714192 on Build GRCh38
rs1034263498 Chr.9: 130714123-130714123 on Build GRCh38
rs181237438 Chr.9: 130713951-130713951 on Build GRCh38
rs888806844 Chr.9: 130714061-130714061 on Build GRCh38
rs565585208 Chr.9: 130713755-130713755 on Build GRCh38
rs1041000360 Chr.9: 130714051-130714051 on Build GRCh38
rs971620837 Chr.9: 130714124-130714124 on Build GRCh38
rs186471720 Chr.9: 130714010-130714010 on Build GRCh38
rs781533380 Chr.9: 130714120-130714120 on Build GRCh38
rs1050044120 Chr.9: 130714009-130714009 on Build GRCh38
rs189920541 Chr.9: 130714104-130714104 on Build GRCh38
rs576831925 Chr.9: 130714133-130714133 on Build GRCh38
rs527941915 Chr.9: 130713760-130713760 on Build GRCh38
rs993909170 Chr.9: 130713805-130713805 on Build GRCh38
rs561156476 Chr.9: 130714250-130714250 on Build GRCh38
rs572791005 Chr.9: 130714230-130714230 on Build GRCh38
rs751238904 Chr.9: 130714032-130714032 on Build GRCh38
rs534191356 Chr.9: 130713908-130713908 on Build GRCh38
rs545123469 Chr.9: 130713772-130713772 on Build GRCh38
rs550312485 Chr.9: 130714257-130714257 on Build GRCh38
rs927338653 Chr.9: 130713882-130713882 on Build GRCh38
rs933205020 Chr.9: 130714246-130714246 on Build GRCh38
rs1041543884 Chr.9: 130714217-130714217 on Build GRCh38
rs1026775135 Chr.9: 130713840-130713840 on Build GRCh38
rs984577245 Chr.9: 130713834-130713834 on Build GRCh38
rs561431448 Chr.9: 130713796-130713796 on Build GRCh38
rs936481503 Chr.9: 130713941-130713941 on Build GRCh38
rs544427943 Chr.9: 130714101-130714101 on Build GRCh38
rs556818986 Chr.9: 130714130-130714130 on Build GRCh38
rs1028659932 Chr.9: 130714038-130714038 on Build GRCh38
rs545483793 Chr.9: 130714167-130714167 on Build GRCh38
rs981685009 Chr.9: 130714125-130714125 on Build GRCh38
rs1014253895 Chr.9: 130714152-130714152 on Build GRCh38
rs537247579 Chr.9: 130714105-130714105 on Build GRCh38
rs1041111476 Chr.9: 130713763-130713763 on Build GRCh38
rs34058168 Chr.9: 130713870-130713870 on Build GRCh38
rs978130939 Chr.9: 130713923-130713923 on Build GRCh38
rs561540111 Chr.9: 130714238-130714238 on Build GRCh38
rs953099978 Chr.9: 130713833-130713833 on Build GRCh38
rs541778425 Chr.9: 130714235-130714235 on Build GRCh38
rs943959845 Chr.9: 130714043-130714043 on Build GRCh38
rs1015983612 Chr.9: 130714090-130714090 on Build GRCh38
rs538666682 Chr.9: 130713873-130713873 on Build GRCh38
rs746279642 Chr.9: 130714213-130714213 on Build GRCh38
rs925401405 Chr.9: 130713928-130713928 on Build GRCh38
rs985276256 Chr.9: 130714243-130714243 on Build GRCh38
rs757961608 Chr.9: 130713825-130713825 on Build GRCh38
rs1054499644 Chr.9: 130713925-130713925 on Build GRCh38
rs1005959436 Chr.9: 130714088-130714088 on Build GRCh38
rs575463515 Chr.9: 130714014-130714014 on Build GRCh38

View Full Product Details