Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18510741-18511325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL5
Entrez Gene ID
6792
Gene Name
cyclin dependent kinase like 5
Gene Aliases
CFAP247, EIEE2, ISSX, STK9
UniGene
Hs.659851
Gene Chromosome Location
Chr.X: 18425605-18653629 on Build GRCh38
Transcripts
NM_001037343.1 NM_001323289.1 NM_003159.2

SNP Detail

SNP ID Chromosome Location
rs368320902 Chr.X: 18510771-18510771 on Build GRCh38
rs769668864 Chr.X: 18510790-18510790 on Build GRCh38
rs184941366 Chr.X: 18511302-18511302 on Build GRCh38
rs925272454 Chr.X: 18511205-18511205 on Build GRCh38
rs1011601149 Chr.X: 18510938-18510938 on Build GRCh38
rs587783130 Chr.X: 18510828-18510828 on Build GRCh38
rs142471598 Chr.X: 18510919-18510919 on Build GRCh38
rs768085119 Chr.X: 18510928-18510928 on Build GRCh38
rs891192393 Chr.X: 18510845-18510845 on Build GRCh38
rs752513906 Chr.X: 18510791-18510791 on Build GRCh38
rs748694628 Chr.X: 18511184-18511184 on Build GRCh38
rs758609121 Chr.X: 18510975-18510975 on Build GRCh38
rs760186283 Chr.X: 18510927-18510927 on Build GRCh38
rs751531623 Chr.X: 18511090-18511090 on Build GRCh38
rs587783131 Chr.X: 18510859-18510859 on Build GRCh38
rs1025254528 Chr.X: 18511143-18511143 on Build GRCh38
rs188701247 Chr.X: 18511024-18511024 on Build GRCh38
rs192846065 Chr.X: 18511167-18511167 on Build GRCh38
rs371743051 Chr.X: 18510782-18510782 on Build GRCh38
rs786204991 Chr.X: 18510846-18510846 on Build GRCh38
rs768612597 Chr.X: 18510900-18510900 on Build GRCh38
rs770248960 Chr.X: 18511256-18511256 on Build GRCh38
rs777490768 Chr.X: 18510816-18510816 on Build GRCh38
rs532222548 Chr.X: 18510807-18510807 on Build GRCh38
rs756095341 Chr.X: 18510792-18510792 on Build GRCh38
rs267608422 Chr.X: 18510883-18510883 on Build GRCh38
rs906141160 Chr.X: 18511118-18511118 on Build GRCh38
rs988595567 Chr.X: 18511273-18511273 on Build GRCh38
rs267608420 Chr.X: 18510820-18510821 on Build GRCh38
rs898955201 Chr.X: 18511001-18511001 on Build GRCh38
rs140332992 Chr.X: 18510848-18510848 on Build GRCh38
rs767125302 Chr.X: 18510787-18510787 on Build GRCh38
rs776283418 Chr.X: 18510968-18510968 on Build GRCh38
rs369016064 Chr.X: 18510904-18510904 on Build GRCh38
rs757081261 Chr.X: 18510877-18510877 on Build GRCh38
rs958149516 Chr.X: 18511206-18511206 on Build GRCh38
rs776029783 Chr.X: 18510803-18510803 on Build GRCh38
rs754436143 Chr.X: 18510786-18510786 on Build GRCh38
rs993614979 Chr.X: 18511035-18511035 on Build GRCh38
rs267608421 Chr.X: 18510855-18510855 on Build GRCh38
rs1001682844 Chr.X: 18511135-18511135 on Build GRCh38
rs755096347 Chr.X: 18511121-18511121 on Build GRCh38
rs1025820299 Chr.X: 18511040-18511040 on Build GRCh38
rs137874941 Chr.X: 18510888-18510888 on Build GRCh38
rs750625299 Chr.X: 18510936-18510936 on Build GRCh38
rs951497747 Chr.X: 18511068-18511068 on Build GRCh38
rs907569051 Chr.X: 18510774-18510774 on Build GRCh38
rs980829432 Chr.X: 18511187-18511187 on Build GRCh38

View Full Product Details