Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80442809-80443404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46D
Entrez Gene ID
169966
Gene Name
family with sequence similarity 46 member D
Gene Aliases
CT1.26, CT112
UniGene
Hs.367959
Gene Chromosome Location
Chr.X: 80335504-80445311 on Build GRCh38
Transcripts
NM_001170574.1 NM_152630.4

SNP Detail

SNP ID Chromosome Location
rs61995724 Chr.X: 80443166-80443166 on Build GRCh38
rs762766220 Chr.X: 80442962-80442962 on Build GRCh38
rs752494195 Chr.X: 80442863-80442863 on Build GRCh38
rs1008391005 Chr.X: 80442996-80442996 on Build GRCh38
rs781393813 Chr.X: 80442882-80442882 on Build GRCh38
rs769022513 Chr.X: 80442907-80442907 on Build GRCh38
rs767337141 Chr.X: 80443373-80443373 on Build GRCh38
rs775490438 Chr.X: 80443141-80443141 on Build GRCh38
rs148082423 Chr.X: 80443079-80443079 on Build GRCh38
rs757605310 Chr.X: 80443049-80443049 on Build GRCh38
rs750478659 Chr.X: 80443074-80443074 on Build GRCh38
rs908650103 Chr.X: 80443368-80443368 on Build GRCh38
rs189602496 Chr.X: 80442879-80442879 on Build GRCh38
rs763963851 Chr.X: 80443159-80443159 on Build GRCh38
rs112952835 Chr.X: 80443174-80443174 on Build GRCh38
rs186689398 Chr.X: 80442862-80442862 on Build GRCh38
rs781105250 Chr.X: 80443070-80443070 on Build GRCh38
rs898769200 Chr.X: 80442908-80442908 on Build GRCh38
rs374933664 Chr.X: 80443253-80443253 on Build GRCh38
rs201756726 Chr.X: 80443220-80443220 on Build GRCh38
rs188396741 Chr.X: 80442930-80442930 on Build GRCh38
rs768827836 Chr.X: 80442865-80442865 on Build GRCh38
rs753705045 Chr.X: 80442846-80442846 on Build GRCh38
rs765171207 Chr.X: 80442956-80442956 on Build GRCh38
rs368878211 Chr.X: 80442983-80442983 on Build GRCh38
rs780459324 Chr.X: 80442895-80442895 on Build GRCh38
rs779051485 Chr.X: 80442913-80442913 on Build GRCh38
rs138588734 Chr.X: 80442878-80442878 on Build GRCh38
rs867317346 Chr.X: 80443344-80443344 on Build GRCh38
rs764908562 Chr.X: 80442857-80442857 on Build GRCh38
rs149316031 Chr.X: 80442893-80442893 on Build GRCh38
rs763046111 Chr.X: 80443147-80443147 on Build GRCh38
rs754990371 Chr.X: 80442852-80442852 on Build GRCh38
rs140859426 Chr.X: 80443116-80443116 on Build GRCh38
rs751822975 Chr.X: 80443016-80443016 on Build GRCh38
rs150129047 Chr.X: 80442870-80442870 on Build GRCh38
rs139370296 Chr.X: 80443158-80443158 on Build GRCh38
rs776244562 Chr.X: 80442932-80442932 on Build GRCh38
rs113640425 Chr.X: 80442850-80442850 on Build GRCh38
rs754320744 Chr.X: 80443269-80443269 on Build GRCh38
rs758997189 Chr.X: 80442948-80442948 on Build GRCh38
rs755519479 Chr.X: 80443306-80443306 on Build GRCh38
rs776167652 Chr.X: 80443130-80443130 on Build GRCh38
rs752587473 Chr.X: 80442961-80442961 on Build GRCh38
rs780554117 Chr.X: 80443083-80443083 on Build GRCh38
rs779425399 Chr.X: 80443356-80443356 on Build GRCh38
rs372634428 Chr.X: 80443214-80443214 on Build GRCh38
rs143610675 Chr.X: 80442929-80442929 on Build GRCh38
rs61755189 Chr.X: 80443135-80443135 on Build GRCh38
rs746631425 Chr.X: 80443099-80443099 on Build GRCh38
rs182581837 Chr.X: 80442887-80442887 on Build GRCh38
rs750626708 Chr.X: 80443216-80443216 on Build GRCh38
rs201318635 Chr.X: 80442955-80442955 on Build GRCh38
rs959936581 Chr.X: 80443146-80443146 on Build GRCh38
rs749617118 Chr.X: 80442906-80442906 on Build GRCh38
rs749800137 Chr.X: 80443086-80443086 on Build GRCh38
rs61755190 Chr.X: 80443136-80443136 on Build GRCh38
rs143862091 Chr.X: 80443093-80443093 on Build GRCh38
rs778079784 Chr.X: 80442873-80442873 on Build GRCh38
rs374685215 Chr.X: 80442923-80442923 on Build GRCh38
rs1113265 Chr.X: 80443094-80443094 on Build GRCh38
rs774402950 Chr.X: 80443199-80443199 on Build GRCh38

View Full Product Details