Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 79406178-79406774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GK2
Entrez Gene ID
2712
Gene Name
glycerol kinase 2
Gene Aliases
GKP2, GKTA
UniGene
Hs.98008
Gene Chromosome Location
Chr.4: 79406353-79408218 on Build GRCh38
Transcripts
NM_033214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996466 c.1583C>T p.S528F Chr.4: 79406618-79406618 on Build GRCh38
5866825 c.1453G>A p.E485K Chr.4: 79406748-79406748 on Build GRCh38
1486114 c.1640G>C p.R547T Chr.4: 79406561-79406561 on Build GRCh38
4941873 c.1441G>A p.V481I Chr.4: 79406760-79406760 on Build GRCh38
4394560 c.1439C>T p.S480L Chr.4: 79406762-79406762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200207795 Chr.4: 79406686-79406686 on Build GRCh38
rs768973049 Chr.4: 79406739-79406739 on Build GRCh38
rs183594819 Chr.4: 79406457-79406457 on Build GRCh38
rs374538936 Chr.4: 79406277-79406277 on Build GRCh38
rs752104063 Chr.4: 79406569-79406569 on Build GRCh38
rs150707757 Chr.4: 79406245-79406245 on Build GRCh38
rs967797679 Chr.4: 79406330-79406330 on Build GRCh38
rs371254913 Chr.4: 79406705-79406705 on Build GRCh38
rs1008375938 Chr.4: 79406286-79406286 on Build GRCh38
rs774493694 Chr.4: 79406499-79406499 on Build GRCh38
rs939902839 Chr.4: 79406479-79406479 on Build GRCh38
rs753659323 Chr.4: 79406608-79406608 on Build GRCh38
rs931149193 Chr.4: 79406230-79406230 on Build GRCh38
rs974054799 Chr.4: 79406483-79406483 on Build GRCh38
rs34557444 Chr.4: 79406632-79406632 on Build GRCh38
rs956919128 Chr.4: 79406418-79406418 on Build GRCh38
rs147312231 Chr.4: 79406542-79406542 on Build GRCh38
rs761863743 Chr.4: 79406583-79406583 on Build GRCh38
rs960852815 Chr.4: 79406425-79406425 on Build GRCh38
rs763349038 Chr.4: 79406648-79406648 on Build GRCh38
rs11747 Chr.4: 79406472-79406472 on Build GRCh38
rs997577549 Chr.4: 79406329-79406330 on Build GRCh38
rs901112580 Chr.4: 79406557-79406557 on Build GRCh38
rs867193773 Chr.4: 79406634-79406634 on Build GRCh38
rs376757153 Chr.4: 79406507-79406507 on Build GRCh38
rs746526407 Chr.4: 79406725-79406725 on Build GRCh38
rs774794718 Chr.4: 79406662-79406662 on Build GRCh38
rs377582691 Chr.4: 79406604-79406604 on Build GRCh38
rs189734448 Chr.4: 79406495-79406495 on Build GRCh38
rs764583001 Chr.4: 79406565-79406565 on Build GRCh38
rs779899163 Chr.4: 79406490-79406490 on Build GRCh38
rs1036327211 Chr.4: 79406314-79406314 on Build GRCh38
rs778969212 Chr.4: 79406616-79406616 on Build GRCh38
rs773629437 Chr.4: 79406751-79406751 on Build GRCh38
rs771757204 Chr.4: 79406533-79406533 on Build GRCh38
rs137892990 Chr.4: 79406398-79406398 on Build GRCh38
rs766589876 Chr.4: 79406683-79406683 on Build GRCh38
rs765454049 Chr.4: 79406545-79406545 on Build GRCh38
rs558110551 Chr.4: 79406207-79406207 on Build GRCh38
rs144748131 Chr.4: 79406558-79406558 on Build GRCh38
rs561494537 Chr.4: 79406332-79406332 on Build GRCh38
rs373182459 Chr.4: 79406680-79406680 on Build GRCh38
rs776198316 Chr.4: 79406489-79406489 on Build GRCh38
rs267600272 Chr.4: 79406706-79406706 on Build GRCh38
rs377464934 Chr.4: 79406682-79406682 on Build GRCh38
rs1024575376 Chr.4: 79406446-79406446 on Build GRCh38
rs948226334 Chr.4: 79406292-79406292 on Build GRCh38
rs541616938 Chr.4: 79406319-79406319 on Build GRCh38
rs1021928456 Chr.4: 79406324-79406324 on Build GRCh38
rs202051208 Chr.4: 79406679-79406679 on Build GRCh38
rs769060950 Chr.4: 79406650-79406650 on Build GRCh38
rs144143489 Chr.4: 79406538-79406538 on Build GRCh38
rs778881211 Chr.4: 79406697-79406697 on Build GRCh38
rs980549114 Chr.4: 79406343-79406343 on Build GRCh38
rs770605965 Chr.4: 79406643-79406643 on Build GRCh38
rs527360665 Chr.4: 79406336-79406336 on Build GRCh38
rs919963814 Chr.4: 79406485-79406485 on Build GRCh38
rs575841938 Chr.4: 79406282-79406282 on Build GRCh38
rs774708528 Chr.4: 79406744-79406744 on Build GRCh38
rs1032366865 Chr.4: 79406296-79406296 on Build GRCh38
rs1053265927 Chr.4: 79406549-79406549 on Build GRCh38
rs140901841 Chr.4: 79406551-79406551 on Build GRCh38
rs755110124 Chr.4: 79406654-79406654 on Build GRCh38
rs867592537 Chr.4: 79406321-79406321 on Build GRCh38
rs776215042 Chr.4: 79406646-79406646 on Build GRCh38
rs566067569 Chr.4: 79406729-79406729 on Build GRCh38
rs867557237 Chr.4: 79406692-79406692 on Build GRCh38
rs754769311 Chr.4: 79406611-79406611 on Build GRCh38
rs78602685 Chr.4: 79406209-79406209 on Build GRCh38
rs1052314 Chr.4: 79406512-79406512 on Build GRCh38
rs770390846 Chr.4: 79406727-79406727 on Build GRCh38
rs749118242 Chr.4: 79406494-79406494 on Build GRCh38
rs970350945 Chr.4: 79406453-79406454 on Build GRCh38
rs777631974 Chr.4: 79406723-79406723 on Build GRCh38
rs752070089 Chr.4: 79406595-79406596 on Build GRCh38
rs777427264 Chr.4: 79406639-79406639 on Build GRCh38
rs960464440 Chr.4: 79406670-79406670 on Build GRCh38
rs547114748 Chr.4: 79406430-79406430 on Build GRCh38
rs1009252675 Chr.4: 79406323-79406323 on Build GRCh38
rs536094689 Chr.4: 79406552-79406552 on Build GRCh38
rs762342531 Chr.4: 79406666-79406666 on Build GRCh38
rs370754449 Chr.4: 79406631-79406631 on Build GRCh38
rs768294332 Chr.4: 79406225-79406225 on Build GRCh38
rs754770256 Chr.4: 79406696-79406696 on Build GRCh38
rs781360321 Chr.4: 79406655-79406656 on Build GRCh38
rs141163113 Chr.4: 79406219-79406219 on Build GRCh38
rs757652615 Chr.4: 79406283-79406283 on Build GRCh38
rs201032274 Chr.4: 79406541-79406541 on Build GRCh38
rs748118317 Chr.4: 79406626-79406626 on Build GRCh38
rs1033431733 Chr.4: 79406377-79406377 on Build GRCh38
rs1001851314 Chr.4: 79406653-79406653 on Build GRCh38
rs1032444093 Chr.4: 79406627-79406627 on Build GRCh38
rs147908480 Chr.4: 79406732-79406732 on Build GRCh38
rs200091513 Chr.4: 79406745-79406745 on Build GRCh38
rs926383198 Chr.4: 79406486-79406486 on Build GRCh38
rs867695888 Chr.4: 79406603-79406603 on Build GRCh38
rs756369279 Chr.4: 79406601-79406601 on Build GRCh38
rs908473635 Chr.4: 79406471-79406471 on Build GRCh38
rs566419487 Chr.4: 79406330-79406333 on Build GRCh38
rs369618994 Chr.4: 79406595-79406595 on Build GRCh38
rs200205106 Chr.4: 79406602-79406602 on Build GRCh38
rs755819327 Chr.4: 79406488-79406488 on Build GRCh38

View Full Product Details