Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31267164-31267734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH6
Entrez Gene ID
1004
Gene Name
cadherin 6
Gene Aliases
CAD6, KCAD
UniGene
Hs.124776
Gene Chromosome Location
Chr.5: 31193655-31329146 on Build GRCh38
Transcripts
NM_004932.3 XM_011513921.2 XM_017008910.1 XM_017008911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7413155 c.215A>G p.Q72R Chr.5: 31267688-31267688 on Build GRCh38
5868386 c.125G>A p.G42E Chr.5: 31267598-31267598 on Build GRCh38
4413110 c.170G>T p.W57L Chr.5: 31267643-31267643 on Build GRCh38
5450797 c.117G>T p.E39D Chr.5: 31267590-31267590 on Build GRCh38
74096 c.125G>C p.G42A Chr.5: 31267598-31267598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752593179 Chr.5: 31267231-31267231 on Build GRCh38
rs747260776 Chr.5: 31267591-31267591 on Build GRCh38
rs757802150 Chr.5: 31267571-31267571 on Build GRCh38
rs772327985 Chr.5: 31267460-31267460 on Build GRCh38
rs562537452 Chr.5: 31267602-31267602 on Build GRCh38
rs1005658489 Chr.5: 31267264-31267264 on Build GRCh38
rs537863981 Chr.5: 31267480-31267480 on Build GRCh38
rs762903474 Chr.5: 31267623-31267623 on Build GRCh38
rs771132576 Chr.5: 31267455-31267455 on Build GRCh38
rs144712334 Chr.5: 31267327-31267327 on Build GRCh38
rs769896023 Chr.5: 31267611-31267611 on Build GRCh38
rs188745311 Chr.5: 31267574-31267574 on Build GRCh38
rs368346209 Chr.5: 31267689-31267689 on Build GRCh38
rs905031126 Chr.5: 31267210-31267210 on Build GRCh38
rs750058121 Chr.5: 31267435-31267435 on Build GRCh38
rs768577996 Chr.5: 31267592-31267592 on Build GRCh38
rs370500416 Chr.5: 31267681-31267681 on Build GRCh38
rs201767254 Chr.5: 31267590-31267590 on Build GRCh38
rs746040162 Chr.5: 31267459-31267459 on Build GRCh38
rs1040705742 Chr.5: 31267211-31267211 on Build GRCh38
rs968360638 Chr.5: 31267349-31267349 on Build GRCh38
rs1003282366 Chr.5: 31267244-31267244 on Build GRCh38
rs112368228 Chr.5: 31267461-31267461 on Build GRCh38
rs747152944 Chr.5: 31267710-31267710 on Build GRCh38
rs779462305 Chr.5: 31267682-31267682 on Build GRCh38
rs749715359 Chr.5: 31267568-31267568 on Build GRCh38
rs766205249 Chr.5: 31267503-31267503 on Build GRCh38
rs1009253447 Chr.5: 31267322-31267322 on Build GRCh38
rs115945895 Chr.5: 31267351-31267351 on Build GRCh38
rs759260938 Chr.5: 31267660-31267660 on Build GRCh38
rs752299718 Chr.5: 31267680-31267680 on Build GRCh38
rs373660848 Chr.5: 31267567-31267567 on Build GRCh38
rs368126816 Chr.5: 31267580-31267580 on Build GRCh38
rs761843796 Chr.5: 31267487-31267487 on Build GRCh38
rs1034741023 Chr.5: 31267253-31267253 on Build GRCh38
rs774726977 Chr.5: 31267458-31267458 on Build GRCh38
rs930542289 Chr.5: 31267188-31267188 on Build GRCh38
rs373902933 Chr.5: 31267688-31267688 on Build GRCh38
rs940068893 Chr.5: 31267478-31267478 on Build GRCh38
rs561951224 Chr.5: 31267486-31267486 on Build GRCh38
rs778197559 Chr.5: 31267444-31267444 on Build GRCh38
rs773139830 Chr.5: 31267622-31267622 on Build GRCh38
rs149686511 Chr.5: 31267586-31267586 on Build GRCh38
rs780427917 Chr.5: 31267705-31267705 on Build GRCh38
rs766885014 Chr.5: 31267672-31267672 on Build GRCh38
rs772276938 Chr.5: 31267589-31267589 on Build GRCh38
rs749811705 Chr.5: 31267451-31267451 on Build GRCh38
rs892176803 Chr.5: 31267283-31267283 on Build GRCh38
rs756643915 Chr.5: 31267440-31267440 on Build GRCh38
rs540475400 Chr.5: 31267640-31267640 on Build GRCh38
rs35360913 Chr.5: 31267656-31267656 on Build GRCh38
rs547674648 Chr.5: 31267255-31267255 on Build GRCh38
rs868575517 Chr.5: 31267383-31267383 on Build GRCh38
rs560643887 Chr.5: 31267488-31267488 on Build GRCh38
rs1028537154 Chr.5: 31267357-31267357 on Build GRCh38
rs758095623 Chr.5: 31267501-31267501 on Build GRCh38
rs185851196 Chr.5: 31267202-31267202 on Build GRCh38
rs754385058 Chr.5: 31267546-31267546 on Build GRCh38
rs1044898516 Chr.5: 31267199-31267199 on Build GRCh38
rs555538646 Chr.5: 31267437-31267437 on Build GRCh38
rs776797071 Chr.5: 31267600-31267600 on Build GRCh38
rs200308506 Chr.5: 31267528-31267528 on Build GRCh38
rs779691971 Chr.5: 31267438-31267438 on Build GRCh38
rs750295332 Chr.5: 31267500-31267500 on Build GRCh38
rs35047741 Chr.5: 31267325-31267325 on Build GRCh38
rs974130351 Chr.5: 31267481-31267481 on Build GRCh38

View Full Product Details