Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135115171-135115744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL3
Entrez Gene ID
340351
Gene Name
ATP/GTP binding protein like 3
Gene Aliases
CCP3
UniGene
Hs.648616
Gene Chromosome Location
Chr.7: 134985169-135135782 on Build GRCh38
Transcripts
NM_178563.3 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1 NM_001305630.1 XM_017012595.1

Gene Symbol
C7orf49
Entrez Gene ID
78996
Gene Name
chromosome 7 open reading frame 49
Gene Aliases
MRI
UniGene
Hs.643113
Gene Chromosome Location
Chr.7: 135092303-135170826 on Build GRCh38
Transcripts
NM_178563.3 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1 NM_001305630.1 XM_017012595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792053 c.2342+7C>G p.? Chr.7: 135115618-135115618 on Build GRCh38
7426629 c.2494C>T p.Q832* Chr.7: 135115598-135115598 on Build GRCh38
7003322 c.2118C>T p.H706H Chr.7: 135115387-135115387 on Build GRCh38
6328488 c.2308G>A p.E770K Chr.7: 135115412-135115412 on Build GRCh38
1085807 c.2205C>T p.S735S Chr.7: 135115474-135115474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371770007 Chr.7: 135115638-135115641 on Build GRCh38
rs934961611 Chr.7: 135115329-135115329 on Build GRCh38
rs757729721 Chr.7: 135115461-135115461 on Build GRCh38
rs779054463 Chr.7: 135115568-135115569 on Build GRCh38
rs189696122 Chr.7: 135115477-135115477 on Build GRCh38
rs1028377241 Chr.7: 135115409-135115409 on Build GRCh38
rs755165667 Chr.7: 135115347-135115347 on Build GRCh38
rs1046469020 Chr.7: 135115694-135115695 on Build GRCh38
rs779255050 Chr.7: 135115353-135115353 on Build GRCh38
rs751209446 Chr.7: 135115460-135115460 on Build GRCh38
rs763853516 Chr.7: 135115458-135115458 on Build GRCh38
rs775534165 Chr.7: 135115450-135115450 on Build GRCh38
rs964426294 Chr.7: 135115496-135115496 on Build GRCh38
rs533800152 Chr.7: 135115491-135115491 on Build GRCh38
rs948169143 Chr.7: 135115418-135115418 on Build GRCh38
rs893306877 Chr.7: 135115356-135115356 on Build GRCh38
rs916645177 Chr.7: 135115403-135115403 on Build GRCh38
rs748416839 Chr.7: 135115355-135115355 on Build GRCh38
rs182356886 Chr.7: 135115532-135115532 on Build GRCh38
rs1054549282 Chr.7: 135115334-135115334 on Build GRCh38
rs537794368 Chr.7: 135115416-135115416 on Build GRCh38
rs754445096 Chr.7: 135115631-135115631 on Build GRCh38
rs1049908497 Chr.7: 135115721-135115721 on Build GRCh38
rs990542986 Chr.7: 135115522-135115522 on Build GRCh38
rs906547227 Chr.7: 135115460-135115460 on Build GRCh38
rs565589064 Chr.7: 135115661-135115661 on Build GRCh38
rs978151145 Chr.7: 135115552-135115552 on Build GRCh38
rs527692577 Chr.7: 135115194-135115194 on Build GRCh38
rs750731396 Chr.7: 135115537-135115537 on Build GRCh38
rs117898624 Chr.7: 135115481-135115481 on Build GRCh38
rs974390346 Chr.7: 135115208-135115208 on Build GRCh38
rs114250710 Chr.7: 135115651-135115651 on Build GRCh38
rs113930111 Chr.7: 135115331-135115331 on Build GRCh38
rs937743171 Chr.7: 135115259-135115259 on Build GRCh38
rs572746285 Chr.7: 135115693-135115693 on Build GRCh38
rs576710817 Chr.7: 135115628-135115628 on Build GRCh38
rs959203204 Chr.7: 135115494-135115494 on Build GRCh38
rs776435914 Chr.7: 135115425-135115425 on Build GRCh38
rs995942823 Chr.7: 135115520-135115520 on Build GRCh38
rs963686153 Chr.7: 135115620-135115620 on Build GRCh38
rs746442579 Chr.7: 135115684-135115684 on Build GRCh38
rs773961903 Chr.7: 135115507-135115507 on Build GRCh38
rs1053455496 Chr.7: 135115472-135115472 on Build GRCh38
rs549192523 Chr.7: 135115269-135115269 on Build GRCh38
rs573708799 Chr.7: 135115531-135115531 on Build GRCh38
rs996852264 Chr.7: 135115384-135115384 on Build GRCh38
rs755073596 Chr.7: 135115646-135115646 on Build GRCh38
rs112352940 Chr.7: 135115264-135115264 on Build GRCh38
rs771354848 Chr.7: 135115195-135115195 on Build GRCh38
rs1042077500 Chr.7: 135115319-135115319 on Build GRCh38
rs903418050 Chr.7: 135115322-135115322 on Build GRCh38
rs867176596 Chr.7: 135115443-135115443 on Build GRCh38
rs891105449 Chr.7: 135115426-135115426 on Build GRCh38
rs868252390 Chr.7: 135115641-135115641 on Build GRCh38
rs771017532 Chr.7: 135115654-135115655 on Build GRCh38
rs568700781 Chr.7: 135115284-135115284 on Build GRCh38
rs893275863 Chr.7: 135115685-135115685 on Build GRCh38
rs932840286 Chr.7: 135115717-135115717 on Build GRCh38
rs28640016 Chr.7: 135115262-135115262 on Build GRCh38
rs78930373 Chr.7: 135115493-135115493 on Build GRCh38
rs949088810 Chr.7: 135115275-135115275 on Build GRCh38
rs117730477 Chr.7: 135115246-135115246 on Build GRCh38
rs556838766 Chr.7: 135115576-135115576 on Build GRCh38
rs372057243 Chr.7: 135115431-135115431 on Build GRCh38

View Full Product Details