Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26581809-26582408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL2
Entrez Gene ID
1808
Gene Name
dihydropyrimidinase like 2
Gene Aliases
CRMP-2, CRMP2, DHPRP2, DRP-2, DRP2, N2A3, ULIP-2, ULIP2
UniGene
Hs.593187
Gene Chromosome Location
Chr.8: 26514193-26658177 on Build GRCh38
Transcripts
NM_001197293.2 NM_001244604.1 NM_001386.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204525 c.70A>C p.I24L Chr.8: 26581999-26581999 on Build GRCh38
7083089 c.43G>T p.D15Y Chr.8: 26581972-26581972 on Build GRCh38
6271011 c.82G>T p.D28Y Chr.8: 26582011-26582011 on Build GRCh38
5649015 c.57C>T p.I19I Chr.8: 26581986-26581986 on Build GRCh38
7314319 c.64G>T p.G22C Chr.8: 26581993-26581993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011742197 Chr.8: 26582351-26582351 on Build GRCh38
rs908599798 Chr.8: 26581885-26581885 on Build GRCh38
rs915737254 Chr.8: 26582355-26582355 on Build GRCh38
rs760149612 Chr.8: 26581924-26581924 on Build GRCh38
rs750995710 Chr.8: 26581986-26581986 on Build GRCh38
rs780855720 Chr.8: 26582340-26582340 on Build GRCh38
rs376843343 Chr.8: 26582105-26582105 on Build GRCh38
rs145324050 Chr.8: 26582177-26582177 on Build GRCh38
rs772763159 Chr.8: 26582086-26582086 on Build GRCh38
rs750095391 Chr.8: 26581955-26581955 on Build GRCh38
rs765937076 Chr.8: 26581976-26581976 on Build GRCh38
rs192122791 Chr.8: 26582247-26582247 on Build GRCh38
rs1030459922 Chr.8: 26582181-26582181 on Build GRCh38
rs956418927 Chr.8: 26582128-26582128 on Build GRCh38
rs749543222 Chr.8: 26582070-26582070 on Build GRCh38
rs765049299 Chr.8: 26581954-26581954 on Build GRCh38
rs976187633 Chr.8: 26582162-26582162 on Build GRCh38
rs1049989995 Chr.8: 26582023-26582023 on Build GRCh38
rs552762302 Chr.8: 26581836-26581836 on Build GRCh38
rs753304614 Chr.8: 26582220-26582220 on Build GRCh38
rs1057003691 Chr.8: 26582027-26582027 on Build GRCh38
rs199994960 Chr.8: 26582019-26582019 on Build GRCh38
rs372134054 Chr.8: 26581918-26581918 on Build GRCh38
rs939180619 Chr.8: 26581846-26581846 on Build GRCh38
rs1003905538 Chr.8: 26582127-26582127 on Build GRCh38
rs549488422 Chr.8: 26582253-26582253 on Build GRCh38
rs968231729 Chr.8: 26582385-26582385 on Build GRCh38
rs891958602 Chr.8: 26582312-26582312 on Build GRCh38
rs138340807 Chr.8: 26581971-26581971 on Build GRCh38
rs116076155 Chr.8: 26582148-26582148 on Build GRCh38
rs932654056 Chr.8: 26582271-26582271 on Build GRCh38
rs1053183429 Chr.8: 26582311-26582311 on Build GRCh38
rs770338377 Chr.8: 26582276-26582276 on Build GRCh38
rs769078245 Chr.8: 26582100-26582100 on Build GRCh38
rs908382785 Chr.8: 26582231-26582231 on Build GRCh38
rs369399315 Chr.8: 26582077-26582077 on Build GRCh38
rs986055414 Chr.8: 26581880-26581880 on Build GRCh38
rs563340893 Chr.8: 26582211-26582211 on Build GRCh38
rs199968124 Chr.8: 26582165-26582165 on Build GRCh38
rs112345442 Chr.8: 26582059-26582059 on Build GRCh38
rs187676050 Chr.8: 26582159-26582159 on Build GRCh38
rs754950124 Chr.8: 26582008-26582008 on Build GRCh38
rs745622408 Chr.8: 26581956-26581957 on Build GRCh38
rs183303882 Chr.8: 26581917-26581917 on Build GRCh38
rs113765113 Chr.8: 26581845-26581845 on Build GRCh38
rs1057273914 Chr.8: 26582323-26582323 on Build GRCh38
rs767277545 Chr.8: 26581939-26581939 on Build GRCh38
rs34978863 Chr.8: 26582337-26582338 on Build GRCh38
rs776812636 Chr.8: 26582103-26582103 on Build GRCh38
rs1008897049 Chr.8: 26582302-26582302 on Build GRCh38
rs559932397 Chr.8: 26582313-26582313 on Build GRCh38
rs891352670 Chr.8: 26582279-26582279 on Build GRCh38
rs150442037 Chr.8: 26582090-26582090 on Build GRCh38
rs777369069 Chr.8: 26582064-26582064 on Build GRCh38
rs745918228 Chr.8: 26582079-26582079 on Build GRCh38
rs140494039 Chr.8: 26581938-26581938 on Build GRCh38
rs772673561 Chr.8: 26581933-26581933 on Build GRCh38
rs577876554 Chr.8: 26582174-26582174 on Build GRCh38
rs1021982673 Chr.8: 26582375-26582375 on Build GRCh38
rs755902500 Chr.8: 26582055-26582055 on Build GRCh38
rs977469495 Chr.8: 26582256-26582256 on Build GRCh38
rs947445413 Chr.8: 26581899-26581899 on Build GRCh38
rs576057697 Chr.8: 26582076-26582076 on Build GRCh38
rs903293192 Chr.8: 26581951-26581951 on Build GRCh38
rs969107022 Chr.8: 26582250-26582250 on Build GRCh38
rs938759255 Chr.8: 26582299-26582299 on Build GRCh38
rs754215253 Chr.8: 26581850-26581850 on Build GRCh38
rs914555624 Chr.8: 26581882-26581882 on Build GRCh38
rs926547762 Chr.8: 26582185-26582185 on Build GRCh38
rs1026935615 Chr.8: 26582098-26582098 on Build GRCh38
rs570959014 Chr.8: 26581847-26581847 on Build GRCh38
rs991969776 Chr.8: 26582217-26582217 on Build GRCh38
rs761106927 Chr.8: 26581940-26581940 on Build GRCh38

View Full Product Details