Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 53769645-53770113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1H
Entrez Gene ID
51606
Gene Name
ATPase H+ transporting V1 subunit H
Gene Aliases
CGI-11, MSTP042, NBP1, SFD, SFDalpha, SFDbeta, VMA13
UniGene
Hs.491737
Gene Chromosome Location
Chr.8: 53715543-53843311 on Build GRCh38
Transcripts
NM_015941.3 NM_213619.2 NM_213620.2 XM_006716455.2 XM_011517542.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5893280 c.1070C>T p.S357L Chr.8: 53769723-53769723 on Build GRCh38
3900633 c.1041A>G p.E347E Chr.8: 53769698-53769698 on Build GRCh38
1283582 c.1056C>G p.H352Q Chr.8: 53769683-53769683 on Build GRCh38
1283581 c.1110C>G p.H370Q Chr.8: 53769683-53769683 on Build GRCh38
1648087 c.1065C>G p.Y355* Chr.8: 53769728-53769728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745797084 Chr.8: 53769790-53769790 on Build GRCh38
rs961519773 Chr.8: 53769881-53769881 on Build GRCh38
rs552196320 Chr.8: 53769758-53769758 on Build GRCh38
rs368229604 Chr.8: 53770057-53770057 on Build GRCh38
rs370578376 Chr.8: 53769754-53769754 on Build GRCh38
rs772928546 Chr.8: 53769916-53769916 on Build GRCh38
rs961086640 Chr.8: 53770064-53770064 on Build GRCh38
rs895849204 Chr.8: 53770052-53770052 on Build GRCh38
rs984173189 Chr.8: 53770078-53770078 on Build GRCh38
rs1016722472 Chr.8: 53769876-53769876 on Build GRCh38
rs867048020 Chr.8: 53769686-53769686 on Build GRCh38
rs778612230 Chr.8: 53769789-53769789 on Build GRCh38
rs575224776 Chr.8: 53769718-53769718 on Build GRCh38
rs1025604436 Chr.8: 53770062-53770062 on Build GRCh38
rs952912561 Chr.8: 53770074-53770074 on Build GRCh38
rs532802862 Chr.8: 53770002-53770002 on Build GRCh38
rs911221623 Chr.8: 53769934-53769934 on Build GRCh38
rs182449666 Chr.8: 53769951-53769951 on Build GRCh38
rs74393216 Chr.8: 53769901-53769901 on Build GRCh38
rs551495516 Chr.8: 53770041-53770041 on Build GRCh38
rs189891120 Chr.8: 53769933-53769933 on Build GRCh38
rs199934417 Chr.8: 53769800-53769800 on Build GRCh38
rs922778156 Chr.8: 53769915-53769915 on Build GRCh38
rs142433792 Chr.8: 53769743-53769743 on Build GRCh38
rs57348174 Chr.8: 53769698-53769698 on Build GRCh38
rs3735831 Chr.8: 53769866-53769866 on Build GRCh38
rs893546138 Chr.8: 53769955-53769955 on Build GRCh38
rs757034915 Chr.8: 53769788-53769788 on Build GRCh38
rs567329029 Chr.8: 53769950-53769950 on Build GRCh38
rs765868393 Chr.8: 53769943-53769943 on Build GRCh38
rs550614137 Chr.8: 53769919-53769919 on Build GRCh38
rs1042171335 Chr.8: 53769962-53769962 on Build GRCh38
rs1003880197 Chr.8: 53769759-53769759 on Build GRCh38
rs922318305 Chr.8: 53769937-53769938 on Build GRCh38
rs111414889 Chr.8: 53769905-53769905 on Build GRCh38
rs1033313445 Chr.8: 53769851-53769851 on Build GRCh38
rs752068866 Chr.8: 53769676-53769676 on Build GRCh38
rs1005294910 Chr.8: 53769873-53769873 on Build GRCh38
rs113474015 Chr.8: 53769728-53769728 on Build GRCh38
rs1005654084 Chr.8: 53770031-53770031 on Build GRCh38
rs577657345 Chr.8: 53769939-53769939 on Build GRCh38
rs993925509 Chr.8: 53770085-53770085 on Build GRCh38
rs772058028 Chr.8: 53769795-53769795 on Build GRCh38
rs117884943 Chr.8: 53769971-53769971 on Build GRCh38
rs754887510 Chr.8: 53769965-53769965 on Build GRCh38
rs564202217 Chr.8: 53769771-53769772 on Build GRCh38
rs572223705 Chr.8: 53770030-53770030 on Build GRCh38
rs755483881 Chr.8: 53769678-53769678 on Build GRCh38
rs1009382139 Chr.8: 53769692-53769692 on Build GRCh38
rs530344276 Chr.8: 53769917-53769917 on Build GRCh38
rs748915070 Chr.8: 53769775-53769775 on Build GRCh38
rs555677398 Chr.8: 53769979-53769979 on Build GRCh38
rs946151081 Chr.8: 53769958-53769958 on Build GRCh38

View Full Product Details